Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001268.3 | NP_001259.1 | 13 | 1722 | |
NM_001286830.1 | NP_001273759.1 | 12 | 1657 | ||
NM_001286831.1 | NP_001273760.1 | 10 | 1462 | ||
NM_001286832.1 | NP_001273761.1 | 8 | 1100 | ||
XM_005266242.2 | XP_005266299.1 | 12 | 1575 | ||
XM_005266244.2 | XP_005266301.1 | 11 | 1510 | ||
XM_006719759.3 | XP_006719822.1 | 12 | 1543 | ||
XM_011534894.2 | XP_011533196.1 | 11 | 1470 | ||
XM_011534895.1 | XP_011533197.1 | 9 | 1191 | ||
XM_017020369.1 | XP_016875858.1 | 14 | 1996 | ||
XM_017020370.1 | XP_016875859.1 | 13 | 1868 | ||
XM_017020371.1 | XP_016875860.1 | 12 | 1741 | ||
XM_017020372.1 | XP_016875861.1 | 11 | 1351 | ||
XM_017020373.1 | XP_016875862.1 | 10 | 1533 | ||
XM_017020378.1 | XP_016875867.1 | 12 | 1528 | ||
XM_017020379.1 | XP_016875868.1 | 11 | 1463 | ||
GenBank mRNA | AB209543.1 | - | 10 | 1428 | |
AF060219.1 | - | 12 | 1503 | ||
AK023010.1 | - | 12 | 1465 | ||
AK122778.1 | - | 9 | 1377 | ||
AK301516.1 | - | 12 | 1481 | ||
AK304598.1 | - | 8 | 924 | ||
AK310081.1 | - | 11 | 1457 | ||
AK315703.1 | - | 13 | 1642 | ||
BC029052.1 | - | 13 | 1526 | 5194795 |