Gene Transcripts

Gene Symbol
SPATA17
Entrez Gene ID
128153
Gene Name
spermatogenesis associated 17
Gene Aliases
IQCH, MSRG-11, MSRG11
Chromosome Location
Chr. 1: 217631324 - 217868151 on Build GRCh38
UniGene ID
Hs.171130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138796.3 NP_620151.1 1, 2 128
XM_005273052.1 XP_005273109.1 1, 2 110
XM_006711165.3 XP_006711228.1 1, 2 119
XM_011509194.2 XP_011507496.1 1, 2 110
GenBank mRNA AK098591.1 - 1, 2 108
AK310291.1 - 1, 2 112
AY963797.1 - 1, 2 131
BC014608.2 - 1, 2 99 4824429

View Full Product Details