Gene Transcripts
- Gene Symbol
- FAM69B
- Entrez Gene ID
- 138311
- Gene Name
- family with sequence similarity 69 member B
- Gene Aliases
- C9orf136, pp6977
- Chromosome Location
- Chr. 9: 136712572 - 136724718 on Build GRCh38
- UniGene ID
- Hs.741305
- Species
- Homo sapiens
- Gene Symbol
- SNORA17B
- Entrez Gene ID
- 677824
- Gene Name
- small nucleolar RNA, H/ACA box 17B
- Gene Aliases
- ACA43, SNORA43
- Chromosome Location
- Chr. 9: 136726104 - 136726237 on Build GRCh38
- UniGene ID
- Hs.495480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054908.1 |
- |
6 |
1563 |
|
View Full Product Details