Gene Transcripts

Gene Symbol
TRMT44
Entrez Gene ID
152992
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases
C4orf23, METTL19, TRM44
Chromosome Location
Chr. 4: 8440774 - 8512531 on Build GRCh38
UniGene ID
Hs.566191
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152544.2 NP_689757.2 8 1433
XM_005247950.3 XP_005248007.1 8 1430
XM_011513406.2 XP_011511708.1 8 1462
XM_011513407.2 XP_011511709.1 8 1462
XM_011513408.1 XP_011511710.1 9 1574
XM_011513409.2 XP_011511711.1 9 1545
GenBank mRNA AK093044.1 - 4 439
BC035655.1 - 8 933 5588410

View Full Product Details