Gene Transcripts

Gene Symbol
TIGIT
Entrez Gene ID
201633
Gene Name
T-cell immunoreceptor with Ig and ITIM domains
Gene Aliases
VSIG9, VSTM3, WUCAM
Chromosome Location
Chr. 3: 114293986 - 114310288 on Build GRCh38
UniGene ID
Hs.421750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173799.3 NP_776160.2 1, 2 147
XM_011512538.1 XP_011510840.1 1, 2 135
GenBank mRNA AK097192.1 - 1, 2 105
AL833175.1 - 2, 3 2887
BC101288.1 - 1, 2 104 40024492
BC101289.1 - 1, 2 104 40024494
BC101290.1 - 1, 2 104 40024496
BC101291.1 - 1, 2 104 40024497
BX640915.1 - 11
EU675310.1 - 1, 2 147

View Full Product Details