Gene Transcripts

Gene Symbol
KDM6B
Entrez Gene ID
23135
Gene Name
lysine demethylase 6B
Gene Aliases
JMJD3
Chromosome Location
Chr. 17: 7834210 - 7854800 on Build GRCh38
UniGene ID
Hs.223678
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080424.1 NP_001073893.1 11 1708
XM_005256549.3 XP_005256606.1 12 1860
XM_005256550.4 XP_005256607.1 11 1702
XM_005256551.3 XP_005256608.1 11 1741
XM_005256552.4 XP_005256609.1 11 2714
XM_006721483.3 XP_006721546.1 12 1805
XM_011523750.2 XP_011522052.1 12 3065
XM_011523752.2 XP_011522054.1 11 1705
XM_017024381.1 XP_016879870.1 11 3151
GenBank mRNA AB002344.2 - 11 1713

View Full Product Details