Gene Transcripts

Gene Symbol
WFDC11
Entrez Gene ID
259239
Gene Name
WAP four-disulfide core domain 11
Gene Aliases
WAP11
Chromosome Location
Chr. 20: 45648563 - 45670239 on Build GRCh38
UniGene ID
Hs.374924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147197.2 NP_671730.1 3, 4 304
GenBank mRNA AY047609.1 - 3, 4 304
BC062670.1 - 3, 4 335 6101449

View Full Product Details