Gene Transcripts

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
Chromosome Location
Chr. 11: 18704305 - 18726939 on Build GRCh38
UniGene ID
Hs.434152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173588.3 NP_775859.3 17 2878
XM_011520030.2 XP_011518332.1 15 5782
XM_011520031.2 XP_011518333.1 14 5479
XM_017017607.1 XP_016873096.1 16 5673
XM_017017608.1 XP_016873097.1 16 3696
GenBank mRNA AK095113.1 - 16 2575
BC115396.1 - 17 2829
BC115397.1 - 16 3028 40068579

View Full Product Details