Gene Transcripts

Gene Symbol
TRIM42
Entrez Gene ID
287015
Gene Name
tripartite motif containing 42
Gene Aliases
PPP1R40, T4A1
Chromosome Location
Chr. 3: 140678024 - 140701150 on Build GRCh38
UniGene ID
Hs.343487
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152616.4 NP_689829.3 3 1276
XM_011512740.2 XP_011511042.1 3 1271
GenBank mRNA AF521868.1 - 3 1088
AK097416.1 - 3 1263
AK310536.1 - 3 1255
BC130472.1 - 3 1186 40146503
BC130474.1 - 3 1186 40146505

View Full Product Details