Gene Transcripts

Gene Symbol
PALM2-AKAP2
Entrez Gene ID
445815
Gene Name
PALM2-AKAP2 readthrough
Gene Aliases
AKAP2
Chromosome Location
Chr. 9: 109780297 - 110172512 on Build GRCh38
UniGene ID
Hs.591908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007203.4 NP_009134.1 7 734
NM_147150.2 NP_671492.1 7 734
GenBank mRNA AB023137.1 - 7 734
AJ303079.1 - 7 722
AJ312217.1 - 8 1284
BC140818.1 - 7 575 9021686
BX649126.1 - 5 520

View Full Product Details