Gene Transcripts

Gene Symbol
AKR1E2
Entrez Gene ID
83592
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR
Chromosome Location
Chr. 10: 4824554 - 4908421 on Build GRCh38
UniGene ID
Hs.657944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040177.2 NP_001035267.1 8 970
NM_001271021.1 NP_001257950.1 6 799
NM_001271025.1 NP_001257954.1 5 676
NR_073125.1 8 970
NR_073126.1 8 884
NR_073127.1 6 799
XM_011519715.2 XP_011518017.1 8 1267
XM_011519718.2 XP_011518020.1 8 1267
XM_011519720.2 XP_011518022.1 8 1249
XM_011519722.2 XP_011518024.1 8 1250
XM_011519724.2 XP_011518026.1 6 1095
XM_011519725.2 XP_011518027.1 7 1191
XM_017016743.1 XP_016872232.1 8 1268
XM_017016744.1 XP_016872233.1 8 1250
XM_017016745.1 XP_016872234.1 8 1053
GenBank mRNA AB040820.1 - 6 680
AB040821.1 - 6 680
AB040822.1 - 5 557
AB055603.1 - 8 822
AF263242.1 - 8 842
AK074211.1 - 8 884
AK302155.1 - 5 604
BC002862.2 - 8 913 3941289
BM724648.1 - 1 60

View Full Product Details