Gene Transcripts

Gene Symbol
FYTTD1
Entrez Gene ID
84248
Gene Name
forty-two-three domain containing 1
Gene Aliases
UIF
Chromosome Location
Chr. 3: 197749553 - 197784446 on Build GRCh38
UniGene ID
Hs.277533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011537.2 NP_001011537.2 9 954
NM_032288.6 NP_115664.2 8 1013
NR_027840.1 8 1124
GenBank mRNA AJ344094.1 - 8 859
AK296283.1 - 7 715
AK302546.1 - 8 1124
AK303462.1 - 9 954
AK315019.1 - 8 862
AK316308.1 - 8 754
AL136558.1 - 8 866
BC035006.2 - 8 1016 5267824
BC039734.1 - 8 923 6042926
CR533502.1 - 8 794

View Full Product Details