Gene Transcripts

Gene Symbol
CEACAM21
Entrez Gene ID
90273
Gene Name
carcinoembryonic antigen related cell adhesion molecule 21
Gene Aliases
CEACAM3, R29124_1
Chromosome Location
Chr. 19: 41549518 - 41586844 on Build GRCh38
UniGene ID
Hs.655885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098506.3 NP_001091976.3 2 411
NM_001288773.2 NP_001275702.2 3 661
NM_001290113.1 NP_001277042.1 2 411
NM_033543.5 NP_291021.4 2 411
XM_005278396.4 XP_005278453.1 2 613
XM_005278397.3 XP_005278454.1 2 611
XM_011527450.1 XP_011525752.1 3 699
XM_011527452.1 XP_011525754.1 3 661
XM_017027430.1 XP_016882919.1 2 985
XM_017027431.1 XP_016882920.1 2 985
XM_017027432.1 XP_016882921.1 2 983
XM_017027433.1 XP_016882922.1 3 661
XM_017027434.1 XP_016882923.1 2 626
XM_017027435.1 XP_016882924.1 2 626
XM_017027436.1 XP_016882925.1 6 1808
XM_017027437.1 XP_016882926.1 2 2841
GenBank mRNA AK023602.1 - 3 661
AY358084.1 - 2 355
BC012001.1 - 2 411 4540619
BC106727.1 - 2 341 40015070
BI833768.1 - 2 411
BI913326.1 - 3 652

View Full Product Details