Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112961167-112961663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928847
Entrez Gene ID
101928847
Gene Name
uncharacterized LOC101928847
Gene Aliases
-
UniGene
Hs.711235
Gene Chromosome Location
Chr.11: 112959279-112963460 on Build GRCh38
Transcripts
NR_120563.1 NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NR_120563.1 NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

SNP Detail

SNP ID Chromosome Location
rs782231095 Chr.11: 112961329-112961330 on Build GRCh38
rs782102520 Chr.11: 112961504-112961504 on Build GRCh38
rs782225336 Chr.11: 112961617-112961617 on Build GRCh38
rs782768191 Chr.11: 112961556-112961557 on Build GRCh38
rs6589347 Chr.11: 112961636-112961636 on Build GRCh38
rs1008564169 Chr.11: 112961304-112961304 on Build GRCh38
rs11381910 Chr.11: 112961585-112961586 on Build GRCh38
rs782063881 Chr.11: 112961342-112961342 on Build GRCh38
rs1048810733 Chr.11: 112961554-112961554 on Build GRCh38
rs7938749 Chr.11: 112961324-112961324 on Build GRCh38
rs782245249 Chr.11: 112961562-112961562 on Build GRCh38
rs782563501 Chr.11: 112961322-112961322 on Build GRCh38
rs913319762 Chr.11: 112961201-112961201 on Build GRCh38
rs782353809 Chr.11: 112961476-112961476 on Build GRCh38
rs782563224 Chr.11: 112961587-112961588 on Build GRCh38
rs782704670 Chr.11: 112961515-112961515 on Build GRCh38
rs572184546 Chr.11: 112961556-112961556 on Build GRCh38
rs543798793 Chr.11: 112961604-112961604 on Build GRCh38
rs781786391 Chr.11: 112961338-112961338 on Build GRCh38
rs782569108 Chr.11: 112961325-112961325 on Build GRCh38
rs782639709 Chr.11: 112961395-112961395 on Build GRCh38
rs782674091 Chr.11: 112961535-112961535 on Build GRCh38
rs782795743 Chr.11: 112961625-112961625 on Build GRCh38
rs782178972 Chr.11: 112961427-112961427 on Build GRCh38
rs1036070882 Chr.11: 112961505-112961505 on Build GRCh38
rs897562497 Chr.11: 112961518-112961518 on Build GRCh38
rs781891562 Chr.11: 112961378-112961378 on Build GRCh38
rs782605842 Chr.11: 112961559-112961560 on Build GRCh38
rs781852393 Chr.11: 112961557-112961558 on Build GRCh38
rs1042949461 Chr.11: 112961230-112961230 on Build GRCh38
rs782271253 Chr.11: 112961481-112961481 on Build GRCh38
rs552768555 Chr.11: 112961264-112961264 on Build GRCh38
rs782761878 Chr.11: 112961441-112961441 on Build GRCh38
rs781962609 Chr.11: 112961555-112961556 on Build GRCh38
rs781791930 Chr.11: 112961452-112961452 on Build GRCh38
rs781985812 Chr.11: 112961557-112961557 on Build GRCh38
rs890092278 Chr.11: 112961292-112961292 on Build GRCh38
rs11214438 Chr.11: 112961242-112961243 on Build GRCh38
rs781890897 Chr.11: 112961509-112961509 on Build GRCh38
rs201772924 Chr.11: 112961618-112961618 on Build GRCh38
rs782454521 Chr.11: 112961324-112961325 on Build GRCh38
rs782087726 Chr.11: 112961578-112961578 on Build GRCh38
rs796286007 Chr.11: 112961619-112961619 on Build GRCh38
rs782413774 Chr.11: 112961424-112961424 on Build GRCh38
rs781843808 Chr.11: 112961454-112961454 on Build GRCh38
rs145493999 Chr.11: 112961554-112961555 on Build GRCh38
rs782282074 Chr.11: 112961564-112961565 on Build GRCh38
rs781949758 Chr.11: 112961496-112961496 on Build GRCh38
rs781802202 Chr.11: 112961520-112961520 on Build GRCh38
rs142518273 Chr.11: 112961327-112961327 on Build GRCh38
rs782340704 Chr.11: 112961336-112961337 on Build GRCh38
rs139414593 Chr.11: 112961326-112961326 on Build GRCh38
rs782721655 Chr.11: 112961343-112961343 on Build GRCh38
rs782257271 Chr.11: 112961413-112961413 on Build GRCh38
rs782554805 Chr.11: 112961381-112961381 on Build GRCh38
rs782589694 Chr.11: 112961465-112961465 on Build GRCh38
rs782374331 Chr.11: 112961580-112961580 on Build GRCh38
rs575893571 Chr.11: 112961460-112961460 on Build GRCh38
rs782325149 Chr.11: 112961430-112961430 on Build GRCh38
rs561424527 Chr.11: 112961564-112961564 on Build GRCh38
rs535845126 Chr.11: 112961344-112961344 on Build GRCh38
rs782429101 Chr.11: 112961463-112961463 on Build GRCh38
rs782572905 Chr.11: 112961382-112961382 on Build GRCh38
rs782211394 Chr.11: 112961469-112961469 on Build GRCh38
rs201206514 Chr.11: 112961577-112961577 on Build GRCh38
rs532864012 Chr.11: 112961232-112961232 on Build GRCh38
rs782527116 Chr.11: 112961531-112961531 on Build GRCh38
rs782001399 Chr.11: 112961331-112961331 on Build GRCh38
rs897610555 Chr.11: 112961319-112961319 on Build GRCh38
rs112306738 Chr.11: 112961619-112961619 on Build GRCh38
rs199895300 Chr.11: 112961559-112961559 on Build GRCh38
rs781982002 Chr.11: 112961480-112961480 on Build GRCh38
rs781797220 Chr.11: 112961323-112961323 on Build GRCh38
rs781801429 Chr.11: 112961595-112961595 on Build GRCh38
rs782028396 Chr.11: 112961426-112961426 on Build GRCh38
rs993297944 Chr.11: 112961189-112961189 on Build GRCh38
rs371890526 Chr.11: 112961246-112961247 on Build GRCh38
rs782541618 Chr.11: 112961514-112961514 on Build GRCh38
rs992876248 Chr.11: 112961363-112961363 on Build GRCh38
rs978578168 Chr.11: 112961394-112961394 on Build GRCh38
rs1036489862 Chr.11: 112961311-112961311 on Build GRCh38
rs967382506 Chr.11: 112961576-112961576 on Build GRCh38
rs782311774 Chr.11: 112961555-112961555 on Build GRCh38
rs782810229 Chr.11: 112961334-112961334 on Build GRCh38
rs782094701 Chr.11: 112961439-112961439 on Build GRCh38
rs781792212 Chr.11: 112961558-112961559 on Build GRCh38
rs782055550 Chr.11: 112961494-112961494 on Build GRCh38
rs782778169 Chr.11: 112961506-112961506 on Build GRCh38
rs559828324 Chr.11: 112961639-112961639 on Build GRCh38
rs34274345 Chr.11: 112961396-112961397 on Build GRCh38
rs782148552 Chr.11: 112961447-112961447 on Build GRCh38
rs782062091 Chr.11: 112961555-112961555 on Build GRCh38
rs775631565 Chr.11: 112961219-112961219 on Build GRCh38
rs782602069 Chr.11: 112961526-112961526 on Build GRCh38
rs931913709 Chr.11: 112961252-112961252 on Build GRCh38
rs782710076 Chr.11: 112961495-112961495 on Build GRCh38
rs782323298 Chr.11: 112961565-112961565 on Build GRCh38
rs782151247 Chr.11: 112961584-112961584 on Build GRCh38
rs374931900 Chr.11: 112961561-112961561 on Build GRCh38
rs372842188 Chr.11: 112961398-112961398 on Build GRCh38
rs782198201 Chr.11: 112961385-112961385 on Build GRCh38
rs7105734 Chr.11: 112961631-112961631 on Build GRCh38
rs1033096683 Chr.11: 112961587-112961587 on Build GRCh38
rs782648319 Chr.11: 112961372-112961372 on Build GRCh38
rs782499528 Chr.11: 112961622-112961622 on Build GRCh38
rs558440771 Chr.11: 112961538-112961538 on Build GRCh38
rs781948052 Chr.11: 112961435-112961435 on Build GRCh38
rs563686839 Chr.11: 112961616-112961616 on Build GRCh38
rs538563752 Chr.11: 112961492-112961492 on Build GRCh38
rs782430578 Chr.11: 112961554-112961554 on Build GRCh38
rs782006975 Chr.11: 112961358-112961358 on Build GRCh38
rs782471736 Chr.11: 112961391-112961391 on Build GRCh38
rs782518318 Chr.11: 112961459-112961459 on Build GRCh38
rs945854645 Chr.11: 112961206-112961206 on Build GRCh38
rs782517564 Chr.11: 112961560-112961561 on Build GRCh38
rs782816995 Chr.11: 112961593-112961593 on Build GRCh38
rs781994961 Chr.11: 112961582-112961582 on Build GRCh38
rs555746093 Chr.11: 112961371-112961371 on Build GRCh38
rs782009096 Chr.11: 112961446-112961446 on Build GRCh38
rs782477221 Chr.11: 112961525-112961525 on Build GRCh38
rs201971024 Chr.11: 112961449-112961449 on Build GRCh38

View Full Product Details