Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30720717-30721258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
UniGene
Hs.234355
Gene Chromosome Location
Chr.12: 30709552-30754994 on Build GRCh38
Transcripts
NM_001002259.2 NM_001206856.2 NM_001319842.1 NM_001319843.1 NM_001319844.1 NM_001319845.1 NM_001319846.1 NM_023925.4 NM_032156.4 XM_006719138.3 XM_006719139.3 XM_006719140.3 XM_006719142.3 XM_006719144.3 XM_006719145.3 XM_006719146.3 XM_006719147.3 XM_017019851.1 XM_017019852.1 XM_017019853.1 XM_017019854.1 XM_017019855.1 XM_017019856.1 XM_017019857.1 XM_017019858.1 XM_017019859.1 XM_017019860.1 XM_017019861.1 XM_017019862.1 XM_017019863.1 XM_017019864.1 XM_017019865.1 XM_017019866.1 XM_017019867.1 XM_017019868.1 XM_017019869.1 XM_017019870.1 XM_017019871.1 XM_017019872.1 XM_017019873.1 XM_017019874.1 XM_017019875.1 XM_017019876.1 XM_017019877.1 XM_017019878.1 XM_017019879.1 XM_017019880.1 XM_017019881.1 XM_017019882.1 XM_017019883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415891 c.2130G>T p.M710I Chr.12: 30720829-30720829 on Build GRCh38
6620003 c.2093C>T p.T698I Chr.12: 30720866-30720866 on Build GRCh38
693391 c.2073A>G p.S691S Chr.12: 30720886-30720886 on Build GRCh38
3811836 c.2098C>T p.Q700* Chr.12: 30720861-30720861 on Build GRCh38
6334489 c.2094C>A p.T698T Chr.12: 30720865-30720865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765124823 Chr.12: 30720885-30720885 on Build GRCh38
rs773505253 Chr.12: 30720876-30720876 on Build GRCh38
rs773483476 Chr.12: 30721121-30721122 on Build GRCh38
rs994342584 Chr.12: 30721117-30721117 on Build GRCh38
rs748296074 Chr.12: 30721210-30721210 on Build GRCh38
rs770157500 Chr.12: 30720851-30720851 on Build GRCh38
rs749650843 Chr.12: 30720858-30720858 on Build GRCh38
rs376694164 Chr.12: 30720921-30720921 on Build GRCh38
rs929820330 Chr.12: 30720790-30720790 on Build GRCh38
rs199894738 Chr.12: 30720796-30720796 on Build GRCh38
rs745466108 Chr.12: 30720850-30720850 on Build GRCh38
rs761991032 Chr.12: 30720865-30720865 on Build GRCh38
rs761784802 Chr.12: 30720783-30720783 on Build GRCh38
rs796354350 Chr.12: 30720863-30720863 on Build GRCh38
rs777564624 Chr.12: 30720836-30720836 on Build GRCh38
rs767455191 Chr.12: 30720866-30720866 on Build GRCh38
rs899720874 Chr.12: 30721229-30721229 on Build GRCh38
rs760918490 Chr.12: 30720881-30720881 on Build GRCh38
rs969477700 Chr.12: 30721020-30721020 on Build GRCh38
rs758251146 Chr.12: 30720894-30720894 on Build GRCh38
rs369690765 Chr.12: 30720938-30720938 on Build GRCh38
rs929725390 Chr.12: 30721070-30721070 on Build GRCh38
rs912385592 Chr.12: 30721150-30721150 on Build GRCh38
rs371879635 Chr.12: 30720864-30720864 on Build GRCh38
rs746930091 Chr.12: 30721014-30721014 on Build GRCh38
rs139160988 Chr.12: 30720838-30720838 on Build GRCh38
rs964023999 Chr.12: 30721222-30721222 on Build GRCh38
rs761719235 Chr.12: 30720753-30720754 on Build GRCh38
rs935276917 Chr.12: 30721231-30721231 on Build GRCh38
rs768799934 Chr.12: 30721114-30721114 on Build GRCh38
rs774952623 Chr.12: 30720926-30720926 on Build GRCh38
rs200247569 Chr.12: 30720811-30720811 on Build GRCh38
rs780510734 Chr.12: 30720917-30720917 on Build GRCh38
rs955572896 Chr.12: 30721157-30721157 on Build GRCh38
rs764134238 Chr.12: 30720903-30720903 on Build GRCh38
rs775765874 Chr.12: 30720853-30720853 on Build GRCh38
rs754849907 Chr.12: 30720799-30720799 on Build GRCh38
rs772202346 Chr.12: 30720943-30720943 on Build GRCh38
rs748555079 Chr.12: 30720933-30720933 on Build GRCh38
rs1005865873 Chr.12: 30721086-30721086 on Build GRCh38
rs925184940 Chr.12: 30721228-30721228 on Build GRCh38
rs760830376 Chr.12: 30720958-30720958 on Build GRCh38
rs560930202 Chr.12: 30720839-30720839 on Build GRCh38
rs1040046628 Chr.12: 30721196-30721196 on Build GRCh38
rs372431345 Chr.12: 30720912-30720912 on Build GRCh38
rs560341042 Chr.12: 30721123-30721123 on Build GRCh38
rs368786279 Chr.12: 30720909-30720909 on Build GRCh38
rs925216167 Chr.12: 30720982-30720983 on Build GRCh38
rs751729978 Chr.12: 30720778-30720778 on Build GRCh38
rs758341127 Chr.12: 30720813-30720813 on Build GRCh38
rs771372968 Chr.12: 30720835-30720836 on Build GRCh38
rs1022139988 Chr.12: 30720878-30720878 on Build GRCh38
rs752603335 Chr.12: 30720887-30720887 on Build GRCh38
rs1023762102 Chr.12: 30720743-30720743 on Build GRCh38
rs780932468 Chr.12: 30720844-30720844 on Build GRCh38
rs1013713909 Chr.12: 30721050-30721050 on Build GRCh38
rs992137667 Chr.12: 30720998-30720998 on Build GRCh38
rs564893223 Chr.12: 30721015-30721015 on Build GRCh38
rs368031265 Chr.12: 30720859-30720859 on Build GRCh38
rs543004335 Chr.12: 30720967-30720967 on Build GRCh38
rs371696129 Chr.12: 30720751-30720751 on Build GRCh38
rs781415800 Chr.12: 30721165-30721165 on Build GRCh38
rs755832558 Chr.12: 30720920-30720920 on Build GRCh38
rs752691194 Chr.12: 30720805-30720805 on Build GRCh38
rs745525881 Chr.12: 30720919-30720919 on Build GRCh38
rs576115546 Chr.12: 30720950-30720950 on Build GRCh38
rs896235441 Chr.12: 30721041-30721041 on Build GRCh38
rs149800449 Chr.12: 30721037-30721037 on Build GRCh38
rs768710336 Chr.12: 30721082-30721082 on Build GRCh38
rs111415714 Chr.12: 30721051-30721051 on Build GRCh38
rs375108290 Chr.12: 30720807-30720807 on Build GRCh38
rs979458853 Chr.12: 30721226-30721226 on Build GRCh38
rs1017126741 Chr.12: 30721091-30721091 on Build GRCh38
rs935140455 Chr.12: 30721002-30721002 on Build GRCh38
rs371864898 Chr.12: 30720802-30720802 on Build GRCh38
rs749700002 Chr.12: 30720923-30720923 on Build GRCh38
rs1043643461 Chr.12: 30720968-30720968 on Build GRCh38
rs960312428 Chr.12: 30720991-30720991 on Build GRCh38
rs1026192384 Chr.12: 30721142-30721142 on Build GRCh38
rs767827955 Chr.12: 30720791-30720791 on Build GRCh38
rs879680543 Chr.12: 30721203-30721203 on Build GRCh38

View Full Product Details