Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2588841-2589343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213B
Entrez Gene ID
127281
Gene Name
family with sequence similarity 213 member B
Gene Aliases
C1orf93
UniGene
Hs.462033
Gene Chromosome Location
Chr.1: 2586460-2591469 on Build GRCh38
Transcripts
NM_001195736.1 NM_001195737.1 NM_001195738.1 NM_001195740.1 NM_001195741.1 NM_152371.3 NR_036637.1 NR_036638.1 XM_006710354.3 XM_011540664.1 XM_011540665.1 XM_011540666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3487355 c.625C>T p.P209S Chr.1: 2589032-2589032 on Build GRCh38
6659459 c.511G>A p.V171I Chr.1: 2588972-2588972 on Build GRCh38
6659464 c.552G>A p.A184A Chr.1: 2589013-2589013 on Build GRCh38
907454 c.486C>T p.F162F Chr.1: 2588947-2588947 on Build GRCh38
4235963 c.551C>T p.S184F Chr.1: 2588958-2588958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374933565 Chr.1: 2589012-2589012 on Build GRCh38
rs989874706 Chr.1: 2589169-2589169 on Build GRCh38
rs768849870 Chr.1: 2589028-2589028 on Build GRCh38
rs78004063 Chr.1: 2589184-2589184 on Build GRCh38
rs977347681 Chr.1: 2588965-2588965 on Build GRCh38
rs998663560 Chr.1: 2589291-2589291 on Build GRCh38
rs767840240 Chr.1: 2589136-2589136 on Build GRCh38
rs760220237 Chr.1: 2589051-2589051 on Build GRCh38
rs781130096 Chr.1: 2589004-2589004 on Build GRCh38
rs755775415 Chr.1: 2588921-2588921 on Build GRCh38
rs529258905 Chr.1: 2588885-2588885 on Build GRCh38
rs199801631 Chr.1: 2589064-2589064 on Build GRCh38
rs749994297 Chr.1: 2588916-2588916 on Build GRCh38
rs141147759 Chr.1: 2589024-2589024 on Build GRCh38
rs749146460 Chr.1: 2588929-2588929 on Build GRCh38
rs751055105 Chr.1: 2588902-2588902 on Build GRCh38
rs779140263 Chr.1: 2589090-2589090 on Build GRCh38
rs948132852 Chr.1: 2589174-2589174 on Build GRCh38
rs139936708 Chr.1: 2589077-2589077 on Build GRCh38
rs528065205 Chr.1: 2588912-2588912 on Build GRCh38
rs371656711 Chr.1: 2588961-2588961 on Build GRCh38
rs758257557 Chr.1: 2589068-2589068 on Build GRCh38
rs778792541 Chr.1: 2588995-2588995 on Build GRCh38
rs762160893 Chr.1: 2589033-2589033 on Build GRCh38
rs536957046 Chr.1: 2589191-2589191 on Build GRCh38
rs566881045 Chr.1: 2589086-2589086 on Build GRCh38
rs766878796 Chr.1: 2588974-2588974 on Build GRCh38
rs941648587 Chr.1: 2589238-2589238 on Build GRCh38
rs763457229 Chr.1: 2588971-2588971 on Build GRCh38
rs766070835 Chr.1: 2588986-2588986 on Build GRCh38
rs1031495295 Chr.1: 2589298-2589298 on Build GRCh38
rs759829520 Chr.1: 2588889-2588889 on Build GRCh38
rs1038423957 Chr.1: 2589263-2589263 on Build GRCh38
rs565565761 Chr.1: 2588972-2588972 on Build GRCh38
rs551150393 Chr.1: 2589038-2589038 on Build GRCh38
rs780299564 Chr.1: 2589085-2589085 on Build GRCh38
rs542482574 Chr.1: 2589293-2589293 on Build GRCh38
rs184894716 Chr.1: 2589275-2589275 on Build GRCh38
rs980914726 Chr.1: 2589182-2589182 on Build GRCh38
rs749952576 Chr.1: 2588979-2588979 on Build GRCh38
rs770211827 Chr.1: 2588967-2588967 on Build GRCh38
rs955242260 Chr.1: 2589111-2589111 on Build GRCh38
rs112011977 Chr.1: 2589200-2589200 on Build GRCh38
rs895256183 Chr.1: 2588893-2588897 on Build GRCh38
rs1031552259 Chr.1: 2589137-2589137 on Build GRCh38
rs369682917 Chr.1: 2589029-2589029 on Build GRCh38
rs78851285 Chr.1: 2589304-2589304 on Build GRCh38
rs1046531419 Chr.1: 2589162-2589162 on Build GRCh38
rs868071995 Chr.1: 2588891-2588891 on Build GRCh38
rs6701238 Chr.1: 2589061-2589061 on Build GRCh38
rs748149450 Chr.1: 2588951-2588951 on Build GRCh38
rs768583008 Chr.1: 2588877-2588877 on Build GRCh38
rs758364600 Chr.1: 2589001-2589001 on Build GRCh38
rs544710926 Chr.1: 2588970-2588970 on Build GRCh38
rs770115843 Chr.1: 2589008-2589008 on Build GRCh38
rs200164303 Chr.1: 2588948-2588948 on Build GRCh38
rs997856343 Chr.1: 2589283-2589283 on Build GRCh38
rs753118730 Chr.1: 2588897-2588897 on Build GRCh38
rs746014507 Chr.1: 2588966-2588966 on Build GRCh38
rs370643221 Chr.1: 2589266-2589266 on Build GRCh38
rs963886410 Chr.1: 2588906-2588906 on Build GRCh38
rs115817728 Chr.1: 2588989-2588989 on Build GRCh38
rs375013831 Chr.1: 2588898-2588898 on Build GRCh38
rs746005922 Chr.1: 2589005-2589005 on Build GRCh38
rs776677627 Chr.1: 2588888-2588888 on Build GRCh38
rs1047322689 Chr.1: 2589220-2589220 on Build GRCh38
rs1048298001 Chr.1: 2589196-2589197 on Build GRCh38
rs760718442 Chr.1: 2588883-2588883 on Build GRCh38
rs374419742 Chr.1: 2588911-2588911 on Build GRCh38
rs915268615 Chr.1: 2589172-2589172 on Build GRCh38
rs373074035 Chr.1: 2589034-2589034 on Build GRCh38
rs1050808904 Chr.1: 2589278-2589278 on Build GRCh38
rs755758710 Chr.1: 2588980-2588980 on Build GRCh38
rs770867206 Chr.1: 2588952-2588952 on Build GRCh38
rs556702529 Chr.1: 2589197-2589197 on Build GRCh38
rs775286283 Chr.1: 2589140-2589141 on Build GRCh38
rs569040418 Chr.1: 2589208-2589208 on Build GRCh38
rs775892312 Chr.1: 2589013-2589013 on Build GRCh38
rs1029905788 Chr.1: 2589107-2589107 on Build GRCh38
rs1011533178 Chr.1: 2589312-2589312 on Build GRCh38
rs890920482 Chr.1: 2589279-2589279 on Build GRCh38
rs749548230 Chr.1: 2589025-2589025 on Build GRCh38
rs750497324 Chr.1: 2589084-2589084 on Build GRCh38
rs747981119 Chr.1: 2588998-2588998 on Build GRCh38
rs955271310 Chr.1: 2588919-2588919 on Build GRCh38
rs1047355667 Chr.1: 2589039-2589039 on Build GRCh38
rs752608434 Chr.1: 2589066-2589066 on Build GRCh38
rs1042632483 Chr.1: 2588864-2588864 on Build GRCh38
rs765907216 Chr.1: 2589060-2589060 on Build GRCh38
rs558729234 Chr.1: 2589117-2589117 on Build GRCh38
rs6667564 Chr.1: 2588863-2588863 on Build GRCh38
rs956680922 Chr.1: 2589138-2589138 on Build GRCh38
rs373599830 Chr.1: 2588882-2588882 on Build GRCh38
rs143473292 Chr.1: 2588947-2588947 on Build GRCh38
rs779756816 Chr.1: 2588924-2588924 on Build GRCh38
rs754640027 Chr.1: 2589062-2589062 on Build GRCh38
rs765643404 Chr.1: 2588892-2588892 on Build GRCh38
rs887368733 Chr.1: 2589221-2589221 on Build GRCh38
rs368991978 Chr.1: 2588899-2588899 on Build GRCh38

View Full Product Details