Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103305832-103306351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf42
Entrez Gene ID
374470
Gene Name
chromosome 12 open reading frame 42
Gene Aliases
-
UniGene
Hs.534649
Gene Chromosome Location
Chr.12: 103047669-103525258 on Build GRCh38
Transcripts
NM_001099336.2 NM_001278419.1 NM_001278420.1 NM_198521.3 NR_103526.1 XM_006719377.3 XM_011538291.1 XM_011538292.2 XM_011538293.2 XM_011538294.2 XM_011538295.1 XM_011538297.2 XM_011538300.1 XM_011538305.1 XM_011538306.1 XM_011538308.2 XM_011538312.2 XM_011538315.2 XM_017019276.1 XM_017019277.1 XM_017019278.1 XM_017019279.1 XM_017019280.1 XM_017019281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4333423 c.559G>T p.A187S Chr.12: 103306046-103306046 on Build GRCh38
5469123 c.395C>A p.A132E Chr.12: 103306210-103306210 on Build GRCh38
271009 c.382C>T p.R128C Chr.12: 103306223-103306223 on Build GRCh38
5596376 c.402C>T p.S134S Chr.12: 103306203-103306203 on Build GRCh38
4502564 c.616C>T p.P206S Chr.12: 103305989-103305989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370514326 Chr.12: 103306214-103306214 on Build GRCh38
rs757549027 Chr.12: 103306281-103306281 on Build GRCh38
rs759166157 Chr.12: 103305977-103305977 on Build GRCh38
rs760427771 Chr.12: 103305996-103305996 on Build GRCh38
rs746339721 Chr.12: 103306183-103306183 on Build GRCh38
rs756366371 Chr.12: 103306039-103306039 on Build GRCh38
rs762265739 Chr.12: 103306314-103306314 on Build GRCh38
rs779221742 Chr.12: 103306074-103306074 on Build GRCh38
rs775116204 Chr.12: 103306107-103306107 on Build GRCh38
rs373580415 Chr.12: 103305966-103305966 on Build GRCh38
rs7488867 Chr.12: 103305907-103305907 on Build GRCh38
rs753726249 Chr.12: 103306007-103306007 on Build GRCh38
rs765087581 Chr.12: 103306129-103306129 on Build GRCh38
rs774213107 Chr.12: 103305958-103305960 on Build GRCh38
rs965739864 Chr.12: 103306026-103306026 on Build GRCh38
rs756311422 Chr.12: 103306172-103306172 on Build GRCh38
rs78107859 Chr.12: 103306169-103306169 on Build GRCh38
rs778102758 Chr.12: 103306041-103306041 on Build GRCh38
rs746416298 Chr.12: 103306083-103306083 on Build GRCh38
rs1046279266 Chr.12: 103305871-103305871 on Build GRCh38
rs767358170 Chr.12: 103306144-103306145 on Build GRCh38
rs757257534 Chr.12: 103306010-103306010 on Build GRCh38
rs771435436 Chr.12: 103306086-103306086 on Build GRCh38
rs369523668 Chr.12: 103305999-103305999 on Build GRCh38
rs558454593 Chr.12: 103306253-103306253 on Build GRCh38
rs754948537 Chr.12: 103306031-103306031 on Build GRCh38
rs767461340 Chr.12: 103306159-103306159 on Build GRCh38
rs764188160 Chr.12: 103306261-103306261 on Build GRCh38
rs778250846 Chr.12: 103305940-103305940 on Build GRCh38
rs966526947 Chr.12: 103306278-103306278 on Build GRCh38
rs951583958 Chr.12: 103306075-103306075 on Build GRCh38
rs58399247 Chr.12: 103306226-103306226 on Build GRCh38
rs772867120 Chr.12: 103306326-103306326 on Build GRCh38
rs761646627 Chr.12: 103306128-103306128 on Build GRCh38
rs760372689 Chr.12: 103306111-103306111 on Build GRCh38
rs769332092 Chr.12: 103305991-103305991 on Build GRCh38
rs746712205 Chr.12: 103306245-103306245 on Build GRCh38
rs372635785 Chr.12: 103306199-103306199 on Build GRCh38
rs190115385 Chr.12: 103306289-103306289 on Build GRCh38
rs776357950 Chr.12: 103306218-103306218 on Build GRCh38
rs781367117 Chr.12: 103306035-103306035 on Build GRCh38
rs137908805 Chr.12: 103305945-103305945 on Build GRCh38
rs201864057 Chr.12: 103306293-103306293 on Build GRCh38
rs776558001 Chr.12: 103306121-103306121 on Build GRCh38
rs762790379 Chr.12: 103306233-103306233 on Build GRCh38
rs940095831 Chr.12: 103306005-103306005 on Build GRCh38
rs1048963339 Chr.12: 103305972-103305972 on Build GRCh38
rs7488894 Chr.12: 103305866-103305866 on Build GRCh38
rs753036505 Chr.12: 103306276-103306277 on Build GRCh38
rs768490783 Chr.12: 103306117-103306117 on Build GRCh38
rs1053526067 Chr.12: 103305956-103305956 on Build GRCh38
rs377122822 Chr.12: 103305969-103305969 on Build GRCh38
rs750868580 Chr.12: 103306291-103306291 on Build GRCh38
rs376898243 Chr.12: 103306208-103306208 on Build GRCh38
rs112647633 Chr.12: 103305870-103305870 on Build GRCh38
rs754010210 Chr.12: 103306275-103306275 on Build GRCh38
rs772676234 Chr.12: 103306188-103306188 on Build GRCh38
rs949060557 Chr.12: 103306209-103306209 on Build GRCh38
rs769552310 Chr.12: 103306319-103306319 on Build GRCh38
rs771297869 Chr.12: 103306072-103306072 on Build GRCh38
rs999410491 Chr.12: 103305934-103305934 on Build GRCh38
rs916222287 Chr.12: 103305869-103305869 on Build GRCh38
rs944297973 Chr.12: 103305856-103305856 on Build GRCh38
rs557524390 Chr.12: 103306313-103306313 on Build GRCh38
rs777914047 Chr.12: 103306177-103306177 on Build GRCh38
rs773095957 Chr.12: 103306229-103306229 on Build GRCh38
rs749759295 Chr.12: 103305973-103305973 on Build GRCh38
rs773897801 Chr.12: 103305981-103305981 on Build GRCh38
rs779426620 Chr.12: 103306192-103306192 on Build GRCh38
rs199701137 Chr.12: 103306222-103306222 on Build GRCh38
rs958939239 Chr.12: 103306140-103306140 on Build GRCh38
rs754063412 Chr.12: 103306181-103306181 on Build GRCh38
rs747525647 Chr.12: 103306302-103306302 on Build GRCh38
rs775024106 Chr.12: 103306058-103306058 on Build GRCh38
rs766395571 Chr.12: 103306234-103306234 on Build GRCh38
rs200919713 Chr.12: 103306131-103306131 on Build GRCh38
rs887231179 Chr.12: 103305982-103305982 on Build GRCh38
rs781216290 Chr.12: 103305957-103305957 on Build GRCh38
rs984373007 Chr.12: 103306077-103306077 on Build GRCh38
rs756393301 Chr.12: 103306283-103306283 on Build GRCh38
rs780653238 Chr.12: 103306300-103306300 on Build GRCh38
rs747720849 Chr.12: 103306315-103306315 on Build GRCh38
rs7484376 Chr.12: 103306060-103306060 on Build GRCh38
rs376295502 Chr.12: 103306182-103306182 on Build GRCh38
rs765148696 Chr.12: 103306018-103306018 on Build GRCh38
rs762994360 Chr.12: 103306147-103306147 on Build GRCh38
rs748293570 Chr.12: 103306038-103306038 on Build GRCh38
rs935210215 Chr.12: 103306257-103306258 on Build GRCh38
rs751508320 Chr.12: 103306236-103306236 on Build GRCh38
rs200707105 Chr.12: 103305971-103305971 on Build GRCh38
rs758912596 Chr.12: 103305988-103305988 on Build GRCh38
rs767056248 Chr.12: 103305990-103305990 on Build GRCh38

View Full Product Details