Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110636957-110637455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAXD
Entrez Gene ID
55739
Gene Name
NAD(P)HX dehydratase
Gene Aliases
CARKD, LP3298
UniGene
Hs.408324
Gene Chromosome Location
Chr.13: 110615460-110639996 on Build GRCh38
Transcripts
NM_001242881.1 NM_001242882.1 NM_001242883.1 NM_018210.3 NR_040103.1 NR_040104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4045969 c.833C>T p.S278L Chr.13: 110637189-110637189 on Build GRCh38
2263234 c.863C>T p.A288V Chr.13: 110637219-110637219 on Build GRCh38
3467250 c.834G>A p.S278S Chr.13: 110637190-110637190 on Build GRCh38
7294391 c.846C>T p.G282G Chr.13: 110637202-110637202 on Build GRCh38
4600145 c.847G>A p.V283I Chr.13: 110637203-110637203 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751836366 Chr.13: 110637104-110637106 on Build GRCh38
rs746651071 Chr.13: 110637081-110637081 on Build GRCh38
rs774315079 Chr.13: 110637249-110637249 on Build GRCh38
rs567458785 Chr.13: 110637309-110637309 on Build GRCh38
rs758770785 Chr.13: 110637285-110637285 on Build GRCh38
rs892175537 Chr.13: 110637046-110637046 on Build GRCh38
rs773029675 Chr.13: 110637237-110637237 on Build GRCh38
rs1016362973 Chr.13: 110637200-110637200 on Build GRCh38
rs184339299 Chr.13: 110637346-110637346 on Build GRCh38
rs750925994 Chr.13: 110637273-110637273 on Build GRCh38
rs531045384 Chr.13: 110637350-110637350 on Build GRCh38
rs934155545 Chr.13: 110637280-110637280 on Build GRCh38
rs368407833 Chr.13: 110637258-110637258 on Build GRCh38
rs1006325282 Chr.13: 110637189-110637189 on Build GRCh38
rs189115635 Chr.13: 110637397-110637397 on Build GRCh38
rs565496908 Chr.13: 110637071-110637071 on Build GRCh38
rs901477753 Chr.13: 110637149-110637149 on Build GRCh38
rs767572333 Chr.13: 110637255-110637255 on Build GRCh38
rs113797046 Chr.13: 110637400-110637400 on Build GRCh38
rs928624591 Chr.13: 110637011-110637011 on Build GRCh38
rs759461319 Chr.13: 110637252-110637252 on Build GRCh38
rs779207809 Chr.13: 110637272-110637272 on Build GRCh38
rs372132546 Chr.13: 110637063-110637063 on Build GRCh38
rs201905540 Chr.13: 110637104-110637104 on Build GRCh38
rs553247594 Chr.13: 110637338-110637338 on Build GRCh38
rs1021887568 Chr.13: 110637057-110637057 on Build GRCh38
rs909906346 Chr.13: 110637228-110637228 on Build GRCh38
rs1016456054 Chr.13: 110636997-110636997 on Build GRCh38
rs374886391 Chr.13: 110637220-110637220 on Build GRCh38
rs116033188 Chr.13: 110637032-110637032 on Build GRCh38
rs534574113 Chr.13: 110637320-110637320 on Build GRCh38
rs330573 Chr.13: 110637045-110637045 on Build GRCh38
rs768097312 Chr.13: 110637101-110637101 on Build GRCh38
rs773082677 Chr.13: 110637118-110637118 on Build GRCh38
rs147252532 Chr.13: 110637202-110637202 on Build GRCh38
rs561275871 Chr.13: 110637412-110637412 on Build GRCh38
rs770681179 Chr.13: 110637248-110637248 on Build GRCh38
rs757607976 Chr.13: 110637269-110637269 on Build GRCh38
rs537226052 Chr.13: 110637223-110637223 on Build GRCh38
rs1039378722 Chr.13: 110637088-110637088 on Build GRCh38
rs143155796 Chr.13: 110637404-110637404 on Build GRCh38
rs191908670 Chr.13: 110637260-110637260 on Build GRCh38
rs896371498 Chr.13: 110637385-110637385 on Build GRCh38
rs199637796 Chr.13: 110637190-110637190 on Build GRCh38
rs932973949 Chr.13: 110637335-110637335 on Build GRCh38
rs3825467 Chr.13: 110637351-110637351 on Build GRCh38
rs61745623 Chr.13: 110637159-110637159 on Build GRCh38
rs777545221 Chr.13: 110637300-110637300 on Build GRCh38
rs1043609691 Chr.13: 110637050-110637050 on Build GRCh38
rs1035939660 Chr.13: 110637402-110637402 on Build GRCh38
rs567699584 Chr.13: 110637389-110637389 on Build GRCh38
rs536741459 Chr.13: 110637433-110637433 on Build GRCh38
rs1055454588 Chr.13: 110637358-110637358 on Build GRCh38
rs970643319 Chr.13: 110637201-110637201 on Build GRCh38
rs921521143 Chr.13: 110637263-110637263 on Build GRCh38
rs945104436 Chr.13: 110637314-110637314 on Build GRCh38
rs1038914294 Chr.13: 110637321-110637321 on Build GRCh38
rs370194200 Chr.13: 110637111-110637111 on Build GRCh38
rs542848282 Chr.13: 110637021-110637021 on Build GRCh38
rs1039959044 Chr.13: 110636984-110636984 on Build GRCh38
rs766317263 Chr.13: 110637094-110637095 on Build GRCh38
rs763102231 Chr.13: 110637081-110637082 on Build GRCh38
rs747619640 Chr.13: 110637288-110637288 on Build GRCh38
rs756387024 Chr.13: 110637170-110637170 on Build GRCh38
rs1013851760 Chr.13: 110637394-110637394 on Build GRCh38
rs1020273860 Chr.13: 110637427-110637427 on Build GRCh38
rs762659161 Chr.13: 110637122-110637122 on Build GRCh38
rs978861471 Chr.13: 110637264-110637264 on Build GRCh38
rs943279606 Chr.13: 110637064-110637064 on Build GRCh38
rs938688942 Chr.13: 110637016-110637016 on Build GRCh38
rs751472798 Chr.13: 110637143-110637143 on Build GRCh38
rs200659241 Chr.13: 110637296-110637296 on Build GRCh38
rs147247524 Chr.13: 110637009-110637009 on Build GRCh38
rs780904587 Chr.13: 110637208-110637208 on Build GRCh38
rs764265790 Chr.13: 110637259-110637259 on Build GRCh38
rs970357671 Chr.13: 110637008-110637008 on Build GRCh38
rs913665945 Chr.13: 110637311-110637311 on Build GRCh38
rs780710325 Chr.13: 110637286-110637286 on Build GRCh38
rs779430450 Chr.13: 110637197-110637197 on Build GRCh38
rs771326264 Chr.13: 110637158-110637158 on Build GRCh38
rs531114784 Chr.13: 110637203-110637203 on Build GRCh38
rs913360005 Chr.13: 110637037-110637037 on Build GRCh38
rs1033593802 Chr.13: 110637116-110637116 on Build GRCh38
rs771785434 Chr.13: 110637024-110637024 on Build GRCh38
rs923212545 Chr.13: 110637322-110637322 on Build GRCh38
rs769695070 Chr.13: 110637114-110637114 on Build GRCh38
rs1009194229 Chr.13: 110637053-110637053 on Build GRCh38
rs563465580 Chr.13: 110636985-110636985 on Build GRCh38
rs997596703 Chr.13: 110637157-110637157 on Build GRCh38

View Full Product Details