Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49992693-49993187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01588
Entrez Gene ID
283551
Gene Name
long intergenic non-protein coding RNA 1588
Gene Aliases
C14orf182
UniGene
Hs.292580
Gene Chromosome Location
Chr.14: 49981669-50007520 on Build GRCh38
Transcripts
NR_126496.1 NR_126497.1 NR_126498.1 NR_126499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419831 c.285C>T p.D95D Chr.14: 49992795-49992795 on Build GRCh38
2198217 c.225G>A p.M75I Chr.14: 49992855-49992855 on Build GRCh38
955983 c.240C>T p.L80L Chr.14: 49992840-49992840 on Build GRCh38
5470241 c.260T>G p.V87G Chr.14: 49992820-49992820 on Build GRCh38
7092417 c.235A>G p.K79E Chr.14: 49992845-49992845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372407267 Chr.14: 49992916-49992916 on Build GRCh38
rs751038236 Chr.14: 49992945-49992945 on Build GRCh38
rs562782021 Chr.14: 49992856-49992856 on Build GRCh38
rs747391857 Chr.14: 49992833-49992833 on Build GRCh38
rs769062126 Chr.14: 49992838-49992840 on Build GRCh38
rs759364941 Chr.14: 49992868-49992868 on Build GRCh38
rs769414680 Chr.14: 49992925-49992925 on Build GRCh38
rs368686710 Chr.14: 49992775-49992775 on Build GRCh38
rs952037454 Chr.14: 49992905-49992905 on Build GRCh38
rs61749566 Chr.14: 49993022-49993022 on Build GRCh38
rs762499945 Chr.14: 49992861-49992861 on Build GRCh38
rs995226301 Chr.14: 49993078-49993078 on Build GRCh38
rs546585412 Chr.14: 49992797-49992797 on Build GRCh38
rs775017614 Chr.14: 49992783-49992784 on Build GRCh38
rs34909517 Chr.14: 49992846-49992847 on Build GRCh38
rs61748277 Chr.14: 49992880-49992880 on Build GRCh38
rs769467881 Chr.14: 49992857-49992857 on Build GRCh38
rs777682916 Chr.14: 49993025-49993026 on Build GRCh38
rs10134096 Chr.14: 49992955-49992955 on Build GRCh38
rs754278041 Chr.14: 49992788-49992788 on Build GRCh38
rs554547022 Chr.14: 49993060-49993060 on Build GRCh38
rs965072416 Chr.14: 49992723-49992723 on Build GRCh38
rs1039523320 Chr.14: 49992973-49992973 on Build GRCh38
rs780595005 Chr.14: 49992913-49992913 on Build GRCh38
rs761220278 Chr.14: 49992832-49992832 on Build GRCh38
rs754771670 Chr.14: 49992752-49992752 on Build GRCh38
rs940749490 Chr.14: 49992926-49992926 on Build GRCh38
rs566418769 Chr.14: 49992821-49992821 on Build GRCh38
rs765601610 Chr.14: 49992887-49992887 on Build GRCh38
rs777065654 Chr.14: 49992850-49992850 on Build GRCh38
rs202106679 Chr.14: 49992737-49992737 on Build GRCh38
rs781370229 Chr.14: 49992922-49992922 on Build GRCh38
rs369591169 Chr.14: 49993083-49993083 on Build GRCh38
rs146890607 Chr.14: 49992734-49992734 on Build GRCh38
rs534058959 Chr.14: 49993151-49993151 on Build GRCh38
rs1050663756 Chr.14: 49993015-49993015 on Build GRCh38
rs1052076094 Chr.14: 49993047-49993047 on Build GRCh38
rs745326375 Chr.14: 49992727-49992728 on Build GRCh38
rs10134002 Chr.14: 49992891-49992891 on Build GRCh38
rs892149506 Chr.14: 49993077-49993077 on Build GRCh38
rs936310457 Chr.14: 49993044-49993044 on Build GRCh38
rs976207729 Chr.14: 49992930-49992930 on Build GRCh38
rs771559701 Chr.14: 49992781-49992781 on Build GRCh38
rs937665350 Chr.14: 49992881-49992881 on Build GRCh38
rs758614459 Chr.14: 49992908-49992908 on Build GRCh38
rs774223261 Chr.14: 49992865-49992865 on Build GRCh38
rs768958021 Chr.14: 49992837-49992837 on Build GRCh38
rs527635410 Chr.14: 49992796-49992796 on Build GRCh38
rs772679789 Chr.14: 49992813-49992813 on Build GRCh38
rs778052981 Chr.14: 49992756-49992756 on Build GRCh38
rs949144981 Chr.14: 49992846-49992846 on Build GRCh38
rs904141022 Chr.14: 49993024-49993024 on Build GRCh38
rs766310159 Chr.14: 49992864-49992864 on Build GRCh38
rs749995552 Chr.14: 49992933-49992933 on Build GRCh38
rs751347397 Chr.14: 49992746-49992746 on Build GRCh38
rs908352575 Chr.14: 49992824-49992824 on Build GRCh38
rs554621805 Chr.14: 49992931-49992931 on Build GRCh38
rs537563122 Chr.14: 49992921-49992921 on Build GRCh38
rs1009025479 Chr.14: 49992963-49992963 on Build GRCh38
rs764668400 Chr.14: 49992728-49992728 on Build GRCh38
rs116099570 Chr.14: 49992745-49992745 on Build GRCh38
rs188832056 Chr.14: 49992874-49992874 on Build GRCh38
rs140722402 Chr.14: 49992946-49992946 on Build GRCh38
rs952641588 Chr.14: 49993160-49993160 on Build GRCh38
rs900531049 Chr.14: 49992995-49992995 on Build GRCh38
rs1042587915 Chr.14: 49992950-49992950 on Build GRCh38
rs752548335 Chr.14: 49992875-49992875 on Build GRCh38

View Full Product Details