Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99502613-99503112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNK
Entrez Gene ID
8812
Gene Name
cyclin K
Gene Aliases
CPR4
UniGene
Hs.510409
Gene Chromosome Location
Chr.14: 99481402-99511515 on Build GRCh38
Transcripts
NM_001099402.1 XM_005268154.4 XM_011537275.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4514438 c.974C>T p.P325L Chr.14: 99502947-99502947 on Build GRCh38
4053400 c.978G>T p.Q326H Chr.14: 99502951-99502951 on Build GRCh38
6271618 c.858G>T p.P286P Chr.14: 99502831-99502831 on Build GRCh38
5740210 c.976C>T p.Q326* Chr.14: 99502949-99502949 on Build GRCh38
4148335 c.838C>A p.Q280K Chr.14: 99502811-99502811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981692579 Chr.14: 99502644-99502644 on Build GRCh38
rs901355751 Chr.14: 99502688-99502688 on Build GRCh38
rs193115185 Chr.14: 99502684-99502684 on Build GRCh38
rs200034824 Chr.14: 99502830-99502830 on Build GRCh38
rs372216197 Chr.14: 99502779-99502779 on Build GRCh38
rs534989573 Chr.14: 99502908-99502908 on Build GRCh38
rs771816459 Chr.14: 99503065-99503065 on Build GRCh38
rs576158425 Chr.14: 99503075-99503075 on Build GRCh38
rs781120753 Chr.14: 99502780-99502780 on Build GRCh38
rs773882638 Chr.14: 99502841-99502841 on Build GRCh38
rs761142587 Chr.14: 99502906-99502907 on Build GRCh38
rs749107194 Chr.14: 99502927-99502927 on Build GRCh38
rs761124604 Chr.14: 99502969-99502969 on Build GRCh38
rs770288405 Chr.14: 99503080-99503080 on Build GRCh38
rs777791031 Chr.14: 99502894-99502894 on Build GRCh38
rs554258238 Chr.14: 99503083-99503083 on Build GRCh38
rs370035979 Chr.14: 99503009-99503009 on Build GRCh38
rs369597761 Chr.14: 99502852-99502852 on Build GRCh38
rs897973962 Chr.14: 99503006-99503006 on Build GRCh38
rs200503921 Chr.14: 99502973-99502973 on Build GRCh38
rs768325984 Chr.14: 99502823-99502823 on Build GRCh38
rs566277343 Chr.14: 99502864-99502864 on Build GRCh38
rs752948207 Chr.14: 99502982-99502982 on Build GRCh38
rs780030047 Chr.14: 99503022-99503022 on Build GRCh38
rs773891711 Chr.14: 99502680-99502680 on Build GRCh38
rs374411620 Chr.14: 99503015-99503015 on Build GRCh38
rs771462544 Chr.14: 99502970-99502970 on Build GRCh38
rs377647054 Chr.14: 99503003-99503003 on Build GRCh38
rs764363364 Chr.14: 99502907-99502909 on Build GRCh38
rs967791189 Chr.14: 99503057-99503057 on Build GRCh38
rs931964755 Chr.14: 99502877-99502877 on Build GRCh38
rs754025705 Chr.14: 99502710-99502710 on Build GRCh38
rs756058163 Chr.14: 99502909-99502909 on Build GRCh38
rs1011876216 Chr.14: 99503034-99503034 on Build GRCh38
rs778898292 Chr.14: 99502738-99502738 on Build GRCh38
rs184863233 Chr.14: 99503033-99503033 on Build GRCh38
rs759957278 Chr.14: 99502977-99502977 on Build GRCh38
rs748943823 Chr.14: 99502820-99502820 on Build GRCh38
rs776969319 Chr.14: 99503088-99503089 on Build GRCh38
rs762627982 Chr.14: 99502672-99502672 on Build GRCh38
rs756548583 Chr.14: 99502695-99502695 on Build GRCh38
rs775308532 Chr.14: 99502804-99502804 on Build GRCh38
rs752858323 Chr.14: 99502878-99502878 on Build GRCh38
rs972801325 Chr.14: 99503032-99503032 on Build GRCh38
rs372835830 Chr.14: 99502750-99502750 on Build GRCh38
rs200877327 Chr.14: 99502925-99502925 on Build GRCh38
rs189193359 Chr.14: 99503081-99503081 on Build GRCh38
rs768254731 Chr.14: 99502945-99502945 on Build GRCh38
rs747848206 Chr.14: 99503050-99503050 on Build GRCh38
rs372907456 Chr.14: 99502838-99502838 on Build GRCh38
rs754196878 Chr.14: 99502863-99502863 on Build GRCh38
rs751012933 Chr.14: 99502947-99502947 on Build GRCh38
rs777490086 Chr.14: 99503066-99503066 on Build GRCh38
rs145203611 Chr.14: 99503090-99503090 on Build GRCh38
rs776121018 Chr.14: 99502818-99502820 on Build GRCh38
rs778797351 Chr.14: 99503040-99503040 on Build GRCh38
rs935270794 Chr.14: 99502854-99502854 on Build GRCh38
rs765452231 Chr.14: 99502981-99502981 on Build GRCh38
rs909955801 Chr.14: 99502667-99502667 on Build GRCh38
rs1023229603 Chr.14: 99503041-99503041 on Build GRCh38
rs745738824 Chr.14: 99502801-99502801 on Build GRCh38
rs940083894 Chr.14: 99503025-99503025 on Build GRCh38
rs3918090 Chr.14: 99502831-99502831 on Build GRCh38
rs771726550 Chr.14: 99502965-99502965 on Build GRCh38
rs763535876 Chr.14: 99502678-99502678 on Build GRCh38
rs775810991 Chr.14: 99503092-99503092 on Build GRCh38
rs766722787 Chr.14: 99502702-99502702 on Build GRCh38
rs774074339 Chr.14: 99502826-99502826 on Build GRCh38
rs189345231 Chr.14: 99502683-99502683 on Build GRCh38
rs778725600 Chr.14: 99502946-99502946 on Build GRCh38
rs748221414 Chr.14: 99502747-99502747 on Build GRCh38
rs3918091 Chr.14: 99503021-99503021 on Build GRCh38
rs758477791 Chr.14: 99502892-99502892 on Build GRCh38
rs1035122250 Chr.14: 99502660-99502660 on Build GRCh38
rs201983471 Chr.14: 99503024-99503024 on Build GRCh38
rs755202901 Chr.14: 99502726-99502726 on Build GRCh38
rs376026947 Chr.14: 99502851-99502851 on Build GRCh38
rs750879696 Chr.14: 99502690-99502690 on Build GRCh38
rs772822750 Chr.14: 99502966-99502966 on Build GRCh38
rs79410788 Chr.14: 99502636-99502636 on Build GRCh38

View Full Product Details