Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102139646-102140136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSP90AA1
Entrez Gene ID
3320
Gene Name
heat shock protein 90 alpha family class A member 1
Gene Aliases
EL52, HEL-S-65p, HSP86, HSP89A, HSP90A, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP-2, LAP2
UniGene
Hs.525600
Gene Chromosome Location
Chr.14: 102080738-102139749 on Build GRCh38
Transcripts
NM_001017963.2 XM_011536718.2 NM_001242414.1 NM_001242415.1 NM_001242416.1 NM_001242417.1 NM_001242418.1 NM_001320130.1 NM_144574.3 NM_181291.2 NM_181308.2 NR_038354.1 XM_006720308.1 XM_006720309.1 XM_006720311.1 XM_006720313.2 XM_006720318.2 XM_006720320.3 XM_011537335.2 XM_011537336.2 XM_011537337.2 XM_011537338.1 XM_011537339.2 XM_011537340.2 XM_011537341.2 XM_011537343.1 XM_011537344.2 XM_011537345.1 XM_011537352.2 XM_011537353.2 XM_011537355.2 XM_011537356.2 XM_011537358.2 XM_017021766.1 XM_017021767.1 XM_017021768.1 XM_017021770.1 XM_017021771.1 XM_017021772.1 XM_017021773.1 XM_017021774.1 XM_017021775.1 XM_017021776.1 XM_017021777.1 XM_017021778.1 XM_017021779.1 XM_017021780.1 XM_017021781.1 XM_017021782.1 XM_017021783.1 XM_017021784.1 XM_017021785.1

Gene Symbol
WDR20
Entrez Gene ID
91833
Gene Name
WD repeat domain 20
Gene Aliases
DMR
UniGene
Hs.36859
Gene Chromosome Location
Chr.14: 102139518-102224686 on Build GRCh38
Transcripts
NM_001017963.2 XM_011536718.2 NM_001242414.1 NM_001242415.1 NM_001242416.1 NM_001242417.1 NM_001242418.1 NM_001320130.1 NM_144574.3 NM_181291.2 NM_181308.2 NR_038354.1 XM_006720308.1 XM_006720309.1 XM_006720311.1 XM_006720313.2 XM_006720318.2 XM_006720320.3 XM_011537335.2 XM_011537336.2 XM_011537337.2 XM_011537338.1 XM_011537339.2 XM_011537340.2 XM_011537341.2 XM_011537343.1 XM_011537344.2 XM_011537345.1 XM_011537352.2 XM_011537353.2 XM_011537355.2 XM_011537356.2 XM_011537358.2 XM_017021766.1 XM_017021767.1 XM_017021768.1 XM_017021770.1 XM_017021771.1 XM_017021772.1 XM_017021773.1 XM_017021774.1 XM_017021775.1 XM_017021776.1 XM_017021777.1 XM_017021778.1 XM_017021779.1 XM_017021780.1 XM_017021781.1 XM_017021782.1 XM_017021783.1 XM_017021784.1 XM_017021785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7096297 c.195C>T p.L65L Chr.14: 102140118-102140118 on Build GRCh38
7090849 c.195C>G p.L65L Chr.14: 102140118-102140118 on Build GRCh38
7096296 c.195C>T p.L65L Chr.14: 102140118-102140118 on Build GRCh38
697043 c.127C>A p.Q43K Chr.14: 102140050-102140050 on Build GRCh38
2024925 c.165C>G p.L55L Chr.14: 102140088-102140088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752133712 Chr.14: 102139967-102139967 on Build GRCh38
rs551423918 Chr.14: 102139856-102139856 on Build GRCh38
rs777183351 Chr.14: 102140112-102140112 on Build GRCh38
rs113602506 Chr.14: 102139687-102139687 on Build GRCh38
rs368209373 Chr.14: 102139990-102139990 on Build GRCh38
rs187774268 Chr.14: 102139771-102139771 on Build GRCh38
rs777252446 Chr.14: 102139971-102139971 on Build GRCh38
rs962232848 Chr.14: 102139920-102139920 on Build GRCh38
rs1025927516 Chr.14: 102139854-102139854 on Build GRCh38
rs754536997 Chr.14: 102139956-102139956 on Build GRCh38
rs374597485 Chr.14: 102139970-102139970 on Build GRCh38
rs191463145 Chr.14: 102139921-102139921 on Build GRCh38
rs374419515 Chr.14: 102140076-102140076 on Build GRCh38
rs866468682 Chr.14: 102139722-102139722 on Build GRCh38
rs765817316 Chr.14: 102140049-102140049 on Build GRCh38
rs149607593 Chr.14: 102140061-102140061 on Build GRCh38
rs889948345 Chr.14: 102139793-102139793 on Build GRCh38
rs750744397 Chr.14: 102139897-102139897 on Build GRCh38
rs976047833 Chr.14: 102140006-102140006 on Build GRCh38
rs945954203 Chr.14: 102139813-102139813 on Build GRCh38
rs773254962 Chr.14: 102140019-102140019 on Build GRCh38
rs762155932 Chr.14: 102139931-102139931 on Build GRCh38
rs1000055689 Chr.14: 102139847-102139847 on Build GRCh38
rs754336582 Chr.14: 102139951-102139951 on Build GRCh38
rs367961460 Chr.14: 102140113-102140113 on Build GRCh38
rs781026564 Chr.14: 102140075-102140075 on Build GRCh38
rs938659225 Chr.14: 102139705-102139705 on Build GRCh38
rs913917397 Chr.14: 102140042-102140042 on Build GRCh38
rs767010749 Chr.14: 102139946-102139946 on Build GRCh38
rs543182507 Chr.14: 102139747-102139747 on Build GRCh38
rs752291551 Chr.14: 102140052-102140052 on Build GRCh38
rs952141877 Chr.14: 102139680-102139681 on Build GRCh38
rs778631628 Chr.14: 102139959-102139959 on Build GRCh38
rs908993608 Chr.14: 102139863-102139863 on Build GRCh38
rs928553976 Chr.14: 102139696-102139696 on Build GRCh38
rs564942548 Chr.14: 102139859-102139859 on Build GRCh38
rs751723473 Chr.14: 102139876-102139876 on Build GRCh38
rs563998051 Chr.14: 102139666-102139666 on Build GRCh38
rs138455240 Chr.14: 102139749-102139749 on Build GRCh38
rs1053118269 Chr.14: 102139706-102139706 on Build GRCh38
rs757797886 Chr.14: 102140058-102140058 on Build GRCh38
rs773485048 Chr.14: 102140096-102140096 on Build GRCh38
rs200731683 Chr.14: 102140005-102140005 on Build GRCh38
rs757570761 Chr.14: 102139883-102139883 on Build GRCh38
rs766780589 Chr.14: 102140045-102140045 on Build GRCh38
rs928538251 Chr.14: 102139913-102139913 on Build GRCh38
rs371361662 Chr.14: 102139902-102139902 on Build GRCh38
rs1013870743 Chr.14: 102139909-102139909 on Build GRCh38
rs373018913 Chr.14: 102139901-102139901 on Build GRCh38
rs371409567 Chr.14: 102139910-102139910 on Build GRCh38
rs867800996 Chr.14: 102140063-102140063 on Build GRCh38
rs549221821 Chr.14: 102140079-102140079 on Build GRCh38
rs565846168 Chr.14: 102140088-102140088 on Build GRCh38
rs1043134652 Chr.14: 102140039-102140039 on Build GRCh38
rs972165511 Chr.14: 102139678-102139678 on Build GRCh38
rs376293169 Chr.14: 102139905-102139905 on Build GRCh38
rs535319759 Chr.14: 102140013-102140013 on Build GRCh38
rs534709063 Chr.14: 102140091-102140091 on Build GRCh38
rs771617250 Chr.14: 102139914-102139914 on Build GRCh38
rs754486063 Chr.14: 102139906-102139906 on Build GRCh38
rs760903759 Chr.14: 102139943-102139943 on Build GRCh38
rs769708239 Chr.14: 102140004-102140004 on Build GRCh38
rs966016417 Chr.14: 102139835-102139835 on Build GRCh38
rs984484749 Chr.14: 102139860-102139860 on Build GRCh38
rs771983268 Chr.14: 102140095-102140095 on Build GRCh38
rs1019840973 Chr.14: 102139832-102139832 on Build GRCh38
rs756889105 Chr.14: 102140053-102140053 on Build GRCh38
rs778719353 Chr.14: 102139907-102139907 on Build GRCh38
rs763820486 Chr.14: 102140065-102140065 on Build GRCh38
rs747298783 Chr.14: 102140109-102140109 on Build GRCh38
rs761112138 Chr.14: 102140033-102140033 on Build GRCh38
rs777681670 Chr.14: 102139915-102139915 on Build GRCh38
rs762981900 Chr.14: 102139874-102139874 on Build GRCh38
rs745875623 Chr.14: 102139999-102139999 on Build GRCh38
rs1007483945 Chr.14: 102139799-102139799 on Build GRCh38
rs944697852 Chr.14: 102139718-102139718 on Build GRCh38
rs138281148 Chr.14: 102139983-102139983 on Build GRCh38
rs962306062 Chr.14: 102139668-102139668 on Build GRCh38
rs746843473 Chr.14: 102139974-102139974 on Build GRCh38
rs752516665 Chr.14: 102140097-102140097 on Build GRCh38
rs762043809 Chr.14: 102139719-102139719 on Build GRCh38
rs767616468 Chr.14: 102139934-102139934 on Build GRCh38
rs1053066547 Chr.14: 102139960-102139960 on Build GRCh38
rs376176088 Chr.14: 102139738-102139738 on Build GRCh38
rs768656710 Chr.14: 102139929-102139929 on Build GRCh38
rs948582336 Chr.14: 102139684-102139684 on Build GRCh38
rs746756774 Chr.14: 102139919-102139919 on Build GRCh38
rs574183547 Chr.14: 102139735-102139735 on Build GRCh38
rs1023925483 Chr.14: 102139912-102139912 on Build GRCh38
rs750504705 Chr.14: 102139930-102139930 on Build GRCh38
rs750843842 Chr.14: 102139941-102139941 on Build GRCh38
rs770657800 Chr.14: 102139979-102139979 on Build GRCh38
rs1043188156 Chr.14: 102139781-102139781 on Build GRCh38
rs201411514 Chr.14: 102140070-102140070 on Build GRCh38
rs763907807 Chr.14: 102139702-102139702 on Build GRCh38
rs751108491 Chr.14: 102140069-102140069 on Build GRCh38
rs893656138 Chr.14: 102139839-102139839 on Build GRCh38
rs767996622 Chr.14: 102139885-102139885 on Build GRCh38
rs761770982 Chr.14: 102140010-102140010 on Build GRCh38
rs997103900 Chr.14: 102139840-102139840 on Build GRCh38
rs764323062 Chr.14: 102139875-102139875 on Build GRCh38
rs765243769 Chr.14: 102139928-102139928 on Build GRCh38
rs771264414 Chr.14: 102140110-102140110 on Build GRCh38

View Full Product Details