Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30908003-30908531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_001146094.1 NM_001146095.1 NM_001146096.1 NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_011521372.2 XM_017022012.1 XM_017022013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5675613 c.1303G>A p.E435K Chr.15: 30908186-30908186 on Build GRCh38
7264078 c.1294A>C p.T432P Chr.15: 30908177-30908177 on Build GRCh38
6445974 c.1352C>T p.T451M Chr.15: 30908235-30908235 on Build GRCh38
6521933 c.1362C>A p.G454G Chr.15: 30908245-30908245 on Build GRCh38
6445973 c.1352C>T p.T451M Chr.15: 30908235-30908235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755358716 Chr.15: 30908142-30908142 on Build GRCh38
rs368417113 Chr.15: 30908153-30908153 on Build GRCh38
rs374895181 Chr.15: 30908147-30908147 on Build GRCh38
rs748007364 Chr.15: 30908269-30908269 on Build GRCh38
rs909887254 Chr.15: 30908354-30908354 on Build GRCh38
rs138954416 Chr.15: 30908233-30908233 on Build GRCh38
rs745320524 Chr.15: 30908239-30908239 on Build GRCh38
rs543551458 Chr.15: 30908328-30908328 on Build GRCh38
rs776559468 Chr.15: 30908261-30908261 on Build GRCh38
rs149661264 Chr.15: 30908073-30908073 on Build GRCh38
rs888850195 Chr.15: 30908484-30908484 on Build GRCh38
rs768326389 Chr.15: 30908258-30908258 on Build GRCh38
rs375324676 Chr.15: 30908440-30908440 on Build GRCh38
rs35162264 Chr.15: 30908480-30908480 on Build GRCh38
rs984114432 Chr.15: 30908352-30908352 on Build GRCh38
rs75820658 Chr.15: 30908208-30908208 on Build GRCh38
rs753840240 Chr.15: 30908114-30908114 on Build GRCh38
rs967656496 Chr.15: 30908313-30908313 on Build GRCh38
rs771893997 Chr.15: 30908087-30908088 on Build GRCh38
rs771896426 Chr.15: 30908181-30908181 on Build GRCh38
rs138307818 Chr.15: 30908154-30908154 on Build GRCh38
rs777369804 Chr.15: 30908195-30908195 on Build GRCh38
rs751740552 Chr.15: 30908231-30908231 on Build GRCh38
rs79759675 Chr.15: 30908193-30908193 on Build GRCh38
rs755441131 Chr.15: 30908422-30908422 on Build GRCh38
rs751970854 Chr.15: 30908135-30908135 on Build GRCh38
rs776377863 Chr.15: 30908194-30908194 on Build GRCh38
rs1034339759 Chr.15: 30908462-30908462 on Build GRCh38
rs552320033 Chr.15: 30908432-30908432 on Build GRCh38
rs11633433 Chr.15: 30908307-30908307 on Build GRCh38
rs201748708 Chr.15: 30908078-30908078 on Build GRCh38
rs148577234 Chr.15: 30908399-30908399 on Build GRCh38
rs920983726 Chr.15: 30908357-30908357 on Build GRCh38
rs781484762 Chr.15: 30908143-30908143 on Build GRCh38
rs779949313 Chr.15: 30908361-30908361 on Build GRCh38
rs994812144 Chr.15: 30908450-30908450 on Build GRCh38
rs751811292 Chr.15: 30908228-30908228 on Build GRCh38
rs745484031 Chr.15: 30908174-30908174 on Build GRCh38
rs940887069 Chr.15: 30908404-30908404 on Build GRCh38
rs760501655 Chr.15: 30908192-30908192 on Build GRCh38
rs762981672 Chr.15: 30908278-30908278 on Build GRCh38
rs182455628 Chr.15: 30908235-30908235 on Build GRCh38
rs765230239 Chr.15: 30908212-30908212 on Build GRCh38
rs909154680 Chr.15: 30908047-30908047 on Build GRCh38
rs372026378 Chr.15: 30908338-30908338 on Build GRCh38
rs371629533 Chr.15: 30908276-30908276 on Build GRCh38
rs992695172 Chr.15: 30908053-30908053 on Build GRCh38
rs945974072 Chr.15: 30908060-30908060 on Build GRCh38
rs769821081 Chr.15: 30908275-30908275 on Build GRCh38
rs917127564 Chr.15: 30908055-30908055 on Build GRCh38
rs765375809 Chr.15: 30908120-30908120 on Build GRCh38
rs541343503 Chr.15: 30908244-30908244 on Build GRCh38
rs1021937419 Chr.15: 30908504-30908504 on Build GRCh38
rs760664761 Chr.15: 30908104-30908104 on Build GRCh38
rs1015629980 Chr.15: 30908220-30908220 on Build GRCh38
rs750633479 Chr.15: 30908124-30908124 on Build GRCh38
rs775301410 Chr.15: 30908188-30908188 on Build GRCh38
rs558967415 Chr.15: 30908215-30908215 on Build GRCh38
rs1031758745 Chr.15: 30908263-30908263 on Build GRCh38
rs781679930 Chr.15: 30908459-30908459 on Build GRCh38
rs200759965 Chr.15: 30908130-30908130 on Build GRCh38
rs144455048 Chr.15: 30908351-30908351 on Build GRCh38
rs771990319 Chr.15: 30908089-30908089 on Build GRCh38
rs761736097 Chr.15: 30908119-30908119 on Build GRCh38
rs1007251410 Chr.15: 30908163-30908163 on Build GRCh38
rs748701476 Chr.15: 30908151-30908151 on Build GRCh38
rs568070731 Chr.15: 30908046-30908046 on Build GRCh38
rs770479368 Chr.15: 30908070-30908070 on Build GRCh38
rs372815502 Chr.15: 30908165-30908165 on Build GRCh38
rs916024362 Chr.15: 30908335-30908335 on Build GRCh38
rs374747721 Chr.15: 30908294-30908294 on Build GRCh38
rs1051950452 Chr.15: 30908179-30908179 on Build GRCh38
rs368966606 Chr.15: 30908293-30908293 on Build GRCh38
rs919084163 Chr.15: 30908412-30908412 on Build GRCh38
rs530040866 Chr.15: 30908327-30908327 on Build GRCh38
rs758689259 Chr.15: 30908128-30908128 on Build GRCh38
rs1051311707 Chr.15: 30908373-30908373 on Build GRCh38
rs1014340379 Chr.15: 30908496-30908496 on Build GRCh38
rs763884861 Chr.15: 30908107-30908107 on Build GRCh38
rs761870607 Chr.15: 30908209-30908209 on Build GRCh38
rs888767298 Chr.15: 30908157-30908157 on Build GRCh38
rs775468946 Chr.15: 30908095-30908095 on Build GRCh38
rs200504684 Chr.15: 30908216-30908216 on Build GRCh38
rs755654276 Chr.15: 30908154-30908155 on Build GRCh38
rs746677293 Chr.15: 30908252-30908252 on Build GRCh38
rs142501642 Chr.15: 30908236-30908236 on Build GRCh38
rs766540040 Chr.15: 30908225-30908225 on Build GRCh38

View Full Product Details