Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55490647-55491184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYX1C1
Entrez Gene ID
161582
Gene Name
dyslexia susceptibility 1 candidate 1
Gene Aliases
CILD25, DNAAF4, DYX1, DYXC1, EKN1, RD
UniGene
Hs.126403
Gene Chromosome Location
Chr.15: 55417755-55508234 on Build GRCh38
Transcripts
NM_001033559.2 NM_001033560.1 NM_130810.3 NR_037923.1

Gene Symbol
DYX1C1-CCPG1
Entrez Gene ID
100533483
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.285051
Gene Chromosome Location
Chr.15: 55355223-55498584 on Build GRCh38
Transcripts
NM_001033559.2 NM_001033560.1 NM_130810.3 NR_037923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7310396 c.370G>T p.E124* Chr.15: 55491158-55491158 on Build GRCh38
7189973 c.400A>T p.M134L Chr.15: 55491128-55491128 on Build GRCh38
7424681 c.360A>T p.A120A Chr.15: 55491168-55491168 on Build GRCh38
4920840 c.345A>T p.T115T Chr.15: 55491183-55491183 on Build GRCh38
7310395 c.370G>T p.E124* Chr.15: 55491158-55491158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531034378 Chr.15: 55490952-55490952 on Build GRCh38
rs146913170 Chr.15: 55490675-55490675 on Build GRCh38
rs960288016 Chr.15: 55490847-55490847 on Build GRCh38
rs750345738 Chr.15: 55490700-55490700 on Build GRCh38
rs574218105 Chr.15: 55490789-55490789 on Build GRCh38
rs573994909 Chr.15: 55490862-55490862 on Build GRCh38
rs996360479 Chr.15: 55490943-55490943 on Build GRCh38
rs1057455849 Chr.15: 55490821-55490821 on Build GRCh38
rs944647296 Chr.15: 55491122-55491122 on Build GRCh38
rs1035708352 Chr.15: 55490989-55490989 on Build GRCh38
rs192844280 Chr.15: 55491055-55491055 on Build GRCh38
rs940631014 Chr.15: 55490945-55490945 on Build GRCh38
rs769744140 Chr.15: 55491106-55491106 on Build GRCh38
rs761697978 Chr.15: 55491098-55491098 on Build GRCh38
rs1044224442 Chr.15: 55490878-55490878 on Build GRCh38
rs533401735 Chr.15: 55490981-55490981 on Build GRCh38
rs762693941 Chr.15: 55491113-55491113 on Build GRCh38
rs150525906 Chr.15: 55491132-55491132 on Build GRCh38
rs796784326 Chr.15: 55490771-55490771 on Build GRCh38
rs995597175 Chr.15: 55490966-55490967 on Build GRCh38
rs749092579 Chr.15: 55491161-55491161 on Build GRCh38
rs1051666557 Chr.15: 55490745-55490745 on Build GRCh38
rs62019995 Chr.15: 55490879-55490879 on Build GRCh38
rs917359683 Chr.15: 55490858-55490858 on Build GRCh38
rs969325265 Chr.15: 55491064-55491064 on Build GRCh38
rs968879467 Chr.15: 55490963-55490963 on Build GRCh38
rs57914590 Chr.15: 55491144-55491144 on Build GRCh38
rs143462903 Chr.15: 55490884-55490884 on Build GRCh38
rs921458935 Chr.15: 55490760-55490760 on Build GRCh38
rs980596143 Chr.15: 55491000-55491000 on Build GRCh38
rs769685504 Chr.15: 55491160-55491160 on Build GRCh38
rs776516127 Chr.15: 55491086-55491086 on Build GRCh38
rs534539144 Chr.15: 55490749-55490749 on Build GRCh38
rs917267659 Chr.15: 55490993-55490993 on Build GRCh38
rs756017665 Chr.15: 55490803-55490803 on Build GRCh38
rs368804132 Chr.15: 55491140-55491140 on Build GRCh38
rs1036181161 Chr.15: 55490957-55490957 on Build GRCh38
rs567876245 Chr.15: 55491069-55491069 on Build GRCh38
rs576727789 Chr.15: 55490885-55490885 on Build GRCh38
rs554324913 Chr.15: 55490755-55490755 on Build GRCh38
rs767771520 Chr.15: 55491114-55491114 on Build GRCh38
rs192071791 Chr.15: 55490799-55490799 on Build GRCh38
rs565668410 Chr.15: 55491046-55491046 on Build GRCh38
rs900207517 Chr.15: 55491137-55491137 on Build GRCh38
rs765886556 Chr.15: 55491141-55491141 on Build GRCh38
rs920393904 Chr.15: 55491052-55491052 on Build GRCh38
rs1025933827 Chr.15: 55490694-55490694 on Build GRCh38
rs137881700 Chr.15: 55490723-55490723 on Build GRCh38
rs934657887 Chr.15: 55490809-55490809 on Build GRCh38
rs950389552 Chr.15: 55490719-55490719 on Build GRCh38
rs1027525011 Chr.15: 55490967-55490967 on Build GRCh38
rs1015185207 Chr.15: 55490767-55490767 on Build GRCh38
rs538558567 Chr.15: 55490682-55490682 on Build GRCh38
rs1047644696 Chr.15: 55490902-55490902 on Build GRCh38
rs890284161 Chr.15: 55490754-55490754 on Build GRCh38
rs576855321 Chr.15: 55491115-55491115 on Build GRCh38
rs747703690 Chr.15: 55491156-55491156 on Build GRCh38
rs886077202 Chr.15: 55490905-55490905 on Build GRCh38
rs996020438 Chr.15: 55490777-55490777 on Build GRCh38
rs978976258 Chr.15: 55490966-55490966 on Build GRCh38
rs994746268 Chr.15: 55490691-55490691 on Build GRCh38
rs879214268 Chr.15: 55490724-55490724 on Build GRCh38
rs947136787 Chr.15: 55490883-55490883 on Build GRCh38
rs987074610 Chr.15: 55491134-55491134 on Build GRCh38
rs542716024 Chr.15: 55490907-55490907 on Build GRCh38
rs899064531 Chr.15: 55490681-55490681 on Build GRCh38
rs750814401 Chr.15: 55491143-55491143 on Build GRCh38
rs565824651 Chr.15: 55490741-55490741 on Build GRCh38
rs367828329 Chr.15: 55490911-55490911 on Build GRCh38
rs899023231 Chr.15: 55490961-55490961 on Build GRCh38
rs975587566 Chr.15: 55490736-55490736 on Build GRCh38
rs534579615 Chr.15: 55491048-55491048 on Build GRCh38
rs183465816 Chr.15: 55490912-55490912 on Build GRCh38
rs7180984 Chr.15: 55490975-55490975 on Build GRCh38
rs74792041 Chr.15: 55490972-55490972 on Build GRCh38
rs1035906271 Chr.15: 55490833-55490833 on Build GRCh38
rs965903456 Chr.15: 55490725-55490725 on Build GRCh38
rs1004633274 Chr.15: 55490761-55490761 on Build GRCh38
rs73411032 Chr.15: 55490996-55490996 on Build GRCh38
rs931740024 Chr.15: 55491097-55491097 on Build GRCh38
rs746516117 Chr.15: 55491077-55491077 on Build GRCh38
rs545480000 Chr.15: 55490896-55490896 on Build GRCh38
rs948841068 Chr.15: 55490859-55490859 on Build GRCh38
rs780630130 Chr.15: 55491152-55491152 on Build GRCh38
rs188412550 Chr.15: 55490932-55490932 on Build GRCh38
rs374404403 Chr.15: 55491073-55491073 on Build GRCh38
rs900187853 Chr.15: 55490733-55490733 on Build GRCh38
rs536752081 Chr.15: 55491090-55491090 on Build GRCh38
rs781156334 Chr.15: 55491135-55491138 on Build GRCh38
rs1028490639 Chr.15: 55490947-55490947 on Build GRCh38
rs553854743 Chr.15: 55490853-55490853 on Build GRCh38
rs557066254 Chr.15: 55491111-55491111 on Build GRCh38
rs776121861 Chr.15: 55491094-55491094 on Build GRCh38
rs996036342 Chr.15: 55490737-55490737 on Build GRCh38
rs747493543 Chr.15: 55491126-55491126 on Build GRCh38
rs74830467 Chr.15: 55490957-55490957 on Build GRCh38

View Full Product Details