Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67254437-67254932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1 NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 NM_022784.2 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521915.2 XM_011521918.2 XM_011521922.2 XM_011521923.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
UniGene
Hs.657894
Gene Chromosome Location
Chr.15: 67254772-67501804 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1 NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 NM_022784.2 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521915.2 XM_011521918.2 XM_011521922.2 XM_011521923.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645902 c.60C>T p.D20D Chr.15: 67254572-67254572 on Build GRCh38
5363084 c.15T>C p.T5T Chr.15: 67254911-67254911 on Build GRCh38
5636864 c.45C>A p.S15S Chr.15: 67254587-67254587 on Build GRCh38
2257451 c.7C>T p.Q3* Chr.15: 67254903-67254903 on Build GRCh38
2257450 c.7C>T p.Q3* Chr.15: 67254903-67254903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937732358 Chr.15: 67254881-67254881 on Build GRCh38
rs34746914 Chr.15: 67254733-67254734 on Build GRCh38
rs749693035 Chr.15: 67254630-67254630 on Build GRCh38
rs370216523 Chr.15: 67254872-67254872 on Build GRCh38
rs990427540 Chr.15: 67254626-67254626 on Build GRCh38
rs1011747179 Chr.15: 67254801-67254801 on Build GRCh38
rs527525799 Chr.15: 67254597-67254597 on Build GRCh38
rs1034900836 Chr.15: 67254760-67254760 on Build GRCh38
rs756813133 Chr.15: 67254616-67254616 on Build GRCh38
rs907902517 Chr.15: 67254662-67254662 on Build GRCh38
rs376484021 Chr.15: 67254878-67254878 on Build GRCh38
rs1001985975 Chr.15: 67254749-67254749 on Build GRCh38
rs939941137 Chr.15: 67254875-67254875 on Build GRCh38
rs563959836 Chr.15: 67254842-67254842 on Build GRCh38
rs921056710 Chr.15: 67254839-67254839 on Build GRCh38
rs371342150 Chr.15: 67254615-67254615 on Build GRCh38
rs765012111 Chr.15: 67254847-67254847 on Build GRCh38
rs113546620 Chr.15: 67254647-67254647 on Build GRCh38
rs912358276 Chr.15: 67254491-67254491 on Build GRCh38
rs959025113 Chr.15: 67254460-67254460 on Build GRCh38
rs758655425 Chr.15: 67254871-67254871 on Build GRCh38
rs373466809 Chr.15: 67254654-67254654 on Build GRCh38
rs369049544 Chr.15: 67254624-67254624 on Build GRCh38
rs750789718 Chr.15: 67254904-67254904 on Build GRCh38
rs757709854 Chr.15: 67254867-67254867 on Build GRCh38
rs894664153 Chr.15: 67254761-67254761 on Build GRCh38
rs751793473 Chr.15: 67254656-67254657 on Build GRCh38
rs903428420 Chr.15: 67254901-67254901 on Build GRCh38
rs767540102 Chr.15: 67254605-67254605 on Build GRCh38
rs766048528 Chr.15: 67254851-67254851 on Build GRCh38
rs564840614 Chr.15: 67254471-67254471 on Build GRCh38
rs754435742 Chr.15: 67254863-67254863 on Build GRCh38
rs572038348 Chr.15: 67254734-67254734 on Build GRCh38
rs578254920 Chr.15: 67254807-67254807 on Build GRCh38
rs559840295 Chr.15: 67254900-67254900 on Build GRCh38
rs752166368 Chr.15: 67254897-67254897 on Build GRCh38
rs761539733 Chr.15: 67254778-67254778 on Build GRCh38
rs939658757 Chr.15: 67254715-67254715 on Build GRCh38
rs751036390 Chr.15: 67254853-67254853 on Build GRCh38
rs931200015 Chr.15: 67254658-67254658 on Build GRCh38
rs776686661 Chr.15: 67254675-67254675 on Build GRCh38
rs374064896 Chr.15: 67254581-67254581 on Build GRCh38
rs977390382 Chr.15: 67254821-67254821 on Build GRCh38
rs180799838 Chr.15: 67254623-67254623 on Build GRCh38
rs889712018 Chr.15: 67254762-67254762 on Build GRCh38
rs533628412 Chr.15: 67254481-67254481 on Build GRCh38
rs990535236 Chr.15: 67254470-67254470 on Build GRCh38
rs368197105 Chr.15: 67254591-67254591 on Build GRCh38
rs1040506379 Chr.15: 67254696-67254696 on Build GRCh38
rs764498221 Chr.15: 67254856-67254856 on Build GRCh38
rs752740271 Chr.15: 67254608-67254608 on Build GRCh38
rs747120630 Chr.15: 67254666-67254666 on Build GRCh38
rs925738103 Chr.15: 67254486-67254486 on Build GRCh38
rs778231997 Chr.15: 67254551-67254551 on Build GRCh38
rs756022692 Chr.15: 67254535-67254535 on Build GRCh38
rs539301011 Chr.15: 67254631-67254631 on Build GRCh38
rs771204932 Chr.15: 67254600-67254600 on Build GRCh38
rs764780013 Chr.15: 67254642-67254642 on Build GRCh38
rs746262191 Chr.15: 67254869-67254869 on Build GRCh38
rs867166731 Chr.15: 67254835-67254835 on Build GRCh38
rs766409174 Chr.15: 67254526-67254527 on Build GRCh38
rs746039568 Chr.15: 67254659-67254659 on Build GRCh38
rs772440973 Chr.15: 67254660-67254660 on Build GRCh38
rs955947267 Chr.15: 67254505-67254505 on Build GRCh38
rs1041804504 Chr.15: 67254485-67254485 on Build GRCh38
rs374516781 Chr.15: 67254614-67254614 on Build GRCh38
rs1028231476 Chr.15: 67254475-67254475 on Build GRCh38
rs202246610 Chr.15: 67254492-67254493 on Build GRCh38
rs201942506 Chr.15: 67254638-67254638 on Build GRCh38
rs541621490 Chr.15: 67254758-67254758 on Build GRCh38
rs547264525 Chr.15: 67254613-67254613 on Build GRCh38
rs374270407 Chr.15: 67254635-67254635 on Build GRCh38
rs188441125 Chr.15: 67254590-67254590 on Build GRCh38
rs558335276 Chr.15: 67254798-67254798 on Build GRCh38
rs1003448430 Chr.15: 67254622-67254622 on Build GRCh38
rs762543048 Chr.15: 67254850-67254850 on Build GRCh38
rs370626700 Chr.15: 67254572-67254572 on Build GRCh38
rs369542537 Chr.15: 67254628-67254628 on Build GRCh38
rs113435495 Chr.15: 67254857-67254857 on Build GRCh38
rs775665288 Chr.15: 67254664-67254664 on Build GRCh38
rs891483667 Chr.15: 67254803-67254803 on Build GRCh38
rs975537183 Chr.15: 67254834-67254834 on Build GRCh38
rs750152610 Chr.15: 67254643-67254643 on Build GRCh38
rs1046134763 Chr.15: 67254724-67254724 on Build GRCh38
rs774257097 Chr.15: 67254604-67254604 on Build GRCh38
rs752444584 Chr.15: 67254868-67254868 on Build GRCh38
rs762775024 Chr.15: 67254885-67254885 on Build GRCh38
rs996920871 Chr.15: 67254752-67254752 on Build GRCh38
rs879768611 Chr.15: 67254582-67254582 on Build GRCh38
rs1039574865 Chr.15: 67254502-67254502 on Build GRCh38
rs901904492 Chr.15: 67254703-67254703 on Build GRCh38
rs763918850 Chr.15: 67254611-67254611 on Build GRCh38
rs35281350 Chr.15: 67254652-67254653 on Build GRCh38
rs776575211 Chr.15: 67254880-67254880 on Build GRCh38
rs529424803 Chr.15: 67254536-67254536 on Build GRCh38
rs774684811 Chr.15: 67254632-67254632 on Build GRCh38
rs759522737 Chr.15: 67254633-67254633 on Build GRCh38
rs1000559378 Chr.15: 67254789-67254789 on Build GRCh38
rs150409242 Chr.15: 67254888-67254888 on Build GRCh38
rs886739089 Chr.15: 67254705-67254705 on Build GRCh38
rs886791512 Chr.15: 67254612-67254612 on Build GRCh38
rs544092628 Chr.15: 67254831-67254831 on Build GRCh38
rs758889010 Chr.15: 67254907-67254907 on Build GRCh38
rs777168127 Chr.15: 67254587-67254587 on Build GRCh38
rs773018358 Chr.15: 67254884-67254884 on Build GRCh38
rs1019636309 Chr.15: 67254829-67254829 on Build GRCh38
rs745408900 Chr.15: 67254556-67254556 on Build GRCh38
rs756942730 Chr.15: 67254545-67254545 on Build GRCh38
rs753539938 Chr.15: 67254637-67254637 on Build GRCh38
rs371940692 Chr.15: 67254625-67254625 on Build GRCh38
rs376857654 Chr.15: 67254627-67254627 on Build GRCh38
rs749596536 Chr.15: 67254650-67254650 on Build GRCh38
rs927515697 Chr.15: 67254602-67254602 on Build GRCh38
rs967639353 Chr.15: 67254815-67254815 on Build GRCh38
rs1027238783 Chr.15: 67254858-67254858 on Build GRCh38
rs115599985 Chr.15: 67254641-67254641 on Build GRCh38
rs555486899 Chr.15: 67254667-67254667 on Build GRCh38
rs761504906 Chr.15: 67254846-67254846 on Build GRCh38
rs780417908 Chr.15: 67254909-67254909 on Build GRCh38
rs971089843 Chr.15: 67254832-67254832 on Build GRCh38
rs779000422 Chr.15: 67254655-67254655 on Build GRCh38
rs931169918 Chr.15: 67254840-67254840 on Build GRCh38
rs778665736 Chr.15: 67254619-67254619 on Build GRCh38
rs556663683 Chr.15: 67254596-67254596 on Build GRCh38
rs757738757 Chr.15: 67254898-67254898 on Build GRCh38
rs529318414 Chr.15: 67254883-67254883 on Build GRCh38
rs766986629 Chr.15: 67254890-67254890 on Build GRCh38
rs778385934 Chr.15: 67254649-67254649 on Build GRCh38
rs932015152 Chr.15: 67254891-67254891 on Build GRCh38
rs563647824 Chr.15: 67254528-67254528 on Build GRCh38
rs749019807 Chr.15: 67254540-67254540 on Build GRCh38
rs371952028 Chr.15: 67254609-67254609 on Build GRCh38
rs899746075 Chr.15: 67254504-67254504 on Build GRCh38
rs543843488 Chr.15: 67254498-67254498 on Build GRCh38

View Full Product Details