Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67357256-67357755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
UniGene
Hs.657894
Gene Chromosome Location
Chr.15: 67254772-67501804 on Build GRCh38
Transcripts
NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521914.2 XM_011521915.2 XM_011521916.2 XM_011521917.2 XM_011521918.2 XM_011521922.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338966 c.653C>T p.P218L Chr.15: 67357360-67357360 on Build GRCh38
7181047 c.645C>T p.F215F Chr.15: 67357352-67357352 on Build GRCh38
3503374 c.655C>T p.R219W Chr.15: 67357362-67357362 on Build GRCh38
5033934 c.674C>T p.P225L Chr.15: 67357381-67357381 on Build GRCh38
5849608 c.638-1G>A p.? Chr.15: 67357344-67357344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779906669 Chr.15: 67357362-67357362 on Build GRCh38
rs368941939 Chr.15: 67357419-67357419 on Build GRCh38
rs926812157 Chr.15: 67357349-67357349 on Build GRCh38
rs776270707 Chr.15: 67357388-67357388 on Build GRCh38
rs958407914 Chr.15: 67357604-67357604 on Build GRCh38
rs781445526 Chr.15: 67357630-67357630 on Build GRCh38
rs911855547 Chr.15: 67357686-67357686 on Build GRCh38
rs775293136 Chr.15: 67357352-67357353 on Build GRCh38
rs1045914808 Chr.15: 67357589-67357589 on Build GRCh38
rs950325560 Chr.15: 67357300-67357300 on Build GRCh38
rs1032779542 Chr.15: 67357591-67357591 on Build GRCh38
rs989641996 Chr.15: 67357622-67357622 on Build GRCh38
rs771781551 Chr.15: 67357303-67357305 on Build GRCh38
rs750204010 Chr.15: 67357440-67357440 on Build GRCh38
rs936867932 Chr.15: 67357355-67357355 on Build GRCh38
rs570694882 Chr.15: 67357315-67357315 on Build GRCh38
rs1036350180 Chr.15: 67357688-67357688 on Build GRCh38
rs766593408 Chr.15: 67357297-67357297 on Build GRCh38
rs778526529 Chr.15: 67357345-67357345 on Build GRCh38
rs548671466 Chr.15: 67357583-67357583 on Build GRCh38
rs937614290 Chr.15: 67357596-67357596 on Build GRCh38
rs761693040 Chr.15: 67357438-67357438 on Build GRCh38
rs780890510 Chr.15: 67357467-67357467 on Build GRCh38
rs879880302 Chr.15: 67357522-67357522 on Build GRCh38
rs751188590 Chr.15: 67357584-67357584 on Build GRCh38
rs1029505865 Chr.15: 67357728-67357728 on Build GRCh38
rs747655064 Chr.15: 67357468-67357468 on Build GRCh38
rs558317058 Chr.15: 67357721-67357721 on Build GRCh38
rs771363943 Chr.15: 67357472-67357472 on Build GRCh38
rs775622696 Chr.15: 67357431-67357431 on Build GRCh38
rs764214714 Chr.15: 67357434-67357434 on Build GRCh38
rs928766128 Chr.15: 67357593-67357593 on Build GRCh38
rs760495284 Chr.15: 67357455-67357457 on Build GRCh38
rs869304 Chr.15: 67357640-67357640 on Build GRCh38
rs1029494435 Chr.15: 67357476-67357476 on Build GRCh38
rs765177601 Chr.15: 67357439-67357439 on Build GRCh38
rs1014992532 Chr.15: 67357414-67357414 on Build GRCh38
rs756245638 Chr.15: 67357336-67357336 on Build GRCh38
rs761777679 Chr.15: 67357453-67357453 on Build GRCh38
rs576776467 Chr.15: 67357478-67357478 on Build GRCh38
rs377534345 Chr.15: 67357371-67357371 on Build GRCh38
rs910898346 Chr.15: 67357516-67357516 on Build GRCh38
rs759895938 Chr.15: 67357424-67357424 on Build GRCh38
rs186371447 Chr.15: 67357582-67357582 on Build GRCh38
rs1026200295 Chr.15: 67357521-67357521 on Build GRCh38
rs966957125 Chr.15: 67357634-67357634 on Build GRCh38
rs745710910 Chr.15: 67357348-67357348 on Build GRCh38
rs375240983 Chr.15: 67357498-67357498 on Build GRCh38
rs776509221 Chr.15: 67357356-67357356 on Build GRCh38
rs1024052033 Chr.15: 67357307-67357307 on Build GRCh38
rs7174933 Chr.15: 67357581-67357581 on Build GRCh38
rs772330465 Chr.15: 67357298-67357298 on Build GRCh38
rs772295121 Chr.15: 67357430-67357430 on Build GRCh38
rs34980358 Chr.15: 67357569-67357569 on Build GRCh38
rs914154613 Chr.15: 67357633-67357633 on Build GRCh38
rs771439713 Chr.15: 67357417-67357417 on Build GRCh38
rs374725274 Chr.15: 67357415-67357415 on Build GRCh38
rs755325327 Chr.15: 67357605-67357605 on Build GRCh38
rs368768816 Chr.15: 67357304-67357304 on Build GRCh38
rs202039610 Chr.15: 67357316-67357316 on Build GRCh38
rs117045074 Chr.15: 67357499-67357499 on Build GRCh38
rs746526010 Chr.15: 67357367-67357367 on Build GRCh38
rs758149730 Chr.15: 67357441-67357441 on Build GRCh38
rs779994497 Chr.15: 67357451-67357451 on Build GRCh38
rs757235384 Chr.15: 67357341-67357341 on Build GRCh38
rs764016018 Chr.15: 67357337-67357337 on Build GRCh38
rs534949146 Chr.15: 67357704-67357704 on Build GRCh38
rs764702633 Chr.15: 67357483-67357483 on Build GRCh38
rs112896105 Chr.15: 67357700-67357700 on Build GRCh38
rs1013923961 Chr.15: 67357557-67357557 on Build GRCh38
rs768351709 Chr.15: 67357368-67357368 on Build GRCh38
rs749989731 Chr.15: 67357575-67357575 on Build GRCh38
rs753710656 Chr.15: 67357338-67357338 on Build GRCh38
rs970248963 Chr.15: 67357308-67357308 on Build GRCh38
rs945372739 Chr.15: 67357363-67357363 on Build GRCh38
rs751317651 Chr.15: 67357452-67357452 on Build GRCh38
rs943404070 Chr.15: 67357694-67357694 on Build GRCh38

View Full Product Details