Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30273915-30274428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLMH
Entrez Gene ID
642
Gene Name
bleomycin hydrolase
Gene Aliases
BH, BMH
UniGene
Hs.371914
Gene Chromosome Location
Chr.17: 30248195-30292166 on Build GRCh38
Transcripts
NM_000386.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357562 c.722A>T p.N241I Chr.17: 30274121-30274121 on Build GRCh38
1710076 c.679C>T p.P227S Chr.17: 30274164-30274164 on Build GRCh38
4065258 c.652C>T p.R218* Chr.17: 30274191-30274191 on Build GRCh38
7095061 c.693C>T p.F231F Chr.17: 30274150-30274150 on Build GRCh38
6246223 c.651C>A p.F217L Chr.17: 30274192-30274192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958572482 Chr.17: 30274376-30274376 on Build GRCh38
rs1020268511 Chr.17: 30274350-30274350 on Build GRCh38
rs1018747929 Chr.17: 30273958-30273958 on Build GRCh38
rs751486628 Chr.17: 30273988-30273988 on Build GRCh38
rs758033039 Chr.17: 30274033-30274033 on Build GRCh38
rs749434555 Chr.17: 30274038-30274038 on Build GRCh38
rs991808532 Chr.17: 30274257-30274257 on Build GRCh38
rs747421308 Chr.17: 30274211-30274211 on Build GRCh38
rs774979440 Chr.17: 30274096-30274096 on Build GRCh38
rs757189424 Chr.17: 30274121-30274121 on Build GRCh38
rs903171409 Chr.17: 30274358-30274358 on Build GRCh38
rs755178186 Chr.17: 30274191-30274191 on Build GRCh38
rs761108875 Chr.17: 30274070-30274070 on Build GRCh38
rs942830441 Chr.17: 30274297-30274297 on Build GRCh38
rs868030761 Chr.17: 30274150-30274150 on Build GRCh38
rs1035985778 Chr.17: 30274361-30274361 on Build GRCh38
rs891308485 Chr.17: 30274277-30274277 on Build GRCh38
rs371345449 Chr.17: 30274222-30274222 on Build GRCh38
rs137855289 Chr.17: 30274063-30274063 on Build GRCh38
rs778865567 Chr.17: 30274197-30274197 on Build GRCh38
rs749502447 Chr.17: 30274190-30274190 on Build GRCh38
rs756390997 Chr.17: 30274012-30274012 on Build GRCh38
rs373475025 Chr.17: 30274100-30274100 on Build GRCh38
rs908920732 Chr.17: 30274143-30274143 on Build GRCh38
rs745591487 Chr.17: 30273995-30273995 on Build GRCh38
rs763160875 Chr.17: 30274017-30274018 on Build GRCh38
rs146981308 Chr.17: 30274010-30274011 on Build GRCh38
rs1011611297 Chr.17: 30274336-30274336 on Build GRCh38
rs537954134 Chr.17: 30274387-30274387 on Build GRCh38
rs557263621 Chr.17: 30274394-30274394 on Build GRCh38
rs761297330 Chr.17: 30274238-30274238 on Build GRCh38
rs991267016 Chr.17: 30274128-30274128 on Build GRCh38
rs780363919 Chr.17: 30274163-30274163 on Build GRCh38
rs763537830 Chr.17: 30274112-30274112 on Build GRCh38
rs958651972 Chr.17: 30274240-30274240 on Build GRCh38
rs377358564 Chr.17: 30274109-30274109 on Build GRCh38
rs781276975 Chr.17: 30274341-30274341 on Build GRCh38
rs780975834 Chr.17: 30273994-30273994 on Build GRCh38
rs779473617 Chr.17: 30274209-30274209 on Build GRCh38
rs377543913 Chr.17: 30274148-30274148 on Build GRCh38
rs370227006 Chr.17: 30274113-30274113 on Build GRCh38
rs865924244 Chr.17: 30274094-30274094 on Build GRCh38
rs149455631 Chr.17: 30274227-30274227 on Build GRCh38
rs373400008 Chr.17: 30274136-30274136 on Build GRCh38
rs1020242316 Chr.17: 30273986-30273986 on Build GRCh38
rs1039795285 Chr.17: 30274300-30274300 on Build GRCh38
rs752358021 Chr.17: 30274230-30274230 on Build GRCh38
rs748234744 Chr.17: 30274201-30274201 on Build GRCh38
rs370640347 Chr.17: 30274204-30274204 on Build GRCh38
rs764993456 Chr.17: 30274089-30274089 on Build GRCh38
rs375546556 Chr.17: 30274030-30274030 on Build GRCh38
rs369965286 Chr.17: 30274062-30274062 on Build GRCh38
rs16965663 Chr.17: 30273987-30273987 on Build GRCh38
rs112288201 Chr.17: 30273991-30273991 on Build GRCh38
rs375009285 Chr.17: 30274221-30274221 on Build GRCh38
rs868352189 Chr.17: 30274154-30274154 on Build GRCh38
rs766457116 Chr.17: 30274233-30274233 on Build GRCh38
rs552170159 Chr.17: 30274305-30274305 on Build GRCh38
rs755880091 Chr.17: 30274160-30274160 on Build GRCh38
rs150484910 Chr.17: 30273959-30273959 on Build GRCh38
rs181868675 Chr.17: 30274359-30274359 on Build GRCh38
rs768284725 Chr.17: 30274229-30274229 on Build GRCh38
rs570290540 Chr.17: 30274206-30274206 on Build GRCh38
rs768585859 Chr.17: 30274046-30274046 on Build GRCh38
rs200660785 Chr.17: 30274069-30274069 on Build GRCh38
rs974205870 Chr.17: 30274389-30274389 on Build GRCh38
rs1011495117 Chr.17: 30273984-30273984 on Build GRCh38

View Full Product Details