Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18246216-18246711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLII
Entrez Gene ID
2314
Gene Name
FLII, actin remodeling protein
Gene Aliases
FLI, FLIL, Fli1
UniGene
Hs.513984
Gene Chromosome Location
Chr.17: 18244815-18258916 on Build GRCh38
Transcripts
NM_001256264.1 NM_001256265.1 NM_002018.3 XM_005256555.3 XM_005256556.3 XM_005256558.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664646 c.2949C>T p.I983I Chr.17: 18246696-18246696 on Build GRCh38
3889392 c.3003C>T p.T1001T Chr.17: 18246642-18246642 on Build GRCh38
2157704 c.3163_3164insC p.S1055fs*39 Chr.17: 18246350-18246351 on Build GRCh38
6664643 c.3057A>G p.V1019V Chr.17: 18246457-18246457 on Build GRCh38
6664649 c.3163A>G p.S1055G Chr.17: 18246351-18246351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867528265 Chr.17: 18246374-18246374 on Build GRCh38
rs376758689 Chr.17: 18246350-18246350 on Build GRCh38
rs879568374 Chr.17: 18246624-18246626 on Build GRCh38
rs551167459 Chr.17: 18246600-18246600 on Build GRCh38
rs1050547923 Chr.17: 18246351-18246352 on Build GRCh38
rs74508005 Chr.17: 18246632-18246632 on Build GRCh38
rs761250060 Chr.17: 18246547-18246547 on Build GRCh38
rs778026726 Chr.17: 18246305-18246305 on Build GRCh38
rs761454237 Chr.17: 18246442-18246442 on Build GRCh38
rs769194407 Chr.17: 18246411-18246411 on Build GRCh38
rs764698657 Chr.17: 18246363-18246363 on Build GRCh38
rs140455902 Chr.17: 18246452-18246452 on Build GRCh38
rs368988400 Chr.17: 18246276-18246276 on Build GRCh38
rs762159677 Chr.17: 18246561-18246561 on Build GRCh38
rs764546674 Chr.17: 18246467-18246467 on Build GRCh38
rs1000537794 Chr.17: 18246515-18246515 on Build GRCh38
rs766717243 Chr.17: 18246567-18246567 on Build GRCh38
rs751594817 Chr.17: 18246686-18246686 on Build GRCh38
rs374093119 Chr.17: 18246581-18246581 on Build GRCh38
rs1031472056 Chr.17: 18246517-18246517 on Build GRCh38
rs758504048 Chr.17: 18246394-18246396 on Build GRCh38
rs1048370552 Chr.17: 18246256-18246256 on Build GRCh38
rs770947928 Chr.17: 18246245-18246245 on Build GRCh38
rs377505637 Chr.17: 18246449-18246449 on Build GRCh38
rs769392349 Chr.17: 18246498-18246498 on Build GRCh38
rs766370090 Chr.17: 18246572-18246572 on Build GRCh38
rs776130105 Chr.17: 18246514-18246514 on Build GRCh38
rs757477726 Chr.17: 18246389-18246389 on Build GRCh38
rs564079069 Chr.17: 18246424-18246424 on Build GRCh38
rs539123081 Chr.17: 18246283-18246283 on Build GRCh38
rs780955337 Chr.17: 18246491-18246491 on Build GRCh38
rs760026918 Chr.17: 18246365-18246365 on Build GRCh38
rs375498127 Chr.17: 18246336-18246336 on Build GRCh38
rs78659863 Chr.17: 18246533-18246533 on Build GRCh38
rs189440439 Chr.17: 18246400-18246400 on Build GRCh38
rs373738329 Chr.17: 18246348-18246348 on Build GRCh38
rs371073180 Chr.17: 18246557-18246557 on Build GRCh38
rs552950931 Chr.17: 18246285-18246285 on Build GRCh38
rs771111506 Chr.17: 18246319-18246319 on Build GRCh38
rs776308524 Chr.17: 18246439-18246439 on Build GRCh38
rs769246541 Chr.17: 18246662-18246662 on Build GRCh38
rs760364052 Chr.17: 18246274-18246274 on Build GRCh38
rs372936022 Chr.17: 18246448-18246448 on Build GRCh38
rs754081390 Chr.17: 18246382-18246382 on Build GRCh38
rs370427382 Chr.17: 18246468-18246468 on Build GRCh38
rs753121182 Chr.17: 18246458-18246458 on Build GRCh38
rs908049138 Chr.17: 18246436-18246436 on Build GRCh38
rs201437083 Chr.17: 18246328-18246328 on Build GRCh38
rs774309595 Chr.17: 18246255-18246255 on Build GRCh38
rs969752638 Chr.17: 18246387-18246387 on Build GRCh38
rs148305879 Chr.17: 18246396-18246396 on Build GRCh38
rs1028235007 Chr.17: 18246322-18246322 on Build GRCh38
rs777520323 Chr.17: 18246589-18246589 on Build GRCh38
rs779124227 Chr.17: 18246393-18246393 on Build GRCh38
rs753062139 Chr.17: 18246372-18246372 on Build GRCh38
rs763077210 Chr.17: 18246684-18246684 on Build GRCh38
rs980344277 Chr.17: 18246395-18246395 on Build GRCh38
rs779140678 Chr.17: 18246474-18246474 on Build GRCh38
rs761366897 Chr.17: 18246355-18246355 on Build GRCh38
rs142476596 Chr.17: 18246318-18246318 on Build GRCh38
rs201997103 Chr.17: 18246608-18246608 on Build GRCh38
rs748231034 Chr.17: 18246530-18246530 on Build GRCh38
rs765490816 Chr.17: 18246683-18246683 on Build GRCh38
rs555225002 Chr.17: 18246371-18246371 on Build GRCh38
rs184572488 Chr.17: 18246346-18246346 on Build GRCh38
rs200998752 Chr.17: 18246455-18246455 on Build GRCh38
rs146517088 Chr.17: 18246364-18246364 on Build GRCh38
rs751576700 Chr.17: 18246580-18246580 on Build GRCh38
rs750478444 Chr.17: 18246478-18246478 on Build GRCh38
rs200974370 Chr.17: 18246465-18246465 on Build GRCh38
rs908017544 Chr.17: 18246639-18246639 on Build GRCh38
rs748328577 Chr.17: 18246297-18246297 on Build GRCh38
rs753279393 Chr.17: 18246279-18246279 on Build GRCh38
rs752042607 Chr.17: 18246290-18246290 on Build GRCh38
rs747523576 Chr.17: 18246649-18246649 on Build GRCh38
rs756287955 Chr.17: 18246300-18246300 on Build GRCh38
rs780091994 Chr.17: 18246483-18246483 on Build GRCh38
rs775298698 Chr.17: 18246344-18246344 on Build GRCh38
rs891649090 Chr.17: 18246521-18246521 on Build GRCh38
rs766743924 Chr.17: 18246551-18246552 on Build GRCh38
rs766497862 Chr.17: 18246685-18246685 on Build GRCh38
rs748886473 Chr.17: 18246590-18246590 on Build GRCh38
rs774789331 Chr.17: 18246520-18246520 on Build GRCh38
rs768421099 Chr.17: 18246427-18246427 on Build GRCh38
rs745813040 Chr.17: 18246335-18246335 on Build GRCh38
rs757498156 Chr.17: 18246471-18246471 on Build GRCh38
rs762318935 Chr.17: 18246666-18246666 on Build GRCh38
rs375385173 Chr.17: 18246559-18246559 on Build GRCh38
rs768015316 Chr.17: 18246456-18246456 on Build GRCh38
rs759452097 Chr.17: 18246263-18246263 on Build GRCh38
rs779746027 Chr.17: 18246409-18246409 on Build GRCh38
rs199790278 Chr.17: 18246271-18246271 on Build GRCh38
rs904408485 Chr.17: 18246507-18246507 on Build GRCh38
rs2856289 Chr.17: 18246540-18246540 on Build GRCh38
rs972281105 Chr.17: 18246428-18246429 on Build GRCh38
rs886486799 Chr.17: 18246260-18246260 on Build GRCh38
rs900539410 Chr.17: 18246288-18246288 on Build GRCh38
rs117072920 Chr.17: 18246664-18246664 on Build GRCh38
rs781604192 Chr.17: 18246296-18246296 on Build GRCh38
rs770315054 Chr.17: 18246510-18246510 on Build GRCh38
rs758292233 Chr.17: 18246481-18246481 on Build GRCh38
rs377765496 Chr.17: 18246493-18246493 on Build GRCh38
rs756308856 Chr.17: 18246373-18246373 on Build GRCh38
rs150392959 Chr.17: 18246648-18246648 on Build GRCh38
rs746891608 Chr.17: 18246485-18246485 on Build GRCh38
rs747887753 Chr.17: 18246584-18246584 on Build GRCh38
rs754838517 Chr.17: 18246489-18246489 on Build GRCh38
rs372477896 Chr.17: 18246293-18246293 on Build GRCh38
rs778189993 Chr.17: 18246380-18246380 on Build GRCh38
rs749331004 Chr.17: 18246316-18246316 on Build GRCh38
rs145824429 Chr.17: 18246587-18246587 on Build GRCh38
rs372325815 Chr.17: 18246569-18246569 on Build GRCh38
rs77114693 Chr.17: 18246568-18246568 on Build GRCh38
rs530284418 Chr.17: 18246532-18246532 on Build GRCh38
rs748875151 Chr.17: 18246504-18246504 on Build GRCh38
rs374888370 Chr.17: 18246273-18246273 on Build GRCh38

View Full Product Details