Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48725069-48725575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB13
Entrez Gene ID
10481
Gene Name
homeobox B13
Gene Aliases
PSGD
UniGene
Hs.66731
Gene Chromosome Location
Chr.17: 48724763-48728749 on Build GRCh38
Transcripts
NM_006361.5

SNP Detail

SNP ID Chromosome Location
rs3107299 Chr.17: 48725199-48725199 on Build GRCh38
rs8065028 Chr.17: 48725173-48725173 on Build GRCh38
rs374232757 Chr.17: 48725457-48725457 on Build GRCh38
rs540946232 Chr.17: 48725433-48725433 on Build GRCh38
rs976334016 Chr.17: 48725473-48725473 on Build GRCh38
rs900865731 Chr.17: 48725405-48725405 on Build GRCh38
rs933640225 Chr.17: 48725409-48725409 on Build GRCh38
rs988433790 Chr.17: 48725256-48725256 on Build GRCh38
rs754351804 Chr.17: 48725293-48725293 on Build GRCh38
rs368411572 Chr.17: 48725555-48725555 on Build GRCh38
rs750930408 Chr.17: 48725219-48725219 on Build GRCh38
rs942391182 Chr.17: 48725395-48725395 on Build GRCh38
rs1016779330 Chr.17: 48725135-48725135 on Build GRCh38
rs779730634 Chr.17: 48725344-48725344 on Build GRCh38
rs887076880 Chr.17: 48725467-48725467 on Build GRCh38
rs542966654 Chr.17: 48725556-48725556 on Build GRCh38
rs1046989070 Chr.17: 48725413-48725413 on Build GRCh38
rs926145936 Chr.17: 48725334-48725334 on Build GRCh38
rs1006340317 Chr.17: 48725253-48725254 on Build GRCh38
rs8069252 Chr.17: 48725268-48725268 on Build GRCh38
rs8069269 Chr.17: 48725307-48725307 on Build GRCh38
rs961912699 Chr.17: 48725426-48725426 on Build GRCh38
rs887222883 Chr.17: 48725110-48725110 on Build GRCh38
rs8065051 Chr.17: 48725218-48725218 on Build GRCh38
rs1029603693 Chr.17: 48725109-48725109 on Build GRCh38
rs1005388361 Chr.17: 48725431-48725432 on Build GRCh38
rs934226305 Chr.17: 48725456-48725456 on Build GRCh38
rs1017392613 Chr.17: 48725482-48725482 on Build GRCh38
rs905027813 Chr.17: 48725441-48725441 on Build GRCh38
rs1017038946 Chr.17: 48725198-48725198 on Build GRCh38
rs1004681436 Chr.17: 48725112-48725112 on Build GRCh38
rs8081840 Chr.17: 48725177-48725177 on Build GRCh38
rs980714994 Chr.17: 48725186-48725186 on Build GRCh38
rs142428986 Chr.17: 48725171-48725171 on Build GRCh38
rs577367530 Chr.17: 48725133-48725133 on Build GRCh38
rs560597408 Chr.17: 48725452-48725452 on Build GRCh38
rs968664569 Chr.17: 48725495-48725495 on Build GRCh38
rs946917673 Chr.17: 48725335-48725335 on Build GRCh38
rs529786232 Chr.17: 48725552-48725552 on Build GRCh38
rs766431405 Chr.17: 48725251-48725251 on Build GRCh38
rs557502169 Chr.17: 48725107-48725107 on Build GRCh38
rs936207240 Chr.17: 48725398-48725398 on Build GRCh38
rs928002768 Chr.17: 48725244-48725244 on Build GRCh38
rs553929053 Chr.17: 48725461-48725463 on Build GRCh38
rs1014776717 Chr.17: 48725422-48725422 on Build GRCh38
rs887002827 Chr.17: 48725430-48725430 on Build GRCh38
rs1004306352 Chr.17: 48725480-48725480 on Build GRCh38
rs979629609 Chr.17: 48725539-48725539 on Build GRCh38
rs925692719 Chr.17: 48725381-48725381 on Build GRCh38
rs991850221 Chr.17: 48725252-48725252 on Build GRCh38
rs914125961 Chr.17: 48725324-48725324 on Build GRCh38
rs979558289 Chr.17: 48725305-48725305 on Build GRCh38
rs368424992 Chr.17: 48725419-48725419 on Build GRCh38
rs574104328 Chr.17: 48725498-48725498 on Build GRCh38
rs980747215 Chr.17: 48725201-48725201 on Build GRCh38
rs945161120 Chr.17: 48725302-48725302 on Build GRCh38
rs1044258018 Chr.17: 48725379-48725379 on Build GRCh38
rs962825253 Chr.17: 48725169-48725169 on Build GRCh38
rs755585408 Chr.17: 48725546-48725546 on Build GRCh38
rs900121258 Chr.17: 48725447-48725447 on Build GRCh38

View Full Product Details