Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50532609-50533098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN3
Entrez Gene ID
55040
Gene Name
epsin 3
Gene Aliases
-
UniGene
Hs.670090
Gene Chromosome Location
Chr.17: 50532687-50543750 on Build GRCh38
Transcripts
NM_017957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130392 c.37G>C p.A13P Chr.17: 50532911-50532911 on Build GRCh38
5960238 c.32_33insT p.P12fs*48 Chr.17: 50532906-50532907 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1025275977 Chr.17: 50532905-50532905 on Build GRCh38
rs1051628920 Chr.17: 50532824-50532824 on Build GRCh38
rs761264863 Chr.17: 50533039-50533039 on Build GRCh38
rs1039912160 Chr.17: 50532998-50532998 on Build GRCh38
rs566870748 Chr.17: 50532720-50532720 on Build GRCh38
rs746329397 Chr.17: 50532799-50532799 on Build GRCh38
rs12601286 Chr.17: 50532672-50532672 on Build GRCh38
rs1001303235 Chr.17: 50532908-50532908 on Build GRCh38
rs950368348 Chr.17: 50532655-50532655 on Build GRCh38
rs116209498 Chr.17: 50532999-50532999 on Build GRCh38
rs951575931 Chr.17: 50532762-50532762 on Build GRCh38
rs757287919 Chr.17: 50532910-50532910 on Build GRCh38
rs928022534 Chr.17: 50532797-50532797 on Build GRCh38
rs761191577 Chr.17: 50532708-50532708 on Build GRCh38
rs886771888 Chr.17: 50532907-50532907 on Build GRCh38
rs763917614 Chr.17: 50532894-50532894 on Build GRCh38
rs922058635 Chr.17: 50532996-50532996 on Build GRCh38
rs111782297 Chr.17: 50532735-50532735 on Build GRCh38
rs926794819 Chr.17: 50532713-50532713 on Build GRCh38
rs981262468 Chr.17: 50532644-50532644 on Build GRCh38
rs936788486 Chr.17: 50532773-50532773 on Build GRCh38
rs111641654 Chr.17: 50532911-50532911 on Build GRCh38
rs945925987 Chr.17: 50532816-50532816 on Build GRCh38
rs569256669 Chr.17: 50533064-50533064 on Build GRCh38
rs936451 Chr.17: 50532737-50532737 on Build GRCh38
rs1037743580 Chr.17: 50533063-50533063 on Build GRCh38
rs980994623 Chr.17: 50532774-50532774 on Build GRCh38
rs549495042 Chr.17: 50533033-50533033 on Build GRCh38
rs758603799 Chr.17: 50532932-50532932 on Build GRCh38
rs544884167 Chr.17: 50532941-50532941 on Build GRCh38
rs760708103 Chr.17: 50532890-50532890 on Build GRCh38
rs886317934 Chr.17: 50533005-50533005 on Build GRCh38
rs1046839988 Chr.17: 50533001-50533001 on Build GRCh38
rs1005205650 Chr.17: 50533021-50533021 on Build GRCh38
rs547286508 Chr.17: 50532976-50532976 on Build GRCh38
rs1002469918 Chr.17: 50533065-50533065 on Build GRCh38
rs988186599 Chr.17: 50532940-50532940 on Build GRCh38
rs749594071 Chr.17: 50532977-50532977 on Build GRCh38
rs1030234051 Chr.17: 50532654-50532654 on Build GRCh38
rs942514840 Chr.17: 50532879-50532879 on Build GRCh38
rs755046 Chr.17: 50532911-50532911 on Build GRCh38
rs2305999 Chr.17: 50532768-50532768 on Build GRCh38
rs1001073477 Chr.17: 50532628-50532628 on Build GRCh38
rs984376158 Chr.17: 50532763-50532763 on Build GRCh38
rs978128686 Chr.17: 50532749-50532749 on Build GRCh38
rs145303593 Chr.17: 50532926-50532926 on Build GRCh38
rs1004591523 Chr.17: 50532656-50532656 on Build GRCh38
rs752434120 Chr.17: 50532862-50532862 on Build GRCh38
rs879453134 Chr.17: 50532837-50532837 on Build GRCh38
rs538269148 Chr.17: 50533067-50533067 on Build GRCh38
rs578200746 Chr.17: 50532790-50532790 on Build GRCh38
rs540849227 Chr.17: 50532807-50532807 on Build GRCh38
rs779697363 Chr.17: 50532849-50532849 on Build GRCh38
rs1002762151 Chr.17: 50532929-50532929 on Build GRCh38
rs564859800 Chr.17: 50532950-50532950 on Build GRCh38
rs144900599 Chr.17: 50533012-50533012 on Build GRCh38
rs770397617 Chr.17: 50532906-50532906 on Build GRCh38
rs768232853 Chr.17: 50532967-50532967 on Build GRCh38
rs1017591979 Chr.17: 50532925-50532925 on Build GRCh38
rs969953221 Chr.17: 50532772-50532772 on Build GRCh38
rs542754638 Chr.17: 50532895-50532895 on Build GRCh38
rs755947861 Chr.17: 50532808-50532808 on Build GRCh38
rs974606357 Chr.17: 50532704-50532704 on Build GRCh38
rs533767707 Chr.17: 50532696-50532696 on Build GRCh38
rs559333532 Chr.17: 50532734-50532734 on Build GRCh38
rs756126272 Chr.17: 50532648-50532648 on Build GRCh38
rs753167824 Chr.17: 50532656-50532657 on Build GRCh38
rs913911289 Chr.17: 50532813-50532813 on Build GRCh38
rs778143247 Chr.17: 50532724-50532724 on Build GRCh38
rs758538252 Chr.17: 50532721-50532721 on Build GRCh38
rs191315585 Chr.17: 50532966-50532966 on Build GRCh38
rs186833208 Chr.17: 50532888-50532888 on Build GRCh38
rs1004221581 Chr.17: 50532993-50532994 on Build GRCh38
rs548125982 Chr.17: 50532752-50532752 on Build GRCh38
rs961042649 Chr.17: 50532936-50532936 on Build GRCh38
rs1052411912 Chr.17: 50532660-50532660 on Build GRCh38
rs966991516 Chr.17: 50532960-50532961 on Build GRCh38
rs766057581 Chr.17: 50532651-50532651 on Build GRCh38
rs945981161 Chr.17: 50532676-50532676 on Build GRCh38
rs767405764 Chr.17: 50532859-50532859 on Build GRCh38
rs1018325724 Chr.17: 50533038-50533038 on Build GRCh38
rs375564066 Chr.17: 50532832-50532832 on Build GRCh38
rs546662252 Chr.17: 50532711-50532711 on Build GRCh38
rs534560535 Chr.17: 50532638-50532638 on Build GRCh38
rs531499673 Chr.17: 50532630-50532630 on Build GRCh38
rs933852637 Chr.17: 50533000-50533000 on Build GRCh38
rs780531550 Chr.17: 50532964-50532964 on Build GRCh38
rs905416061 Chr.17: 50533046-50533046 on Build GRCh38
rs1038060913 Chr.17: 50533022-50533022 on Build GRCh38

View Full Product Details