Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79046463-79046966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF1
Entrez Gene ID
114897
Gene Name
C1q and tumor necrosis factor related protein 1
Gene Aliases
CTRP1, GIP, ZSIG37
UniGene
Hs.201398
Gene Chromosome Location
Chr.17: 79022934-79049788 on Build GRCh38
Transcripts
NM_030968.3 NM_153372.1 NM_198593.3 NM_198594.2 NR_049769.1 XM_006721663.1 XM_006721664.1 XM_006721666.1 XM_017024126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861131 c.233A>C p.D78A Chr.17: 79046632-79046632 on Build GRCh38
4834778 c.245C>A p.S82Y Chr.17: 79046644-79046644 on Build GRCh38
6615408 c.291C>T p.T97T Chr.17: 79046660-79046660 on Build GRCh38
4746038 c.157G>A p.A53T Chr.17: 79046556-79046556 on Build GRCh38
6615404 c.244C>T p.R82W Chr.17: 79046613-79046613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765012138 Chr.17: 79046514-79046514 on Build GRCh38
rs368374363 Chr.17: 79046587-79046587 on Build GRCh38
rs781282070 Chr.17: 79046669-79046669 on Build GRCh38
rs901828719 Chr.17: 79046805-79046805 on Build GRCh38
rs779288561 Chr.17: 79046592-79046592 on Build GRCh38
rs566478936 Chr.17: 79046613-79046613 on Build GRCh38
rs775036302 Chr.17: 79046727-79046727 on Build GRCh38
rs777040057 Chr.17: 79046790-79046790 on Build GRCh38
rs199649620 Chr.17: 79046734-79046734 on Build GRCh38
rs11077409 Chr.17: 79046636-79046636 on Build GRCh38
rs957102171 Chr.17: 79046918-79046918 on Build GRCh38
rs748358025 Chr.17: 79046629-79046629 on Build GRCh38
rs760259627 Chr.17: 79046659-79046659 on Build GRCh38
rs766028634 Chr.17: 79046562-79046562 on Build GRCh38
rs147754695 Chr.17: 79046845-79046845 on Build GRCh38
rs531768528 Chr.17: 79046509-79046509 on Build GRCh38
rs911949137 Chr.17: 79046933-79046933 on Build GRCh38
rs1035847572 Chr.17: 79046749-79046749 on Build GRCh38
rs568819987 Chr.17: 79046946-79046946 on Build GRCh38
rs575004114 Chr.17: 79046637-79046637 on Build GRCh38
rs539583596 Chr.17: 79046520-79046520 on Build GRCh38
rs148895244 Chr.17: 79046622-79046622 on Build GRCh38
rs1001440408 Chr.17: 79046869-79046869 on Build GRCh38
rs754382655 Chr.17: 79046927-79046927 on Build GRCh38
rs768099885 Chr.17: 79046697-79046697 on Build GRCh38
rs746413095 Chr.17: 79046600-79046600 on Build GRCh38
rs994722836 Chr.17: 79046858-79046858 on Build GRCh38
rs562094752 Chr.17: 79046784-79046784 on Build GRCh38
rs148307780 Chr.17: 79046653-79046653 on Build GRCh38
rs546728281 Chr.17: 79046910-79046910 on Build GRCh38
rs749002175 Chr.17: 79046656-79046656 on Build GRCh38
rs367911694 Chr.17: 79046914-79046914 on Build GRCh38
rs776839033 Chr.17: 79046623-79046623 on Build GRCh38
rs577010981 Chr.17: 79046719-79046719 on Build GRCh38
rs759122216 Chr.17: 79046652-79046652 on Build GRCh38
rs545938374 Chr.17: 79046720-79046720 on Build GRCh38
rs748365298 Chr.17: 79046698-79046698 on Build GRCh38
rs770053807 Chr.17: 79046703-79046703 on Build GRCh38
rs762896549 Chr.17: 79046556-79046556 on Build GRCh38
rs537727380 Chr.17: 79046687-79046687 on Build GRCh38
rs533114512 Chr.17: 79046892-79046892 on Build GRCh38
rs754739302 Chr.17: 79046572-79046572 on Build GRCh38
rs141594726 Chr.17: 79046503-79046503 on Build GRCh38
rs755078571 Chr.17: 79046668-79046668 on Build GRCh38
rs773030872 Chr.17: 79046633-79046633 on Build GRCh38
rs766884176 Chr.17: 79046748-79046748 on Build GRCh38
rs760309832 Chr.17: 79046728-79046728 on Build GRCh38
rs200961717 Chr.17: 79046614-79046614 on Build GRCh38
rs371416155 Chr.17: 79046726-79046726 on Build GRCh38
rs182886833 Chr.17: 79046729-79046729 on Build GRCh38
rs780552494 Chr.17: 79046663-79046663 on Build GRCh38
rs367663124 Chr.17: 79046688-79046688 on Build GRCh38
rs749653695 Chr.17: 79046580-79046580 on Build GRCh38
rs950602400 Chr.17: 79046895-79046895 on Build GRCh38
rs763311893 Chr.17: 79046732-79046732 on Build GRCh38
rs557302812 Chr.17: 79046708-79046708 on Build GRCh38
rs375904511 Chr.17: 79046660-79046660 on Build GRCh38
rs751113260 Chr.17: 79046565-79046565 on Build GRCh38
rs767296445 Chr.17: 79046585-79046585 on Build GRCh38
rs77619548 Chr.17: 79046945-79046945 on Build GRCh38
rs769495646 Chr.17: 79046632-79046632 on Build GRCh38
rs141403015 Chr.17: 79046654-79046654 on Build GRCh38
rs745653844 Chr.17: 79046584-79046584 on Build GRCh38
rs546715848 Chr.17: 79046538-79046538 on Build GRCh38
rs372036566 Chr.17: 79046923-79046923 on Build GRCh38
rs550842313 Chr.17: 79046840-79046840 on Build GRCh38
rs878933840 Chr.17: 79046778-79046778 on Build GRCh38
rs145339677 Chr.17: 79046785-79046785 on Build GRCh38
rs773052350 Chr.17: 79046521-79046521 on Build GRCh38
rs554565794 Chr.17: 79046655-79046655 on Build GRCh38
rs1031205423 Chr.17: 79046886-79046886 on Build GRCh38
rs1012985117 Chr.17: 79046804-79046804 on Build GRCh38
rs1035455361 Chr.17: 79046546-79046546 on Build GRCh38
rs372698196 Chr.17: 79046634-79046634 on Build GRCh38
rs758961252 Chr.17: 79046661-79046661 on Build GRCh38
rs996148078 Chr.17: 79046813-79046813 on Build GRCh38
rs971020775 Chr.17: 79046942-79046942 on Build GRCh38
rs752079631 Chr.17: 79046735-79046735 on Build GRCh38
rs201651330 Chr.17: 79046664-79046664 on Build GRCh38
rs1026268700 Chr.17: 79046864-79046864 on Build GRCh38
rs187592493 Chr.17: 79046766-79046766 on Build GRCh38
rs535464469 Chr.17: 79046944-79046944 on Build GRCh38

View Full Product Details