Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1978469-1978976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
UniGene
Hs.651905
Gene Chromosome Location
Chr.19: 1941149-1981338 on Build GRCh38
Transcripts
NM_001319.6 XM_005259498.1 XM_005259499.3 XM_005259500.1 XM_005259501.2 XM_017026297.1 XM_017026298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509217 c.448-9C>G p.? Chr.19: 1978850-1978850 on Build GRCh38
6636799 c.548C>T p.A183V Chr.19: 1978959-1978959 on Build GRCh38
5509216 c.448-10C>G p.? Chr.19: 1978849-1978849 on Build GRCh38
5516801 c.447+5G>A p.? Chr.19: 1978755-1978755 on Build GRCh38
6636803 c.524G>A p.R175H Chr.19: 1978935-1978935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771642864 Chr.19: 1978678-1978678 on Build GRCh38
rs1044006656 Chr.19: 1978767-1978767 on Build GRCh38
rs530717617 Chr.19: 1978841-1978841 on Build GRCh38
rs747829898 Chr.19: 1978844-1978844 on Build GRCh38
rs768168593 Chr.19: 1978823-1978824 on Build GRCh38
rs755824841 Chr.19: 1978939-1978939 on Build GRCh38
rs532979869 Chr.19: 1978759-1978759 on Build GRCh38
rs746697797 Chr.19: 1978774-1978774 on Build GRCh38
rs771550767 Chr.19: 1978757-1978765 on Build GRCh38
rs765398175 Chr.19: 1978596-1978596 on Build GRCh38
rs781390723 Chr.19: 1978620-1978620 on Build GRCh38
rs148091512 Chr.19: 1978887-1978887 on Build GRCh38
rs889420489 Chr.19: 1978787-1978787 on Build GRCh38
rs757423595 Chr.19: 1978555-1978555 on Build GRCh38
rs115802254 Chr.19: 1978580-1978580 on Build GRCh38
rs771042169 Chr.19: 1978821-1978821 on Build GRCh38
rs759031786 Chr.19: 1978915-1978915 on Build GRCh38
rs754448313 Chr.19: 1978842-1978842 on Build GRCh38
rs761425981 Chr.19: 1978829-1978830 on Build GRCh38
rs2277737 Chr.19: 1978834-1978834 on Build GRCh38
rs539820319 Chr.19: 1978579-1978579 on Build GRCh38
rs775715488 Chr.19: 1978831-1978831 on Build GRCh38
rs555939754 Chr.19: 1978633-1978633 on Build GRCh38
rs373180104 Chr.19: 1978757-1978757 on Build GRCh38
rs559620940 Chr.19: 1978492-1978492 on Build GRCh38
rs1048604023 Chr.19: 1978592-1978592 on Build GRCh38
rs759616174 Chr.19: 1978825-1978825 on Build GRCh38
rs775225537 Chr.19: 1978582-1978582 on Build GRCh38
rs760931176 Chr.19: 1978876-1978876 on Build GRCh38
rs760497412 Chr.19: 1978545-1978545 on Build GRCh38
rs763166502 Chr.19: 1978760-1978764 on Build GRCh38
rs112289928 Chr.19: 1978792-1978792 on Build GRCh38
rs769637159 Chr.19: 1978708-1978708 on Build GRCh38
rs761012619 Chr.19: 1978832-1978832 on Build GRCh38
rs772095290 Chr.19: 1978951-1978951 on Build GRCh38
rs778294062 Chr.19: 1978636-1978636 on Build GRCh38
rs756443125 Chr.19: 1978760-1978760 on Build GRCh38
rs574120359 Chr.19: 1978693-1978693 on Build GRCh38
rs751939931 Chr.19: 1978569-1978569 on Build GRCh38
rs780374755 Chr.19: 1978520-1978520 on Build GRCh38
rs1034829277 Chr.19: 1978789-1978789 on Build GRCh38
rs376760598 Chr.19: 1978900-1978900 on Build GRCh38
rs764514642 Chr.19: 1978763-1978763 on Build GRCh38
rs541143980 Chr.19: 1978702-1978702 on Build GRCh38
rs745805274 Chr.19: 1978950-1978950 on Build GRCh38
rs370005594 Chr.19: 1978688-1978688 on Build GRCh38
rs753975205 Chr.19: 1978504-1978504 on Build GRCh38
rs374130056 Chr.19: 1978840-1978840 on Build GRCh38
rs753651278 Chr.19: 1978942-1978942 on Build GRCh38
rs372331895 Chr.19: 1978711-1978711 on Build GRCh38
rs777165253 Chr.19: 1978894-1978894 on Build GRCh38
rs758587163 Chr.19: 1978563-1978563 on Build GRCh38
rs905046084 Chr.19: 1978776-1978776 on Build GRCh38
rs780708950 Chr.19: 1978843-1978843 on Build GRCh38
rs141652429 Chr.19: 1978687-1978687 on Build GRCh38
rs376961305 Chr.19: 1978594-1978594 on Build GRCh38
rs147892032 Chr.19: 1978627-1978627 on Build GRCh38
rs200595669 Chr.19: 1978527-1978527 on Build GRCh38
rs755274972 Chr.19: 1978571-1978571 on Build GRCh38
rs754443192 Chr.19: 1978835-1978835 on Build GRCh38
rs751831667 Chr.19: 1978738-1978738 on Build GRCh38
rs747553413 Chr.19: 1978525-1978525 on Build GRCh38
rs750499976 Chr.19: 1978597-1978597 on Build GRCh38
rs775634169 Chr.19: 1978868-1978868 on Build GRCh38
rs745385169 Chr.19: 1978648-1978648 on Build GRCh38
rs767644859 Chr.19: 1978722-1978722 on Build GRCh38
rs372176315 Chr.19: 1978568-1978568 on Build GRCh38
rs767651270 Chr.19: 1978826-1978826 on Build GRCh38
rs767037697 Chr.19: 1978927-1978927 on Build GRCh38
rs566084947 Chr.19: 1978530-1978530 on Build GRCh38
rs766439465 Chr.19: 1978726-1978726 on Build GRCh38
rs750870831 Chr.19: 1978902-1978902 on Build GRCh38
rs17856624 Chr.19: 1978642-1978642 on Build GRCh38
rs759960518 Chr.19: 1978759-1978760 on Build GRCh38
rs747904813 Chr.19: 1978810-1978810 on Build GRCh38
rs373031517 Chr.19: 1978584-1978584 on Build GRCh38
rs745939601 Chr.19: 1978864-1978864 on Build GRCh38
rs930314857 Chr.19: 1978590-1978590 on Build GRCh38
rs772189864 Chr.19: 1978865-1978865 on Build GRCh38
rs759091146 Chr.19: 1978537-1978537 on Build GRCh38
rs749045334 Chr.19: 1978852-1978852 on Build GRCh38
rs775457030 Chr.19: 1978539-1978539 on Build GRCh38
rs776686742 Chr.19: 1978588-1978588 on Build GRCh38
rs774113662 Chr.19: 1978534-1978534 on Build GRCh38
rs1037188197 Chr.19: 1978606-1978606 on Build GRCh38
rs559163921 Chr.19: 1978754-1978754 on Build GRCh38
rs778889620 Chr.19: 1978949-1978949 on Build GRCh38
rs748592136 Chr.19: 1978575-1978575 on Build GRCh38
rs754618081 Chr.19: 1978775-1978775 on Build GRCh38
rs755399651 Chr.19: 1978609-1978609 on Build GRCh38
rs781722961 Chr.19: 1978572-1978572 on Build GRCh38
rs933683318 Chr.19: 1978521-1978521 on Build GRCh38
rs762967432 Chr.19: 1978598-1978598 on Build GRCh38
rs998579549 Chr.19: 1978517-1978517 on Build GRCh38
rs767226803 Chr.19: 1978847-1978848 on Build GRCh38
rs530792278 Chr.19: 1978847-1978847 on Build GRCh38
rs758818331 Chr.19: 1978518-1978518 on Build GRCh38
rs748842297 Chr.19: 1978528-1978528 on Build GRCh38
rs761629089 Chr.19: 1978706-1978706 on Build GRCh38
rs773754063 Chr.19: 1978825-1978825 on Build GRCh38
rs1004328528 Chr.19: 1978595-1978595 on Build GRCh38
rs904992255 Chr.19: 1978536-1978537 on Build GRCh38
rs144771760 Chr.19: 1978928-1978928 on Build GRCh38
rs376378625 Chr.19: 1978516-1978516 on Build GRCh38
rs753652184 Chr.19: 1978553-1978553 on Build GRCh38
rs764141908 Chr.19: 1978552-1978552 on Build GRCh38
rs749207815 Chr.19: 1978819-1978819 on Build GRCh38
rs143642401 Chr.19: 1978862-1978862 on Build GRCh38
rs765215987 Chr.19: 1978559-1978559 on Build GRCh38
rs1028346801 Chr.19: 1978519-1978519 on Build GRCh38
rs933735468 Chr.19: 1978696-1978696 on Build GRCh38
rs753030863 Chr.19: 1978758-1978758 on Build GRCh38
rs544074723 Chr.19: 1978607-1978607 on Build GRCh38
rs4806824 Chr.19: 1978797-1978797 on Build GRCh38
rs376825133 Chr.19: 1978576-1978576 on Build GRCh38
rs779686141 Chr.19: 1978772-1978772 on Build GRCh38
rs934160560 Chr.19: 1978941-1978941 on Build GRCh38
rs773196915 Chr.19: 1978818-1978818 on Build GRCh38
rs776258099 Chr.19: 1978705-1978705 on Build GRCh38
rs758042298 Chr.19: 1978769-1978769 on Build GRCh38
rs769562349 Chr.19: 1978813-1978813 on Build GRCh38
rs754393831 Chr.19: 1978768-1978768 on Build GRCh38
rs987865145 Chr.19: 1978755-1978755 on Build GRCh38
rs201199524 Chr.19: 1978526-1978526 on Build GRCh38

View Full Product Details