Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1979197-1979705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
UniGene
Hs.651905
Gene Chromosome Location
Chr.19: 1941149-1981338 on Build GRCh38
Transcripts
NM_001319.6 XM_005259498.1 XM_005259499.3 XM_005259500.1 XM_005259501.2 XM_017026297.1 XM_017026298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202501 c.925C>T p.R309W Chr.19: 1979566-1979566 on Build GRCh38
6636798 c.1002+6_1002+7insG p.? Chr.19: 1979649-1979650 on Build GRCh38
1202503 c.787C>T p.R263W Chr.19: 1979337-1979337 on Build GRCh38
6636800 c.779T>G p.L260R Chr.19: 1979329-1979329 on Build GRCh38
5513424 c.777G>A p.T259T Chr.19: 1979327-1979327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043790735 Chr.19: 1979507-1979507 on Build GRCh38
rs773910139 Chr.19: 1979650-1979650 on Build GRCh38
rs746355518 Chr.19: 1979233-1979233 on Build GRCh38
rs762868052 Chr.19: 1979264-1979264 on Build GRCh38
rs756876055 Chr.19: 1979508-1979508 on Build GRCh38
rs553073752 Chr.19: 1979476-1979476 on Build GRCh38
rs376859221 Chr.19: 1979481-1979481 on Build GRCh38
rs35076486 Chr.19: 1979506-1979506 on Build GRCh38
rs749457852 Chr.19: 1979296-1979296 on Build GRCh38
rs921283190 Chr.19: 1979655-1979655 on Build GRCh38
rs760503792 Chr.19: 1979437-1979437 on Build GRCh38
rs781437638 Chr.19: 1979468-1979471 on Build GRCh38
rs371729293 Chr.19: 1979484-1979484 on Build GRCh38
rs779637277 Chr.19: 1979604-1979604 on Build GRCh38
rs778766640 Chr.19: 1979474-1979474 on Build GRCh38
rs143970128 Chr.19: 1979570-1979570 on Build GRCh38
rs758150792 Chr.19: 1979527-1979527 on Build GRCh38
rs191965385 Chr.19: 1979446-1979446 on Build GRCh38
rs747657577 Chr.19: 1979321-1979321 on Build GRCh38
rs756723494 Chr.19: 1979589-1979589 on Build GRCh38
rs766073943 Chr.19: 1979281-1979281 on Build GRCh38
rs781687610 Chr.19: 1979464-1979464 on Build GRCh38
rs11550450 Chr.19: 1979387-1979387 on Build GRCh38
rs759472762 Chr.19: 1979422-1979422 on Build GRCh38
rs765446422 Chr.19: 1979450-1979450 on Build GRCh38
rs764650976 Chr.19: 1979590-1979590 on Build GRCh38
rs781005771 Chr.19: 1979619-1979619 on Build GRCh38
rs778124414 Chr.19: 1979290-1979290 on Build GRCh38
rs752757382 Chr.19: 1979434-1979453 on Build GRCh38
rs754773789 Chr.19: 1979610-1979610 on Build GRCh38
rs574206645 Chr.19: 1979467-1979467 on Build GRCh38
rs778198286 Chr.19: 1979440-1979447 on Build GRCh38
rs776023184 Chr.19: 1979673-1979673 on Build GRCh38
rs754896087 Chr.19: 1979433-1979446 on Build GRCh38
rs541252222 Chr.19: 1979475-1979475 on Build GRCh38
rs149326311 Chr.19: 1979580-1979580 on Build GRCh38
rs771999035 Chr.19: 1979276-1979276 on Build GRCh38
rs746334360 Chr.19: 1979494-1979496 on Build GRCh38
rs35677464 Chr.19: 1979600-1979600 on Build GRCh38
rs767521303 Chr.19: 1979284-1979284 on Build GRCh38
rs770511168 Chr.19: 1979419-1979419 on Build GRCh38
rs974100478 Chr.19: 1979653-1979653 on Build GRCh38
rs780374626 Chr.19: 1979461-1979461 on Build GRCh38
rs754133362 Chr.19: 1979372-1979372 on Build GRCh38
rs749970052 Chr.19: 1979594-1979594 on Build GRCh38
rs1056898992 Chr.19: 1979515-1979515 on Build GRCh38
rs754934394 Chr.19: 1979283-1979283 on Build GRCh38
rs953086134 Chr.19: 1979661-1979661 on Build GRCh38
rs753186272 Chr.19: 1979586-1979586 on Build GRCh38
rs750798326 Chr.19: 1979391-1979391 on Build GRCh38
rs751596129 Chr.19: 1979282-1979282 on Build GRCh38
rs200803486 Chr.19: 1979550-1979550 on Build GRCh38
rs368618107 Chr.19: 1979364-1979364 on Build GRCh38
rs868019413 Chr.19: 1979401-1979401 on Build GRCh38
rs751890447 Chr.19: 1979491-1979491 on Build GRCh38
rs934172875 Chr.19: 1979254-1979254 on Build GRCh38
rs779700928 Chr.19: 1979528-1979528 on Build GRCh38
rs776561765 Chr.19: 1979483-1979483 on Build GRCh38
rs917769449 Chr.19: 1979414-1979414 on Build GRCh38
rs961803729 Chr.19: 1979593-1979593 on Build GRCh38
rs758693294 Chr.19: 1979392-1979392 on Build GRCh38
rs374546538 Chr.19: 1979315-1979315 on Build GRCh38
rs773311679 Chr.19: 1979559-1979559 on Build GRCh38
rs753889632 Chr.19: 1979445-1979445 on Build GRCh38
rs368738419 Chr.19: 1979288-1979288 on Build GRCh38
rs541986661 Chr.19: 1979679-1979680 on Build GRCh38
rs764062633 Chr.19: 1979371-1979371 on Build GRCh38
rs749373021 Chr.19: 1979650-1979650 on Build GRCh38
rs771980201 Chr.19: 1979424-1979424 on Build GRCh38
rs555759847 Chr.19: 1979462-1979462 on Build GRCh38
rs550989112 Chr.19: 1979517-1979517 on Build GRCh38
rs540582543 Chr.19: 1979649-1979650 on Build GRCh38
rs779029842 Chr.19: 1979433-1979439 on Build GRCh38
rs753236076 Chr.19: 1979505-1979505 on Build GRCh38
rs764055678 Chr.19: 1979444-1979444 on Build GRCh38
rs768491176 Chr.19: 1979535-1979535 on Build GRCh38
rs774317639 Chr.19: 1979355-1979355 on Build GRCh38
rs760867567 Chr.19: 1979367-1979367 on Build GRCh38
rs1030387085 Chr.19: 1979631-1979631 on Build GRCh38
rs753769577 Chr.19: 1979413-1979425 on Build GRCh38
rs137921162 Chr.19: 1979396-1979396 on Build GRCh38
rs754395841 Chr.19: 1979683-1979683 on Build GRCh38
rs941452383 Chr.19: 1979457-1979457 on Build GRCh38
rs762969611 Chr.19: 1979563-1979563 on Build GRCh38
rs746787307 Chr.19: 1979532-1979532 on Build GRCh38
rs774985143 Chr.19: 1979226-1979226 on Build GRCh38
rs777115623 Chr.19: 1979413-1979413 on Build GRCh38
rs767902578 Chr.19: 1979581-1979581 on Build GRCh38
rs201840791 Chr.19: 1979496-1979496 on Build GRCh38
rs1043846045 Chr.19: 1979306-1979306 on Build GRCh38
rs775618280 Chr.19: 1979436-1979436 on Build GRCh38
rs201521868 Chr.19: 1979680-1979680 on Build GRCh38
rs757917899 Chr.19: 1979601-1979601 on Build GRCh38
rs768198173 Chr.19: 1979243-1979243 on Build GRCh38
rs368950633 Chr.19: 1979504-1979504 on Build GRCh38
rs138558858 Chr.19: 1979263-1979263 on Build GRCh38
rs1031816961 Chr.19: 1979439-1979439 on Build GRCh38
rs757455663 Chr.19: 1979383-1979383 on Build GRCh38
rs911479741 Chr.19: 1979677-1979677 on Build GRCh38
rs759348180 Chr.19: 1979363-1979363 on Build GRCh38
rs562955675 Chr.19: 1979542-1979542 on Build GRCh38
rs765282962 Chr.19: 1979485-1979485 on Build GRCh38
rs758687862 Chr.19: 1979439-1979439 on Build GRCh38
rs374004002 Chr.19: 1979658-1979658 on Build GRCh38
rs769746203 Chr.19: 1979632-1979632 on Build GRCh38
rs746657543 Chr.19: 1979605-1979605 on Build GRCh38
rs375847481 Chr.19: 1979674-1979674 on Build GRCh38
rs776152304 Chr.19: 1979252-1979252 on Build GRCh38
rs774082363 Chr.19: 1979420-1979420 on Build GRCh38
rs78864193 Chr.19: 1979574-1979574 on Build GRCh38
rs747897974 Chr.19: 1979625-1979625 on Build GRCh38
rs371407966 Chr.19: 1979314-1979314 on Build GRCh38
rs758816150 Chr.19: 1979459-1979459 on Build GRCh38
rs955699683 Chr.19: 1979304-1979304 on Build GRCh38
rs769394939 Chr.19: 1979410-1979410 on Build GRCh38
rs373599985 Chr.19: 1979221-1979221 on Build GRCh38
rs770798173 Chr.19: 1979354-1979354 on Build GRCh38
rs1010177291 Chr.19: 1979492-1979492 on Build GRCh38
rs772975375 Chr.19: 1979336-1979336 on Build GRCh38
rs771350553 Chr.19: 1979453-1979454 on Build GRCh38
rs935308358 Chr.19: 1979511-1979511 on Build GRCh38
rs752677240 Chr.19: 1979366-1979366 on Build GRCh38
rs142134343 Chr.19: 1979327-1979327 on Build GRCh38
rs372348470 Chr.19: 1979418-1979418 on Build GRCh38
rs776486793 Chr.19: 1979541-1979541 on Build GRCh38
rs747247580 Chr.19: 1979447-1979448 on Build GRCh38
rs762490233 Chr.19: 1979351-1979351 on Build GRCh38
rs750635620 Chr.19: 1979453-1979453 on Build GRCh38
rs764952388 Chr.19: 1979262-1979262 on Build GRCh38
rs747454688 Chr.19: 1979409-1979409 on Build GRCh38
rs779221452 Chr.19: 1979310-1979310 on Build GRCh38
rs761260914 Chr.19: 1979255-1979255 on Build GRCh38
rs774488810 Chr.19: 1979652-1979652 on Build GRCh38
rs777627702 Chr.19: 1979641-1979641 on Build GRCh38
rs748753678 Chr.19: 1979465-1979465 on Build GRCh38
rs896431324 Chr.19: 1979365-1979365 on Build GRCh38
rs757693955 Chr.19: 1979309-1979309 on Build GRCh38
rs759773292 Chr.19: 1979656-1979656 on Build GRCh38

View Full Product Details