Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41219097-41219596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382914 c.24G>A p.M8I Chr.19: 41219416-41219416 on Build GRCh38
6917894 c.41C>G p.A14G Chr.19: 41219433-41219433 on Build GRCh38
5855869 c.11G>A p.R4Q Chr.19: 41219403-41219403 on Build GRCh38
6964625 c.77delC p.R28fs*70 Chr.19: 41219469-41219469 on Build GRCh38
4991297 c.75G>T p.M25I Chr.19: 41219467-41219467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532728988 Chr.19: 41219172-41219172 on Build GRCh38
rs774863157 Chr.19: 41219359-41219359 on Build GRCh38
rs978725826 Chr.19: 41219261-41219261 on Build GRCh38
rs554900580 Chr.19: 41219290-41219290 on Build GRCh38
rs781643633 Chr.19: 41219391-41219391 on Build GRCh38
rs1007065743 Chr.19: 41219269-41219269 on Build GRCh38
rs779811231 Chr.19: 41219403-41219403 on Build GRCh38
rs769574605 Chr.19: 41219422-41219422 on Build GRCh38
rs897184491 Chr.19: 41219182-41219182 on Build GRCh38
rs986102949 Chr.19: 41219284-41219284 on Build GRCh38
rs879539383 Chr.19: 41219324-41219324 on Build GRCh38
rs532768963 Chr.19: 41219461-41219461 on Build GRCh38
rs931607661 Chr.19: 41219270-41219270 on Build GRCh38
rs1036585965 Chr.19: 41219323-41219323 on Build GRCh38
rs778406577 Chr.19: 41219453-41219453 on Build GRCh38
rs977982354 Chr.19: 41219152-41219152 on Build GRCh38
rs766690375 Chr.19: 41219376-41219376 on Build GRCh38
rs745532899 Chr.19: 41219358-41219358 on Build GRCh38
rs760191944 Chr.19: 41219149-41219149 on Build GRCh38
rs941902787 Chr.19: 41219318-41219318 on Build GRCh38
rs776668582 Chr.19: 41219412-41219412 on Build GRCh38
rs145867512 Chr.19: 41219445-41219445 on Build GRCh38
rs751579351 Chr.19: 41219380-41219380 on Build GRCh38
rs199900995 Chr.19: 41219354-41219354 on Build GRCh38
rs569949183 Chr.19: 41219325-41219325 on Build GRCh38
rs530407469 Chr.19: 41219428-41219428 on Build GRCh38
rs922946589 Chr.19: 41219230-41219230 on Build GRCh38
rs997221035 Chr.19: 41219409-41219409 on Build GRCh38
rs10411373 Chr.19: 41219397-41219397 on Build GRCh38
rs975773929 Chr.19: 41219180-41219180 on Build GRCh38
rs548131195 Chr.19: 41219173-41219173 on Build GRCh38
rs767609165 Chr.19: 41219446-41219446 on Build GRCh38
rs369717581 Chr.19: 41219353-41219353 on Build GRCh38
rs371082541 Chr.19: 41219441-41219441 on Build GRCh38
rs753148751 Chr.19: 41219450-41219450 on Build GRCh38
rs760325544 Chr.19: 41219360-41219360 on Build GRCh38
rs551974950 Chr.19: 41219124-41219124 on Build GRCh38
rs984558221 Chr.19: 41219134-41219134 on Build GRCh38
rs754135882 Chr.19: 41219460-41219460 on Build GRCh38
rs1052205974 Chr.19: 41219490-41219490 on Build GRCh38
rs759764389 Chr.19: 41219529-41219529 on Build GRCh38
rs969240754 Chr.19: 41219336-41219336 on Build GRCh38
rs768231361 Chr.19: 41219471-41219471 on Build GRCh38
rs373496065 Chr.19: 41219335-41219335 on Build GRCh38
rs112524750 Chr.19: 41219432-41219432 on Build GRCh38
rs764107116 Chr.19: 41219361-41219361 on Build GRCh38
rs73550504 Chr.19: 41219505-41219505 on Build GRCh38
rs770793832 Chr.19: 41219526-41219526 on Build GRCh38
rs1013257352 Chr.19: 41219559-41219559 on Build GRCh38
rs1000076660 Chr.19: 41219472-41219472 on Build GRCh38
rs765153721 Chr.19: 41219367-41219367 on Build GRCh38
rs992207619 Chr.19: 41219313-41219313 on Build GRCh38
rs776755976 Chr.19: 41219363-41219363 on Build GRCh38
rs755101210 Chr.19: 41219387-41219387 on Build GRCh38
rs774019242 Chr.19: 41219357-41219357 on Build GRCh38
rs1013170889 Chr.19: 41219138-41219138 on Build GRCh38
rs941494728 Chr.19: 41219164-41219164 on Build GRCh38
rs747727484 Chr.19: 41219510-41219510 on Build GRCh38
rs182481095 Chr.19: 41219402-41219402 on Build GRCh38
rs563334837 Chr.19: 41219427-41219427 on Build GRCh38
rs774510600 Chr.19: 41219527-41219527 on Build GRCh38
rs115649988 Chr.19: 41219132-41219132 on Build GRCh38
rs933967628 Chr.19: 41219209-41219209 on Build GRCh38
rs540801607 Chr.19: 41219370-41219370 on Build GRCh38
rs563994782 Chr.19: 41219452-41219452 on Build GRCh38
rs1031148650 Chr.19: 41219276-41219276 on Build GRCh38
rs566342269 Chr.19: 41219247-41219247 on Build GRCh38
rs369924410 Chr.19: 41219119-41219119 on Build GRCh38
rs746493416 Chr.19: 41219406-41219406 on Build GRCh38
rs1043985274 Chr.19: 41219468-41219468 on Build GRCh38
rs149864098 Chr.19: 41219398-41219398 on Build GRCh38
rs971941285 Chr.19: 41219561-41219561 on Build GRCh38
rs767309040 Chr.19: 41219343-41219344 on Build GRCh38
rs749007110 Chr.19: 41219517-41219517 on Build GRCh38
rs865972129 Chr.19: 41219298-41219298 on Build GRCh38
rs747483188 Chr.19: 41219351-41219351 on Build GRCh38
rs768319413 Chr.19: 41219410-41219410 on Build GRCh38
rs1022292235 Chr.19: 41219143-41219143 on Build GRCh38
rs373052453 Chr.19: 41219514-41219514 on Build GRCh38
rs10411012 Chr.19: 41219366-41219366 on Build GRCh38
rs780019975 Chr.19: 41219349-41219349 on Build GRCh38
rs555587960 Chr.19: 41219434-41219434 on Build GRCh38
rs761863392 Chr.19: 41219413-41219413 on Build GRCh38
rs909154631 Chr.19: 41219463-41219463 on Build GRCh38
rs536654645 Chr.19: 41219275-41219275 on Build GRCh38
rs746689495 Chr.19: 41219469-41219469 on Build GRCh38

View Full Product Details