Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41230991-41231393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001278599.1 NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130033 c.747G>C p.L249L Chr.19: 41231262-41231262 on Build GRCh38
6882738 c.717G>A p.T239T Chr.19: 41231232-41231232 on Build GRCh38
6970658 c.647_648CC>TT p.S216F Chr.19: 41231027-41231028 on Build GRCh38
6921563 c.637G>A p.V213I Chr.19: 41231017-41231017 on Build GRCh38
4131888 c.728T>G p.V243G Chr.19: 41231243-41231243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756064558 Chr.19: 41231269-41231269 on Build GRCh38
rs144284099 Chr.19: 41231016-41231016 on Build GRCh38
rs943268186 Chr.19: 41231285-41231285 on Build GRCh38
rs895881314 Chr.19: 41231350-41231350 on Build GRCh38
rs775805907 Chr.19: 41231224-41231224 on Build GRCh38
rs746497465 Chr.19: 41231151-41231151 on Build GRCh38
rs754490094 Chr.19: 41231262-41231262 on Build GRCh38
rs372520752 Chr.19: 41231346-41231346 on Build GRCh38
rs762318825 Chr.19: 41231241-41231241 on Build GRCh38
rs752514954 Chr.19: 41231274-41231274 on Build GRCh38
rs150914023 Chr.19: 41231265-41231265 on Build GRCh38
rs913599937 Chr.19: 41231229-41231229 on Build GRCh38
rs780756544 Chr.19: 41231264-41231264 on Build GRCh38
rs760971852 Chr.19: 41231329-41231329 on Build GRCh38
rs993568731 Chr.19: 41231332-41231332 on Build GRCh38
rs780952193 Chr.19: 41231165-41231165 on Build GRCh38
rs773248925 Chr.19: 41231076-41231076 on Build GRCh38
rs921250564 Chr.19: 41231072-41231072 on Build GRCh38
rs140448864 Chr.19: 41231200-41231200 on Build GRCh38
rs200206204 Chr.19: 41231315-41231315 on Build GRCh38
rs771116052 Chr.19: 41231201-41231201 on Build GRCh38
rs751189022 Chr.19: 41231327-41231327 on Build GRCh38
rs763166316 Chr.19: 41231077-41231077 on Build GRCh38
rs201678158 Chr.19: 41231236-41231236 on Build GRCh38
rs775637645 Chr.19: 41231303-41231303 on Build GRCh38
rs370295348 Chr.19: 41231143-41231143 on Build GRCh38
rs975407816 Chr.19: 41231036-41231036 on Build GRCh38
rs765721148 Chr.19: 41231319-41231319 on Build GRCh38
rs749981843 Chr.19: 41231147-41231147 on Build GRCh38
rs753274836 Chr.19: 41231090-41231090 on Build GRCh38
rs367872631 Chr.19: 41231150-41231150 on Build GRCh38
rs376363034 Chr.19: 41231311-41231311 on Build GRCh38
rs142452494 Chr.19: 41231232-41231232 on Build GRCh38
rs376785668 Chr.19: 41231295-41231295 on Build GRCh38
rs777619818 Chr.19: 41231277-41231277 on Build GRCh38
rs756382142 Chr.19: 41231091-41231091 on Build GRCh38
rs535396112 Chr.19: 41231121-41231121 on Build GRCh38
rs1026304928 Chr.19: 41231100-41231100 on Build GRCh38
rs945530203 Chr.19: 41231116-41231116 on Build GRCh38
rs372987501 Chr.19: 41231148-41231148 on Build GRCh38
rs773053200 Chr.19: 41231184-41231184 on Build GRCh38
rs200765254 Chr.19: 41231171-41231171 on Build GRCh38
rs762107336 Chr.19: 41231170-41231170 on Build GRCh38
rs779312028 Chr.19: 41231202-41231202 on Build GRCh38
rs760727969 Chr.19: 41231304-41231304 on Build GRCh38
rs766652458 Chr.19: 41231078-41231078 on Build GRCh38
rs774592224 Chr.19: 41231219-41231219 on Build GRCh38
rs749047403 Chr.19: 41231281-41231281 on Build GRCh38
rs1051415960 Chr.19: 41231266-41231266 on Build GRCh38
rs577354334 Chr.19: 41231298-41231298 on Build GRCh38
rs754575803 Chr.19: 41231152-41231152 on Build GRCh38
rs534638184 Chr.19: 41231103-41231103 on Build GRCh38
rs767132376 Chr.19: 41231336-41231336 on Build GRCh38
rs769662670 Chr.19: 41231053-41231053 on Build GRCh38
rs926104806 Chr.19: 41231158-41231158 on Build GRCh38
rs779718472 Chr.19: 41231008-41231008 on Build GRCh38
rs146774960 Chr.19: 41231101-41231101 on Build GRCh38
rs761251235 Chr.19: 41231231-41231231 on Build GRCh38
rs987881778 Chr.19: 41231217-41231217 on Build GRCh38
rs779055360 Chr.19: 41231289-41231289 on Build GRCh38
rs759658392 Chr.19: 41231081-41231081 on Build GRCh38
rs769184562 Chr.19: 41231308-41231308 on Build GRCh38
rs79266409 Chr.19: 41231112-41231112 on Build GRCh38
rs747864633 Chr.19: 41231040-41231040 on Build GRCh38
rs958023615 Chr.19: 41231337-41231337 on Build GRCh38
rs768702995 Chr.19: 41231029-41231029 on Build GRCh38
rs759570308 Chr.19: 41231221-41231221 on Build GRCh38
rs770739757 Chr.19: 41231287-41231287 on Build GRCh38
rs199702283 Chr.19: 41231339-41231339 on Build GRCh38
rs767654945 Chr.19: 41231222-41231222 on Build GRCh38
rs754244924 Chr.19: 41231240-41231240 on Build GRCh38
rs766099704 Chr.19: 41231256-41231256 on Build GRCh38
rs776682499 Chr.19: 41231030-41231030 on Build GRCh38
rs767979238 Chr.19: 41231083-41231083 on Build GRCh38
rs759219999 Chr.19: 41231335-41231335 on Build GRCh38
rs751290925 Chr.19: 41231259-41231259 on Build GRCh38
rs950685158 Chr.19: 41231246-41231246 on Build GRCh38

View Full Product Details