Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50511501-50512000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPDH
Entrez Gene ID
554235
Gene Name
aspartate dehydrogenase domain containing
Gene Aliases
-
UniGene
Hs.436338
Gene Chromosome Location
Chr.19: 50511600-50514690 on Build GRCh38
Transcripts
NM_001024656.2 NM_001114598.1

SNP Detail

SNP ID Chromosome Location
rs150781238 Chr.19: 50511855-50511855 on Build GRCh38
rs369037329 Chr.19: 50511712-50511712 on Build GRCh38
rs946513319 Chr.19: 50511929-50511929 on Build GRCh38
rs890313954 Chr.19: 50511600-50511600 on Build GRCh38
rs151254015 Chr.19: 50511746-50511746 on Build GRCh38
rs111234385 Chr.19: 50511542-50511542 on Build GRCh38
rs527493722 Chr.19: 50511603-50511603 on Build GRCh38
rs371676761 Chr.19: 50511783-50511783 on Build GRCh38
rs139130158 Chr.19: 50511870-50511870 on Build GRCh38
rs201593378 Chr.19: 50511749-50511749 on Build GRCh38
rs913683289 Chr.19: 50511619-50511619 on Build GRCh38
rs910304475 Chr.19: 50511957-50511957 on Build GRCh38
rs1043507631 Chr.19: 50511959-50511959 on Build GRCh38
rs1048712528 Chr.19: 50511630-50511630 on Build GRCh38
rs953479517 Chr.19: 50511867-50511867 on Build GRCh38
rs140473564 Chr.19: 50511748-50511748 on Build GRCh38
rs1024280568 Chr.19: 50511710-50511710 on Build GRCh38
rs994542963 Chr.19: 50511605-50511605 on Build GRCh38
rs989916982 Chr.19: 50511818-50511818 on Build GRCh38
rs772123865 Chr.19: 50511816-50511818 on Build GRCh38
rs888723092 Chr.19: 50511670-50511670 on Build GRCh38
rs946453616 Chr.19: 50511633-50511633 on Build GRCh38
rs911776824 Chr.19: 50511874-50511874 on Build GRCh38
rs952988657 Chr.19: 50511522-50511522 on Build GRCh38
rs566339613 Chr.19: 50511705-50511705 on Build GRCh38
rs944492715 Chr.19: 50511602-50511602 on Build GRCh38
rs781463340 Chr.19: 50511663-50511663 on Build GRCh38
rs919041531 Chr.19: 50511967-50511967 on Build GRCh38
rs955461976 Chr.19: 50511854-50511854 on Build GRCh38
rs752218853 Chr.19: 50511847-50511847 on Build GRCh38
rs777285502 Chr.19: 50511768-50511768 on Build GRCh38
rs897538136 Chr.19: 50511604-50511604 on Build GRCh38
rs935338476 Chr.19: 50511578-50511578 on Build GRCh38
rs944525289 Chr.19: 50511891-50511891 on Build GRCh38
rs982177118 Chr.19: 50511571-50511571 on Build GRCh38
rs74711311 Chr.19: 50511809-50511809 on Build GRCh38
rs535764210 Chr.19: 50511825-50511825 on Build GRCh38
rs367793901 Chr.19: 50511585-50511585 on Build GRCh38
rs562466381 Chr.19: 50511574-50511574 on Build GRCh38
rs554186481 Chr.19: 50511835-50511835 on Build GRCh38
rs1026226794 Chr.19: 50511781-50511781 on Build GRCh38
rs926336464 Chr.19: 50511975-50511975 on Build GRCh38
rs750506086 Chr.19: 50511821-50511821 on Build GRCh38
rs556748848 Chr.19: 50511776-50511776 on Build GRCh38
rs535890280 Chr.19: 50511926-50511929 on Build GRCh38
rs951534245 Chr.19: 50511808-50511808 on Build GRCh38
rs73062136 Chr.19: 50511842-50511842 on Build GRCh38
rs1036193263 Chr.19: 50511899-50511899 on Build GRCh38
rs923994608 Chr.19: 50511573-50511573 on Build GRCh38
rs7254387 Chr.19: 50511593-50511593 on Build GRCh38
rs892187788 Chr.19: 50511519-50511520 on Build GRCh38
rs576102421 Chr.19: 50511956-50511956 on Build GRCh38
rs202090659 Chr.19: 50511955-50511955 on Build GRCh38
rs769087319 Chr.19: 50511766-50511766 on Build GRCh38
rs984173574 Chr.19: 50511602-50511603 on Build GRCh38
rs913704050 Chr.19: 50511926-50511927 on Build GRCh38
rs1052662132 Chr.19: 50511615-50511615 on Build GRCh38
rs373662143 Chr.19: 50511537-50511537 on Build GRCh38
rs150431672 Chr.19: 50511752-50511752 on Build GRCh38
rs2308272 Chr.19: 50511950-50511950 on Build GRCh38
rs1023788999 Chr.19: 50511525-50511525 on Build GRCh38
rs570526474 Chr.19: 50511753-50511753 on Build GRCh38
rs113025783 Chr.19: 50511773-50511773 on Build GRCh38
rs1010578417 Chr.19: 50511520-50511520 on Build GRCh38
rs991426842 Chr.19: 50511523-50511523 on Build GRCh38
rs370786815 Chr.19: 50511945-50511945 on Build GRCh38
rs931905932 Chr.19: 50511587-50511587 on Build GRCh38
rs770529914 Chr.19: 50511729-50511729 on Build GRCh38
rs1043432790 Chr.19: 50511659-50511659 on Build GRCh38
rs56250057 Chr.19: 50511954-50511955 on Build GRCh38
rs146338297 Chr.19: 50511950-50511951 on Build GRCh38
rs970878995 Chr.19: 50511732-50511732 on Build GRCh38
rs529904861 Chr.19: 50511580-50511580 on Build GRCh38
rs964029426 Chr.19: 50511819-50511819 on Build GRCh38

View Full Product Details