Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27083052-27083576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN1
Entrez Gene ID
11117
Gene Name
elastin microfibril interfacer 1
Gene Aliases
EMI, EMILIN, gp115
UniGene
Hs.63348
Gene Chromosome Location
Chr.2: 27078567-27086403 on Build GRCh38
Transcripts
NM_007046.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019499 c.1780C>T p.R594C Chr.2: 27083351-27083351 on Build GRCh38
5465526 c.1898G>A p.G633E Chr.2: 27083469-27083469 on Build GRCh38
1019501 c.1921C>A p.Q641K Chr.2: 27083492-27083492 on Build GRCh38
6900923 c.1987G>A p.E663K Chr.2: 27083558-27083558 on Build GRCh38
3580501 c.1510G>A p.V504M Chr.2: 27083081-27083081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553403745 Chr.2: 27083399-27083399 on Build GRCh38
rs139944533 Chr.2: 27083339-27083339 on Build GRCh38
rs780673311 Chr.2: 27083544-27083544 on Build GRCh38
rs765535997 Chr.2: 27083498-27083498 on Build GRCh38
rs955650542 Chr.2: 27083231-27083231 on Build GRCh38
rs771549929 Chr.2: 27083312-27083312 on Build GRCh38
rs894151361 Chr.2: 27083084-27083084 on Build GRCh38
rs374940182 Chr.2: 27083119-27083119 on Build GRCh38
rs781500770 Chr.2: 27083189-27083189 on Build GRCh38
rs959396358 Chr.2: 27083340-27083340 on Build GRCh38
rs993771955 Chr.2: 27083362-27083362 on Build GRCh38
rs745565616 Chr.2: 27083372-27083372 on Build GRCh38
rs369967327 Chr.2: 27083346-27083346 on Build GRCh38
rs146042569 Chr.2: 27083127-27083127 on Build GRCh38
rs747577622 Chr.2: 27083117-27083117 on Build GRCh38
rs766052971 Chr.2: 27083138-27083138 on Build GRCh38
rs745504669 Chr.2: 27083309-27083309 on Build GRCh38
rs201549569 Chr.2: 27083534-27083534 on Build GRCh38
rs563926619 Chr.2: 27083106-27083106 on Build GRCh38
rs761704114 Chr.2: 27083402-27083402 on Build GRCh38
rs774000688 Chr.2: 27083205-27083205 on Build GRCh38
rs917814042 Chr.2: 27083366-27083366 on Build GRCh38
rs760787529 Chr.2: 27083466-27083466 on Build GRCh38
rs1024384968 Chr.2: 27083109-27083109 on Build GRCh38
rs534036052 Chr.2: 27083379-27083379 on Build GRCh38
rs754465328 Chr.2: 27083487-27083487 on Build GRCh38
rs372213065 Chr.2: 27083120-27083120 on Build GRCh38
rs569070507 Chr.2: 27083204-27083204 on Build GRCh38
rs748137973 Chr.2: 27083546-27083546 on Build GRCh38
rs769640832 Chr.2: 27083403-27083403 on Build GRCh38
rs765651075 Chr.2: 27083382-27083383 on Build GRCh38
rs753749516 Chr.2: 27083439-27083439 on Build GRCh38
rs748418310 Chr.2: 27083125-27083125 on Build GRCh38
rs758022469 Chr.2: 27083510-27083510 on Build GRCh38
rs373844330 Chr.2: 27083131-27083131 on Build GRCh38
rs11548683 Chr.2: 27083278-27083278 on Build GRCh38
rs759528710 Chr.2: 27083143-27083143 on Build GRCh38
rs756693825 Chr.2: 27083351-27083351 on Build GRCh38
rs778294240 Chr.2: 27083269-27083269 on Build GRCh38
rs766551959 Chr.2: 27083258-27083258 on Build GRCh38
rs752561430 Chr.2: 27083153-27083153 on Build GRCh38
rs138622931 Chr.2: 27083193-27083193 on Build GRCh38
rs753208706 Chr.2: 27083267-27083267 on Build GRCh38
rs759797521 Chr.2: 27083338-27083338 on Build GRCh38
rs757827777 Chr.2: 27083491-27083491 on Build GRCh38
rs759690238 Chr.2: 27083412-27083412 on Build GRCh38
rs776444866 Chr.2: 27083330-27083330 on Build GRCh38
rs770737371 Chr.2: 27083282-27083282 on Build GRCh38
rs775054889 Chr.2: 27083316-27083316 on Build GRCh38
rs868583812 Chr.2: 27083495-27083495 on Build GRCh38
rs749597209 Chr.2: 27083281-27083281 on Build GRCh38
rs763918677 Chr.2: 27083159-27083159 on Build GRCh38
rs767318349 Chr.2: 27083149-27083149 on Build GRCh38
rs773706697 Chr.2: 27083202-27083202 on Build GRCh38
rs750735646 Chr.2: 27083503-27083503 on Build GRCh38
rs541909854 Chr.2: 27083104-27083104 on Build GRCh38
rs1030348145 Chr.2: 27083484-27083484 on Build GRCh38
rs552485083 Chr.2: 27083130-27083130 on Build GRCh38
rs752203222 Chr.2: 27083373-27083373 on Build GRCh38
rs746068703 Chr.2: 27083110-27083110 on Build GRCh38
rs767033564 Chr.2: 27083219-27083219 on Build GRCh38
rs773984791 Chr.2: 27083142-27083142 on Build GRCh38
rs769659278 Chr.2: 27083552-27083552 on Build GRCh38
rs906833541 Chr.2: 27083356-27083356 on Build GRCh38
rs752751079 Chr.2: 27083453-27083453 on Build GRCh38
rs779711180 Chr.2: 27083515-27083515 on Build GRCh38
rs534735729 Chr.2: 27083105-27083105 on Build GRCh38
rs754961210 Chr.2: 27083100-27083100 on Build GRCh38
rs753576764 Chr.2: 27083247-27083247 on Build GRCh38
rs573167279 Chr.2: 27083333-27083333 on Build GRCh38
rs139678896 Chr.2: 27083232-27083232 on Build GRCh38
rs36069611 Chr.2: 27083178-27083178 on Build GRCh38
rs200339477 Chr.2: 27083352-27083352 on Build GRCh38
rs908618660 Chr.2: 27083096-27083096 on Build GRCh38
rs773043123 Chr.2: 27083411-27083411 on Build GRCh38
rs778758313 Chr.2: 27083304-27083304 on Build GRCh38
rs767710118 Chr.2: 27083452-27083452 on Build GRCh38
rs768517659 Chr.2: 27083328-27083328 on Build GRCh38
rs553033062 Chr.2: 27083486-27083486 on Build GRCh38
rs371170446 Chr.2: 27083154-27083154 on Build GRCh38
rs117801783 Chr.2: 27083182-27083182 on Build GRCh38
rs767621068 Chr.2: 27083357-27083357 on Build GRCh38
rs763235759 Chr.2: 27083179-27083179 on Build GRCh38
rs994137663 Chr.2: 27083129-27083129 on Build GRCh38
rs375599106 Chr.2: 27083163-27083163 on Build GRCh38
rs976135721 Chr.2: 27083177-27083177 on Build GRCh38
rs754519478 Chr.2: 27083538-27083538 on Build GRCh38
rs760545096 Chr.2: 27083318-27083318 on Build GRCh38
rs756629138 Chr.2: 27083268-27083268 on Build GRCh38
rs34487849 Chr.2: 27083164-27083164 on Build GRCh38
rs781297451 Chr.2: 27083266-27083266 on Build GRCh38
rs751767343 Chr.2: 27083335-27083335 on Build GRCh38
rs758626189 Chr.2: 27083251-27083251 on Build GRCh38
rs376032560 Chr.2: 27083400-27083400 on Build GRCh38
rs181718238 Chr.2: 27083152-27083152 on Build GRCh38
rs751586097 Chr.2: 27083076-27083076 on Build GRCh38
rs539237417 Chr.2: 27083218-27083218 on Build GRCh38
rs368718986 Chr.2: 27083116-27083116 on Build GRCh38
rs770092227 Chr.2: 27083197-27083197 on Build GRCh38
rs751584697 Chr.2: 27083263-27083263 on Build GRCh38
rs202157641 Chr.2: 27083112-27083112 on Build GRCh38
rs755537123 Chr.2: 27083183-27083183 on Build GRCh38
rs751658031 Chr.2: 27083262-27083262 on Build GRCh38
rs779417501 Chr.2: 27083358-27083358 on Build GRCh38
rs141929856 Chr.2: 27083246-27083246 on Build GRCh38

View Full Product Details