Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29193040-29193574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALK
Entrez Gene ID
238
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Gene Aliases
CD246, NBLST3
UniGene
Hs.654469
Gene Chromosome Location
Chr.2: 29192774-29921611 on Build GRCh38
Transcripts
NM_004304.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6509220 c.1087C>T p.L363L Chr.2: 29193490-29193490 on Build GRCh38
6986477 c.1303G>A p.E435K Chr.2: 29193274-29193274 on Build GRCh38
5719565 c.1041A>G p.K347K Chr.2: 29193536-29193536 on Build GRCh38
6962503 c.1243G>A p.G415S Chr.2: 29193334-29193334 on Build GRCh38
4495941 c.4640C>T p.T1547I Chr.2: 29193447-29193447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs57917930 Chr.2: 29193262-29193262 on Build GRCh38
rs753750497 Chr.2: 29193275-29193275 on Build GRCh38
rs779663817 Chr.2: 29193502-29193502 on Build GRCh38
rs1881423 Chr.2: 29193291-29193291 on Build GRCh38
rs773870096 Chr.2: 29193383-29193383 on Build GRCh38
rs748822593 Chr.2: 29193301-29193301 on Build GRCh38
rs752927575 Chr.2: 29193401-29193401 on Build GRCh38
rs78174819 Chr.2: 29193251-29193251 on Build GRCh38
rs757368506 Chr.2: 29193206-29193206 on Build GRCh38
rs552439944 Chr.2: 29193465-29193465 on Build GRCh38
rs746416960 Chr.2: 29193200-29193201 on Build GRCh38
rs764304191 Chr.2: 29193272-29193272 on Build GRCh38
rs1011355630 Chr.2: 29193501-29193501 on Build GRCh38
rs755556501 Chr.2: 29193512-29193514 on Build GRCh38
rs765866043 Chr.2: 29193328-29193328 on Build GRCh38
rs200715698 Chr.2: 29193164-29193164 on Build GRCh38
rs770217830 Chr.2: 29193367-29193367 on Build GRCh38
rs76150405 Chr.2: 29193302-29193302 on Build GRCh38
rs199947131 Chr.2: 29193159-29193159 on Build GRCh38
rs199646095 Chr.2: 29193441-29193441 on Build GRCh38
rs374733353 Chr.2: 29193550-29193550 on Build GRCh38
rs780454793 Chr.2: 29193280-29193280 on Build GRCh38
rs989939823 Chr.2: 29193167-29193167 on Build GRCh38
rs764298799 Chr.2: 29193489-29193489 on Build GRCh38
rs754214919 Chr.2: 29193274-29193274 on Build GRCh38
rs772461583 Chr.2: 29193219-29193219 on Build GRCh38
rs149694391 Chr.2: 29193542-29193542 on Build GRCh38
rs766019535 Chr.2: 29193180-29193180 on Build GRCh38
rs1033678538 Chr.2: 29193155-29193155 on Build GRCh38
rs186421480 Chr.2: 29193179-29193179 on Build GRCh38
rs1019065457 Chr.2: 29193397-29193397 on Build GRCh38
rs747161831 Chr.2: 29193283-29193283 on Build GRCh38
rs886055927 Chr.2: 29193158-29193159 on Build GRCh38
rs780241185 Chr.2: 29193509-29193510 on Build GRCh38
rs776898697 Chr.2: 29193309-29193309 on Build GRCh38
rs974930400 Chr.2: 29193402-29193402 on Build GRCh38
rs747946035 Chr.2: 29193344-29193344 on Build GRCh38
rs751872583 Chr.2: 29193407-29193407 on Build GRCh38
rs148351049 Chr.2: 29193364-29193364 on Build GRCh38
rs756452385 Chr.2: 29193182-29193182 on Build GRCh38
rs1008766442 Chr.2: 29193316-29193316 on Build GRCh38
rs780399982 Chr.2: 29193220-29193220 on Build GRCh38
rs772438558 Chr.2: 29193280-29193280 on Build GRCh38
rs397706189 Chr.2: 29193163-29193164 on Build GRCh38
rs190639819 Chr.2: 29193319-29193319 on Build GRCh38
rs200829376 Chr.2: 29193358-29193358 on Build GRCh38
rs551779946 Chr.2: 29193172-29193172 on Build GRCh38
rs768295861 Chr.2: 29193221-29193221 on Build GRCh38
rs368300433 Chr.2: 29193249-29193249 on Build GRCh38
rs75681494 Chr.2: 29193552-29193552 on Build GRCh38
rs893771860 Chr.2: 29193362-29193362 on Build GRCh38
rs1881421 Chr.2: 29193500-29193500 on Build GRCh38
rs78868998 Chr.2: 29193444-29193444 on Build GRCh38
rs987583019 Chr.2: 29193198-29193198 on Build GRCh38
rs780976256 Chr.2: 29193513-29193513 on Build GRCh38
rs878854659 Chr.2: 29193462-29193462 on Build GRCh38
rs759166553 Chr.2: 29193306-29193306 on Build GRCh38
rs745349865 Chr.2: 29193363-29193363 on Build GRCh38
rs763613910 Chr.2: 29193384-29193384 on Build GRCh38
rs867422810 Chr.2: 29193496-29193496 on Build GRCh38
rs373037272 Chr.2: 29193292-29193292 on Build GRCh38
rs747978928 Chr.2: 29193242-29193242 on Build GRCh38
rs147241767 Chr.2: 29193479-29193479 on Build GRCh38
rs778914972 Chr.2: 29193215-29193215 on Build GRCh38
rs201645782 Chr.2: 29193158-29193158 on Build GRCh38
rs902642850 Chr.2: 29193521-29193521 on Build GRCh38
rs537603430 Chr.2: 29193263-29193263 on Build GRCh38
rs753622072 Chr.2: 29193409-29193409 on Build GRCh38
rs912173028 Chr.2: 29193211-29193211 on Build GRCh38
rs370947313 Chr.2: 29193349-29193349 on Build GRCh38
rs768933017 Chr.2: 29193542-29193543 on Build GRCh38
rs778371605 Chr.2: 29193183-29193183 on Build GRCh38
rs141242925 Chr.2: 29193498-29193498 on Build GRCh38
rs563126725 Chr.2: 29193493-29193493 on Build GRCh38
rs758556216 Chr.2: 29193335-29193335 on Build GRCh38
rs534201503 Chr.2: 29193216-29193216 on Build GRCh38
rs571671456 Chr.2: 29193398-29193398 on Build GRCh38
rs1046265972 Chr.2: 29193477-29193477 on Build GRCh38
rs958335893 Chr.2: 29193478-29193478 on Build GRCh38
rs368100947 Chr.2: 29193173-29193173 on Build GRCh38
rs376702277 Chr.2: 29193286-29193286 on Build GRCh38
rs767659365 Chr.2: 29193399-29193399 on Build GRCh38
rs1048589952 Chr.2: 29193287-29193287 on Build GRCh38
rs35500880 Chr.2: 29193434-29193435 on Build GRCh38
rs774860532 Chr.2: 29193464-29193464 on Build GRCh38
rs139185626 Chr.2: 29193486-29193486 on Build GRCh38
rs952457982 Chr.2: 29193431-29193431 on Build GRCh38
rs751632696 Chr.2: 29193395-29193395 on Build GRCh38
rs148138515 Chr.2: 29193548-29193548 on Build GRCh38
rs771570579 Chr.2: 29193433-29193433 on Build GRCh38
rs1881422 Chr.2: 29193491-29193491 on Build GRCh38
rs773780435 Chr.2: 29193442-29193442 on Build GRCh38
rs776637020 Chr.2: 29193229-29193229 on Build GRCh38
rs767191161 Chr.2: 29193307-29193307 on Build GRCh38
rs761101687 Chr.2: 29193266-29193266 on Build GRCh38
rs766072801 Chr.2: 29193257-29193257 on Build GRCh38
rs150487874 Chr.2: 29193404-29193404 on Build GRCh38
rs766990201 Chr.2: 29193392-29193392 on Build GRCh38
rs752701859 Chr.2: 29193481-29193481 on Build GRCh38
rs577660779 Chr.2: 29193294-29193294 on Build GRCh38
rs1002240457 Chr.2: 29193118-29193118 on Build GRCh38
rs778797863 Chr.2: 29193428-29193428 on Build GRCh38
rs781448587 Chr.2: 29193340-29193340 on Build GRCh38
rs771352312 Chr.2: 29193522-29193522 on Build GRCh38
rs749162642 Chr.2: 29193353-29193353 on Build GRCh38
rs886055928 Chr.2: 29193439-29193439 on Build GRCh38
rs766508956 Chr.2: 29193338-29193338 on Build GRCh38
rs939750594 Chr.2: 29193467-29193467 on Build GRCh38
rs370701220 Chr.2: 29193304-29193305 on Build GRCh38
rs763403897 Chr.2: 29193455-29193455 on Build GRCh38
rs35364102 Chr.2: 29193262-29193263 on Build GRCh38
rs372440265 Chr.2: 29193276-29193276 on Build GRCh38
rs771622065 Chr.2: 29193460-29193460 on Build GRCh38
rs745390287 Chr.2: 29193430-29193430 on Build GRCh38
rs74669215 Chr.2: 29193113-29193113 on Build GRCh38
rs950584612 Chr.2: 29193084-29193084 on Build GRCh38
rs756989551 Chr.2: 29193490-29193490 on Build GRCh38
rs746470148 Chr.2: 29193503-29193503 on Build GRCh38
rs1021483040 Chr.2: 29193382-29193382 on Build GRCh38
rs372769158 Chr.2: 29193160-29193161 on Build GRCh38
rs751677905 Chr.2: 29193339-29193339 on Build GRCh38
rs774746810 Chr.2: 29193533-29193533 on Build GRCh38
rs878854658 Chr.2: 29193551-29193551 on Build GRCh38
rs750920504 Chr.2: 29193329-29193329 on Build GRCh38
rs754743349 Chr.2: 29193509-29193509 on Build GRCh38
rs764586398 Chr.2: 29193176-29193176 on Build GRCh38
rs369781538 Chr.2: 29193289-29193289 on Build GRCh38
rs200425614 Chr.2: 29193435-29193435 on Build GRCh38
rs750105816 Chr.2: 29193178-29193178 on Build GRCh38
rs542234332 Chr.2: 29193422-29193422 on Build GRCh38
rs374379545 Chr.2: 29193181-29193181 on Build GRCh38
rs1024874247 Chr.2: 29193063-29193063 on Build GRCh38
rs869185071 Chr.2: 29193164-29193165 on Build GRCh38
rs139437088 Chr.2: 29193514-29193514 on Build GRCh38
rs758502850 Chr.2: 29193278-29193278 on Build GRCh38
rs182561050 Chr.2: 29193161-29193161 on Build GRCh38
rs553333897 Chr.2: 29193350-29193350 on Build GRCh38
rs80088378 Chr.2: 29193171-29193171 on Build GRCh38

View Full Product Details