Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68463193-68463724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO48
Entrez Gene ID
554251
Gene Name
F-box protein 48
Gene Aliases
-
UniGene
Hs.164117
Gene Chromosome Location
Chr.2: 68459419-68467325 on Build GRCh38
Transcripts
NM_001024680.2 XM_005264407.2 XM_017004437.1

SNP Detail

SNP ID Chromosome Location
rs910110767 Chr.2: 68463469-68463469 on Build GRCh38
rs535924014 Chr.2: 68463435-68463435 on Build GRCh38
rs905251481 Chr.2: 68463230-68463230 on Build GRCh38
rs1052475916 Chr.2: 68463343-68463343 on Build GRCh38
rs1003787091 Chr.2: 68463269-68463269 on Build GRCh38
rs1010884572 Chr.2: 68463368-68463368 on Build GRCh38
rs1009484781 Chr.2: 68463285-68463285 on Build GRCh38
rs963375115 Chr.2: 68463621-68463621 on Build GRCh38
rs951658261 Chr.2: 68463442-68463442 on Build GRCh38
rs931812492 Chr.2: 68463222-68463222 on Build GRCh38
rs967692684 Chr.2: 68463303-68463303 on Build GRCh38
rs191422086 Chr.2: 68463626-68463626 on Build GRCh38
rs375401028 Chr.2: 68463221-68463221 on Build GRCh38
rs576585050 Chr.2: 68463505-68463505 on Build GRCh38
rs893812866 Chr.2: 68463346-68463346 on Build GRCh38
rs941381634 Chr.2: 68463482-68463482 on Build GRCh38
rs758857969 Chr.2: 68463499-68463502 on Build GRCh38
rs965790460 Chr.2: 68463381-68463381 on Build GRCh38
rs973060262 Chr.2: 68463638-68463638 on Build GRCh38
rs1034823688 Chr.2: 68463270-68463270 on Build GRCh38
rs541006324 Chr.2: 68463695-68463695 on Build GRCh38
rs982984473 Chr.2: 68463460-68463460 on Build GRCh38
rs905443619 Chr.2: 68463426-68463426 on Build GRCh38
rs941479258 Chr.2: 68463217-68463217 on Build GRCh38
rs572093064 Chr.2: 68463620-68463620 on Build GRCh38
rs1005050544 Chr.2: 68463584-68463584 on Build GRCh38
rs1045732115 Chr.2: 68463420-68463420 on Build GRCh38
rs186955425 Chr.2: 68463278-68463278 on Build GRCh38
rs953274850 Chr.2: 68463687-68463687 on Build GRCh38
rs113552873 Chr.2: 68463578-68463578 on Build GRCh38
rs935386820 Chr.2: 68463296-68463296 on Build GRCh38
rs570785151 Chr.2: 68463421-68463421 on Build GRCh38
rs113747004 Chr.2: 68463432-68463432 on Build GRCh38
rs921880454 Chr.2: 68463682-68463682 on Build GRCh38
rs985769985 Chr.2: 68463345-68463345 on Build GRCh38
rs541522132 Chr.2: 68463534-68463534 on Build GRCh38
rs1003706371 Chr.2: 68463440-68463440 on Build GRCh38
rs892280309 Chr.2: 68463615-68463615 on Build GRCh38
rs539776762 Chr.2: 68463459-68463459 on Build GRCh38
rs1051590747 Chr.2: 68463245-68463245 on Build GRCh38
rs553794127 Chr.2: 68463244-68463244 on Build GRCh38
rs1009514472 Chr.2: 68463652-68463652 on Build GRCh38
rs556312069 Chr.2: 68463463-68463463 on Build GRCh38

View Full Product Details