Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97660489-97660977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR1B
Entrez Gene ID
10120
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
Gene Aliases
ARP1B, CTRN2, PC3
UniGene
Hs.98791
Gene Chromosome Location
Chr.2: 97655939-97664098 on Build GRCh38
Transcripts
NM_005735.3 XM_005263854.4 XM_017003116.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181738 c.171C>A p.F57L Chr.2: 97660589-97660589 on Build GRCh38
6092728 c.161G>T p.G54V Chr.2: 97660599-97660599 on Build GRCh38
6593089 c.137G>T p.R46L Chr.2: 97660623-97660623 on Build GRCh38
4659167 c.118G>A p.G40R Chr.2: 97660642-97660642 on Build GRCh38
380951 c.156G>A p.L52L Chr.2: 97660604-97660604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771251640 Chr.2: 97660553-97660553 on Build GRCh38
rs1031109814 Chr.2: 97660898-97660898 on Build GRCh38
rs200290427 Chr.2: 97660665-97660665 on Build GRCh38
rs752709785 Chr.2: 97660567-97660567 on Build GRCh38
rs940469954 Chr.2: 97660809-97660809 on Build GRCh38
rs969721141 Chr.2: 97660752-97660752 on Build GRCh38
rs925447333 Chr.2: 97660797-97660797 on Build GRCh38
rs556203737 Chr.2: 97660870-97660870 on Build GRCh38
rs916235657 Chr.2: 97660746-97660746 on Build GRCh38
rs369909921 Chr.2: 97660673-97660673 on Build GRCh38
rs896269042 Chr.2: 97660907-97660907 on Build GRCh38
rs779437666 Chr.2: 97660525-97660525 on Build GRCh38
rs149669662 Chr.2: 97660959-97660959 on Build GRCh38
rs1001444121 Chr.2: 97660712-97660712 on Build GRCh38
rs750883687 Chr.2: 97660725-97660725 on Build GRCh38
rs368997121 Chr.2: 97660588-97660588 on Build GRCh38
rs992634971 Chr.2: 97660521-97660521 on Build GRCh38
rs368304448 Chr.2: 97660806-97660806 on Build GRCh38
rs772064883 Chr.2: 97660534-97660534 on Build GRCh38
rs1051549691 Chr.2: 97660834-97660834 on Build GRCh38
rs909015218 Chr.2: 97660535-97660535 on Build GRCh38
rs755333171 Chr.2: 97660680-97660682 on Build GRCh38
rs759788778 Chr.2: 97660638-97660638 on Build GRCh38
rs758820949 Chr.2: 97660833-97660833 on Build GRCh38
rs113670297 Chr.2: 97660568-97660568 on Build GRCh38
rs890064747 Chr.2: 97660623-97660623 on Build GRCh38
rs904107120 Chr.2: 97660886-97660886 on Build GRCh38
rs774134186 Chr.2: 97660658-97660658 on Build GRCh38
rs543980691 Chr.2: 97660887-97660887 on Build GRCh38
rs896856040 Chr.2: 97660855-97660855 on Build GRCh38
rs765589704 Chr.2: 97660678-97660678 on Build GRCh38
rs1037078357 Chr.2: 97660562-97660562 on Build GRCh38
rs769730826 Chr.2: 97660639-97660639 on Build GRCh38
rs889770533 Chr.2: 97660846-97660846 on Build GRCh38
rs759450084 Chr.2: 97660563-97660563 on Build GRCh38
rs554129227 Chr.2: 97660705-97660705 on Build GRCh38
rs779201184 Chr.2: 97660618-97660618 on Build GRCh38
rs1049395477 Chr.2: 97660509-97660509 on Build GRCh38
rs146383048 Chr.2: 97660634-97660634 on Build GRCh38
rs770889575 Chr.2: 97660629-97660629 on Build GRCh38
rs574664110 Chr.2: 97660928-97660928 on Build GRCh38
rs918700123 Chr.2: 97660826-97660826 on Build GRCh38
rs985119730 Chr.2: 97660956-97660956 on Build GRCh38
rs775360822 Chr.2: 97660643-97660643 on Build GRCh38
rs546571572 Chr.2: 97660523-97660523 on Build GRCh38
rs749711921 Chr.2: 97660520-97660520 on Build GRCh38
rs942949422 Chr.2: 97660857-97660857 on Build GRCh38
rs979536630 Chr.2: 97660764-97660764 on Build GRCh38
rs559931806 Chr.2: 97660529-97660529 on Build GRCh38
rs992256685 Chr.2: 97660730-97660730 on Build GRCh38
rs377679983 Chr.2: 97660657-97660657 on Build GRCh38
rs763519581 Chr.2: 97660585-97660585 on Build GRCh38
rs780871635 Chr.2: 97660595-97660595 on Build GRCh38
rs750047083 Chr.2: 97660615-97660615 on Build GRCh38
rs554514432 Chr.2: 97660908-97660908 on Build GRCh38
rs772561765 Chr.2: 97660624-97660624 on Build GRCh38
rs532612317 Chr.2: 97660704-97660704 on Build GRCh38
rs913954280 Chr.2: 97660743-97660743 on Build GRCh38
rs201783746 Chr.2: 97660682-97660682 on Build GRCh38
rs73959765 Chr.2: 97660664-97660664 on Build GRCh38
rs747519549 Chr.2: 97660577-97660577 on Build GRCh38
rs960440645 Chr.2: 97660724-97660724 on Build GRCh38
rs1013307414 Chr.2: 97660910-97660910 on Build GRCh38
rs1036196489 Chr.2: 97660819-97660819 on Build GRCh38
rs76631457 Chr.2: 97660586-97660586 on Build GRCh38
rs761426929 Chr.2: 97660663-97660663 on Build GRCh38
rs748665342 Chr.2: 97660621-97660621 on Build GRCh38
rs1001400126 Chr.2: 97660905-97660905 on Build GRCh38
rs1043923132 Chr.2: 97660883-97660883 on Build GRCh38
rs576785765 Chr.2: 97660782-97660782 on Build GRCh38
rs1048205345 Chr.2: 97660865-97660866 on Build GRCh38
rs755368866 Chr.2: 97660616-97660616 on Build GRCh38
rs576163992 Chr.2: 97660877-97660877 on Build GRCh38
rs765090171 Chr.2: 97660564-97660564 on Build GRCh38
rs992187170 Chr.2: 97660733-97660733 on Build GRCh38
rs7574212 Chr.2: 97660518-97660518 on Build GRCh38
rs781661707 Chr.2: 97660690-97660691 on Build GRCh38
rs544285515 Chr.2: 97660929-97660929 on Build GRCh38
rs1026820557 Chr.2: 97660674-97660674 on Build GRCh38
rs761537643 Chr.2: 97660875-97660875 on Build GRCh38
rs895754571 Chr.2: 97660916-97660916 on Build GRCh38
rs538409935 Chr.2: 97660656-97660656 on Build GRCh38
rs79616802 Chr.2: 97660627-97660627 on Build GRCh38
rs376789745 Chr.2: 97660552-97660552 on Build GRCh38
rs201782728 Chr.2: 97660545-97660545 on Build GRCh38

View Full Product Details