Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33675395-33675900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

SNP Detail

SNP ID Chromosome Location
rs1011422750 Chr.20: 33675666-33675666 on Build GRCh38
rs1006693496 Chr.20: 33675810-33675810 on Build GRCh38
rs932389124 Chr.20: 33675806-33675806 on Build GRCh38
rs1037274503 Chr.20: 33675643-33675643 on Build GRCh38
rs3213182 Chr.20: 33675427-33675427 on Build GRCh38
rs943691466 Chr.20: 33675556-33675556 on Build GRCh38
rs184907289 Chr.20: 33675661-33675661 on Build GRCh38
rs968637591 Chr.20: 33675493-33675493 on Build GRCh38
rs976348221 Chr.20: 33675799-33675799 on Build GRCh38
rs771540812 Chr.20: 33675813-33675813 on Build GRCh38
rs906746170 Chr.20: 33675868-33675868 on Build GRCh38
rs1029485868 Chr.20: 33675487-33675487 on Build GRCh38
rs571403469 Chr.20: 33675632-33675633 on Build GRCh38
rs1033685103 Chr.20: 33675660-33675661 on Build GRCh38
rs148012006 Chr.20: 33675612-33675612 on Build GRCh38
rs914399000 Chr.20: 33675490-33675490 on Build GRCh38
rs548876497 Chr.20: 33675766-33675766 on Build GRCh38
rs543924348 Chr.20: 33675603-33675603 on Build GRCh38
rs554955048 Chr.20: 33675547-33675547 on Build GRCh38
rs1043474023 Chr.20: 33675443-33675443 on Build GRCh38
rs374135299 Chr.20: 33675686-33675686 on Build GRCh38
rs983565510 Chr.20: 33675853-33675853 on Build GRCh38
rs999271400 Chr.20: 33675789-33675789 on Build GRCh38
rs576514217 Chr.20: 33675579-33675579 on Build GRCh38
rs946388437 Chr.20: 33675735-33675735 on Build GRCh38
rs906818208 Chr.20: 33675647-33675647 on Build GRCh38
rs1043341295 Chr.20: 33675742-33675742 on Build GRCh38
rs1021433012 Chr.20: 33675677-33675677 on Build GRCh38
rs1003543913 Chr.20: 33675655-33675655 on Build GRCh38
rs764974197 Chr.20: 33675582-33675582 on Build GRCh38
rs764531025 Chr.20: 33675831-33675831 on Build GRCh38
rs768138130 Chr.20: 33675730-33675730 on Build GRCh38
rs924553480 Chr.20: 33675549-33675549 on Build GRCh38
rs1007241903 Chr.20: 33675571-33675571 on Build GRCh38
rs999323662 Chr.20: 33675476-33675476 on Build GRCh38
rs1048601126 Chr.20: 33675807-33675807 on Build GRCh38
rs1040718566 Chr.20: 33675565-33675565 on Build GRCh38
rs902181943 Chr.20: 33675747-33675747 on Build GRCh38
rs991301448 Chr.20: 33675445-33675445 on Build GRCh38
rs147293274 Chr.20: 33675829-33675829 on Build GRCh38
rs532967057 Chr.20: 33675621-33675621 on Build GRCh38
rs3213181 Chr.20: 33675618-33675618 on Build GRCh38
rs902232750 Chr.20: 33675450-33675450 on Build GRCh38
rs1055800789 Chr.20: 33675692-33675692 on Build GRCh38
rs143964465 Chr.20: 33675674-33675674 on Build GRCh38
rs1018177970 Chr.20: 33675580-33675580 on Build GRCh38
rs973375567 Chr.20: 33675599-33675599 on Build GRCh38
rs921009857 Chr.20: 33675589-33675589 on Build GRCh38
rs965085931 Chr.20: 33675592-33675592 on Build GRCh38
rs1050782648 Chr.20: 33675790-33675790 on Build GRCh38
rs983526358 Chr.20: 33675622-33675622 on Build GRCh38
rs545154397 Chr.20: 33675650-33675650 on Build GRCh38
rs3213180 Chr.20: 33675818-33675818 on Build GRCh38
rs552309524 Chr.20: 33675795-33675795 on Build GRCh38
rs759317377 Chr.20: 33675555-33675555 on Build GRCh38
rs994846387 Chr.20: 33675825-33675825 on Build GRCh38
rs920935605 Chr.20: 33675804-33675804 on Build GRCh38
rs898373033 Chr.20: 33675822-33675822 on Build GRCh38
rs916817427 Chr.20: 33675726-33675726 on Build GRCh38
rs181206924 Chr.20: 33675488-33675488 on Build GRCh38

View Full Product Details