Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33676960-33677450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458312 c.1080C>T p.S360S Chr.20: 33676966-33676966 on Build GRCh38
2757812 c.1004C>A p.P335Q Chr.20: 33677167-33677167 on Build GRCh38
4567679 c.819_820CC>TT p.Q274* Chr.20: 33677446-33677447 on Build GRCh38
6092803 c.916A>G p.I306V Chr.20: 33677255-33677255 on Build GRCh38
7295931 c.895C>T p.P299S Chr.20: 33677276-33677276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200825658 Chr.20: 33677006-33677006 on Build GRCh38
rs766615337 Chr.20: 33676990-33676990 on Build GRCh38
rs538498751 Chr.20: 33677094-33677095 on Build GRCh38
rs6057877 Chr.20: 33677090-33677090 on Build GRCh38
rs747336438 Chr.20: 33677059-33677059 on Build GRCh38
rs764704208 Chr.20: 33677283-33677283 on Build GRCh38
rs762920298 Chr.20: 33677004-33677004 on Build GRCh38
rs145759508 Chr.20: 33677135-33677135 on Build GRCh38
rs145231543 Chr.20: 33677057-33677057 on Build GRCh38
rs759041493 Chr.20: 33677280-33677280 on Build GRCh38
rs771807428 Chr.20: 33677140-33677140 on Build GRCh38
rs111268294 Chr.20: 33677160-33677160 on Build GRCh38
rs554294623 Chr.20: 33677265-33677265 on Build GRCh38
rs371744736 Chr.20: 33677408-33677408 on Build GRCh38
rs761517377 Chr.20: 33677019-33677019 on Build GRCh38
rs749348639 Chr.20: 33677218-33677218 on Build GRCh38
rs774135926 Chr.20: 33677007-33677007 on Build GRCh38
rs149272498 Chr.20: 33677168-33677168 on Build GRCh38
rs371719878 Chr.20: 33677337-33677337 on Build GRCh38
rs3213175 Chr.20: 33677042-33677042 on Build GRCh38
rs760562883 Chr.20: 33677250-33677250 on Build GRCh38
rs570268126 Chr.20: 33677093-33677093 on Build GRCh38
rs747031927 Chr.20: 33677161-33677163 on Build GRCh38
rs1001002474 Chr.20: 33677017-33677017 on Build GRCh38
rs752029737 Chr.20: 33677156-33677156 on Build GRCh38
rs372399012 Chr.20: 33677056-33677056 on Build GRCh38
rs756501085 Chr.20: 33677313-33677313 on Build GRCh38
rs267605892 Chr.20: 33677292-33677292 on Build GRCh38
rs776058128 Chr.20: 33677208-33677210 on Build GRCh38
rs903640215 Chr.20: 33677014-33677014 on Build GRCh38
rs375154514 Chr.20: 33677420-33677420 on Build GRCh38
rs775166611 Chr.20: 33677376-33677380 on Build GRCh38
rs775614866 Chr.20: 33677118-33677118 on Build GRCh38
rs367892498 Chr.20: 33677402-33677402 on Build GRCh38
rs190923948 Chr.20: 33677354-33677354 on Build GRCh38
rs776052569 Chr.20: 33676990-33676990 on Build GRCh38
rs777860071 Chr.20: 33677361-33677361 on Build GRCh38
rs548772156 Chr.20: 33677071-33677071 on Build GRCh38
rs778884599 Chr.20: 33677236-33677236 on Build GRCh38
rs755049757 Chr.20: 33677342-33677342 on Build GRCh38
rs770856342 Chr.20: 33677166-33677168 on Build GRCh38
rs764579715 Chr.20: 33677376-33677385 on Build GRCh38
rs755659970 Chr.20: 33677024-33677024 on Build GRCh38
rs553413048 Chr.20: 33677201-33677201 on Build GRCh38
rs551050661 Chr.20: 33677375-33677376 on Build GRCh38
rs750641087 Chr.20: 33677170-33677170 on Build GRCh38
rs750718703 Chr.20: 33677307-33677307 on Build GRCh38
rs138360482 Chr.20: 33677154-33677154 on Build GRCh38
rs374841514 Chr.20: 33677020-33677020 on Build GRCh38
rs3213174 Chr.20: 33677239-33677239 on Build GRCh38
rs764181474 Chr.20: 33677429-33677429 on Build GRCh38
rs369639454 Chr.20: 33677101-33677101 on Build GRCh38
rs888128272 Chr.20: 33677328-33677328 on Build GRCh38
rs201117611 Chr.20: 33677407-33677407 on Build GRCh38
rs774185605 Chr.20: 33677136-33677136 on Build GRCh38
rs1023370024 Chr.20: 33677012-33677012 on Build GRCh38
rs992581181 Chr.20: 33677389-33677389 on Build GRCh38
rs866556043 Chr.20: 33677327-33677327 on Build GRCh38
rs757620294 Chr.20: 33677159-33677159 on Build GRCh38
rs776326343 Chr.20: 33677376-33677376 on Build GRCh38
rs878891941 Chr.20: 33677393-33677393 on Build GRCh38
rs773197012 Chr.20: 33677249-33677249 on Build GRCh38
rs182175727 Chr.20: 33677034-33677034 on Build GRCh38
rs752075116 Chr.20: 33677289-33677289 on Build GRCh38
rs759073647 Chr.20: 33677238-33677264 on Build GRCh38
rs753343055 Chr.20: 33677151-33677151 on Build GRCh38
rs575839838 Chr.20: 33677295-33677295 on Build GRCh38
rs765976516 Chr.20: 33677144-33677144 on Build GRCh38
rs745547146 Chr.20: 33677377-33677377 on Build GRCh38
rs144481343 Chr.20: 33677203-33677203 on Build GRCh38
rs944228695 Chr.20: 33677238-33677238 on Build GRCh38
rs867122904 Chr.20: 33677296-33677296 on Build GRCh38
rs993029218 Chr.20: 33677129-33677129 on Build GRCh38
rs374753083 Chr.20: 33677374-33677374 on Build GRCh38
rs564825532 Chr.20: 33677143-33677143 on Build GRCh38
rs535573433 Chr.20: 33677245-33677245 on Build GRCh38
rs769885322 Chr.20: 33676989-33676989 on Build GRCh38
rs183250565 Chr.20: 33677430-33677430 on Build GRCh38
rs559212050 Chr.20: 33677047-33677047 on Build GRCh38
rs143729545 Chr.20: 33677391-33677391 on Build GRCh38
rs368151951 Chr.20: 33677028-33677028 on Build GRCh38
rs746765090 Chr.20: 33677362-33677362 on Build GRCh38
rs1030566220 Chr.20: 33677049-33677049 on Build GRCh38
rs762296081 Chr.20: 33677291-33677291 on Build GRCh38
rs952341140 Chr.20: 33677061-33677061 on Build GRCh38
rs770643353 Chr.20: 33677264-33677264 on Build GRCh38
rs1031106445 Chr.20: 33677302-33677302 on Build GRCh38
rs754052244 Chr.20: 33677341-33677341 on Build GRCh38
rs764626563 Chr.20: 33677155-33677155 on Build GRCh38
rs960021941 Chr.20: 33677386-33677386 on Build GRCh38
rs748100436 Chr.20: 33677348-33677348 on Build GRCh38
rs764834039 Chr.20: 33677332-33677335 on Build GRCh38
rs768640915 Chr.20: 33677234-33677234 on Build GRCh38
rs761616906 Chr.20: 33677137-33677137 on Build GRCh38
rs373179329 Chr.20: 33677142-33677142 on Build GRCh38
rs574956843 Chr.20: 33677278-33677278 on Build GRCh38
rs749340138 Chr.20: 33677123-33677123 on Build GRCh38
rs778924828 Chr.20: 33677345-33677345 on Build GRCh38

View Full Product Details