Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33677237-33677668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4915933 c.815A>T p.Q272L Chr.20: 33677451-33677451 on Build GRCh38
3545486 c.880G>A p.D294N Chr.20: 33677291-33677291 on Build GRCh38
6651007 c.741G>A p.T247T Chr.20: 33677525-33677525 on Build GRCh38
4567679 c.819_820CC>TT p.Q274* Chr.20: 33677446-33677447 on Build GRCh38
6092803 c.916A>G p.I306V Chr.20: 33677255-33677255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888128272 Chr.20: 33677328-33677328 on Build GRCh38
rs775238660 Chr.20: 33677556-33677556 on Build GRCh38
rs765506787 Chr.20: 33677482-33677482 on Build GRCh38
rs201117611 Chr.20: 33677407-33677407 on Build GRCh38
rs767969405 Chr.20: 33677431-33677431 on Build GRCh38
rs764704208 Chr.20: 33677283-33677283 on Build GRCh38
rs369957716 Chr.20: 33677559-33677559 on Build GRCh38
rs892252132 Chr.20: 33677547-33677548 on Build GRCh38
rs992581181 Chr.20: 33677389-33677389 on Build GRCh38
rs866556043 Chr.20: 33677327-33677327 on Build GRCh38
rs776326343 Chr.20: 33677376-33677376 on Build GRCh38
rs878891941 Chr.20: 33677393-33677393 on Build GRCh38
rs1003887445 Chr.20: 33677623-33677623 on Build GRCh38
rs552816539 Chr.20: 33677592-33677592 on Build GRCh38
rs759041493 Chr.20: 33677280-33677280 on Build GRCh38
rs1008237224 Chr.20: 33677612-33677612 on Build GRCh38
rs376645897 Chr.20: 33677521-33677521 on Build GRCh38
rs773673658 Chr.20: 33677566-33677566 on Build GRCh38
rs913895798 Chr.20: 33677526-33677526 on Build GRCh38
rs752075116 Chr.20: 33677289-33677289 on Build GRCh38
rs35807460 Chr.20: 33677525-33677525 on Build GRCh38
rs755210188 Chr.20: 33677469-33677469 on Build GRCh38
rs748745091 Chr.20: 33677558-33677558 on Build GRCh38
rs373383167 Chr.20: 33677505-33677505 on Build GRCh38
rs575839838 Chr.20: 33677295-33677295 on Build GRCh38
rs554294623 Chr.20: 33677265-33677265 on Build GRCh38
rs768151708 Chr.20: 33677584-33677587 on Build GRCh38
rs371744736 Chr.20: 33677408-33677408 on Build GRCh38
rs3213173 Chr.20: 33677440-33677440 on Build GRCh38
rs745547146 Chr.20: 33677377-33677377 on Build GRCh38
rs1052123927 Chr.20: 33677534-33677534 on Build GRCh38
rs754082291 Chr.20: 33677462-33677462 on Build GRCh38
rs888659779 Chr.20: 33677622-33677622 on Build GRCh38
rs529768589 Chr.20: 33677441-33677441 on Build GRCh38
rs371719878 Chr.20: 33677337-33677337 on Build GRCh38
rs187191843 Chr.20: 33677628-33677628 on Build GRCh38
rs867122904 Chr.20: 33677296-33677296 on Build GRCh38
rs917803687 Chr.20: 33677460-33677460 on Build GRCh38
rs374753083 Chr.20: 33677374-33677374 on Build GRCh38
rs761234206 Chr.20: 33677575-33677575 on Build GRCh38
rs3213172 Chr.20: 33677511-33677511 on Build GRCh38
rs756501085 Chr.20: 33677313-33677313 on Build GRCh38
rs183250565 Chr.20: 33677430-33677430 on Build GRCh38
rs267605892 Chr.20: 33677292-33677292 on Build GRCh38
rs777679476 Chr.20: 33677645-33677645 on Build GRCh38
rs375154514 Chr.20: 33677420-33677420 on Build GRCh38
rs143729545 Chr.20: 33677391-33677391 on Build GRCh38
rs746765090 Chr.20: 33677362-33677362 on Build GRCh38
rs775166611 Chr.20: 33677376-33677380 on Build GRCh38
rs34468325 Chr.20: 33677489-33677489 on Build GRCh38
rs367892498 Chr.20: 33677402-33677402 on Build GRCh38
rs762296081 Chr.20: 33677291-33677291 on Build GRCh38
rs190923948 Chr.20: 33677354-33677354 on Build GRCh38
rs770643353 Chr.20: 33677264-33677264 on Build GRCh38
rs777860071 Chr.20: 33677361-33677361 on Build GRCh38
rs1031106445 Chr.20: 33677302-33677302 on Build GRCh38
rs372963946 Chr.20: 33677564-33677564 on Build GRCh38
rs777808893 Chr.20: 33677504-33677504 on Build GRCh38
rs141063030 Chr.20: 33677531-33677531 on Build GRCh38
rs754052244 Chr.20: 33677341-33677341 on Build GRCh38
rs755049757 Chr.20: 33677342-33677342 on Build GRCh38
rs960021941 Chr.20: 33677386-33677386 on Build GRCh38
rs762315411 Chr.20: 33677579-33677579 on Build GRCh38
rs748100436 Chr.20: 33677348-33677348 on Build GRCh38
rs764579715 Chr.20: 33677376-33677385 on Build GRCh38
rs764834039 Chr.20: 33677332-33677335 on Build GRCh38
rs551050661 Chr.20: 33677375-33677376 on Build GRCh38
rs1048697969 Chr.20: 33677595-33677595 on Build GRCh38
rs750718703 Chr.20: 33677307-33677307 on Build GRCh38
rs758585438 Chr.20: 33677500-33677500 on Build GRCh38
rs764181474 Chr.20: 33677429-33677429 on Build GRCh38
rs574956843 Chr.20: 33677278-33677278 on Build GRCh38
rs541956472 Chr.20: 33677450-33677450 on Build GRCh38
rs778924828 Chr.20: 33677345-33677345 on Build GRCh38

View Full Product Details