Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44332943-44333451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf2
Entrez Gene ID
755
Gene Name
chromosome 21 open reading frame 2
Gene Aliases
LRRC76, YF5/A2
UniGene
Hs.517331
Gene Chromosome Location
Chr.21: 44328944-44339417 on Build GRCh38
Transcripts
NM_001271440.1 NM_001271441.1 NM_001271442.1 NM_004928.2 XM_006724051.3 XM_006724053.3 XM_017028470.1 XM_017028471.1 XM_017028472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358408 c.209G>T p.R70L Chr.21: 44333197-44333197 on Build GRCh38
4800256 c.336G>A p.L112L Chr.21: 44333070-44333070 on Build GRCh38
4754219 c.228C>A p.S76R Chr.21: 44333178-44333178 on Build GRCh38
4602181 c.224C>T p.P75L Chr.21: 44333182-44333182 on Build GRCh38
1291190 c.261G>A p.P87P Chr.21: 44333145-44333145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371211181 Chr.21: 44333177-44333177 on Build GRCh38
rs975687717 Chr.21: 44332980-44332980 on Build GRCh38
rs368674927 Chr.21: 44333124-44333124 on Build GRCh38
rs775668894 Chr.21: 44333292-44333292 on Build GRCh38
rs1021869493 Chr.21: 44332974-44332974 on Build GRCh38
rs750760441 Chr.21: 44332992-44332992 on Build GRCh38
rs755673924 Chr.21: 44333162-44333162 on Build GRCh38
rs922930539 Chr.21: 44333059-44333059 on Build GRCh38
rs377539637 Chr.21: 44333407-44333407 on Build GRCh38
rs76435115 Chr.21: 44333390-44333390 on Build GRCh38
rs567435284 Chr.21: 44333137-44333137 on Build GRCh38
rs368928576 Chr.21: 44333287-44333287 on Build GRCh38
rs759884906 Chr.21: 44333218-44333218 on Build GRCh38
rs368095791 Chr.21: 44332984-44332984 on Build GRCh38
rs778987436 Chr.21: 44333182-44333182 on Build GRCh38
rs1001751336 Chr.21: 44333093-44333093 on Build GRCh38
rs750809604 Chr.21: 44333071-44333071 on Build GRCh38
rs376705985 Chr.21: 44333106-44333106 on Build GRCh38
rs202065452 Chr.21: 44333214-44333214 on Build GRCh38
rs755424242 Chr.21: 44333084-44333084 on Build GRCh38
rs751035924 Chr.21: 44333089-44333089 on Build GRCh38
rs1028502037 Chr.21: 44333116-44333116 on Build GRCh38
rs762918001 Chr.21: 44333063-44333063 on Build GRCh38
rs775496851 Chr.21: 44333057-44333057 on Build GRCh38
rs377185498 Chr.21: 44333153-44333153 on Build GRCh38
rs555164150 Chr.21: 44333075-44333075 on Build GRCh38
rs762159564 Chr.21: 44333068-44333068 on Build GRCh38
rs942969042 Chr.21: 44332972-44332972 on Build GRCh38
rs531036991 Chr.21: 44333017-44333017 on Build GRCh38
rs575547314 Chr.21: 44333312-44333312 on Build GRCh38
rs774614339 Chr.21: 44333120-44333120 on Build GRCh38
rs745430992 Chr.21: 44333067-44333067 on Build GRCh38
rs767756871 Chr.21: 44333069-44333069 on Build GRCh38
rs1047074357 Chr.21: 44333369-44333369 on Build GRCh38
rs952792135 Chr.21: 44333262-44333262 on Build GRCh38
rs543148803 Chr.21: 44333363-44333363 on Build GRCh38
rs762116812 Chr.21: 44333106-44333107 on Build GRCh38
rs549536131 Chr.21: 44333095-44333095 on Build GRCh38
rs1057518441 Chr.21: 44333224-44333224 on Build GRCh38
rs749261239 Chr.21: 44333113-44333113 on Build GRCh38
rs756627311 Chr.21: 44332993-44332993 on Build GRCh38
rs780887106 Chr.21: 44333105-44333105 on Build GRCh38
rs760843025 Chr.21: 44333221-44333222 on Build GRCh38
rs774927073 Chr.21: 44333104-44333105 on Build GRCh38
rs975081399 Chr.21: 44333307-44333307 on Build GRCh38
rs777946402 Chr.21: 44333270-44333270 on Build GRCh38
rs574215463 Chr.21: 44333099-44333099 on Build GRCh38
rs780892823 Chr.21: 44333039-44333039 on Build GRCh38
rs374507502 Chr.21: 44333130-44333130 on Build GRCh38
rs1057356386 Chr.21: 44333229-44333229 on Build GRCh38
rs533273002 Chr.21: 44333425-44333425 on Build GRCh38
rs73907174 Chr.21: 44333385-44333385 on Build GRCh38
rs767796576 Chr.21: 44332991-44332991 on Build GRCh38
rs901339107 Chr.21: 44333149-44333150 on Build GRCh38
rs760747981 Chr.21: 44333132-44333132 on Build GRCh38
rs747034539 Chr.21: 44333277-44333277 on Build GRCh38
rs770527417 Chr.21: 44333033-44333033 on Build GRCh38
rs140451304 Chr.21: 44333188-44333188 on Build GRCh38
rs141427556 Chr.21: 44333009-44333009 on Build GRCh38
rs371653272 Chr.21: 44333022-44333022 on Build GRCh38
rs188379680 Chr.21: 44333308-44333308 on Build GRCh38
rs377552361 Chr.21: 44333189-44333189 on Build GRCh38
rs781390226 Chr.21: 44333280-44333280 on Build GRCh38
rs185111364 Chr.21: 44333298-44333298 on Build GRCh38
rs752698486 Chr.21: 44333268-44333268 on Build GRCh38
rs75087725 Chr.21: 44333234-44333234 on Build GRCh38
rs745656075 Chr.21: 44333046-44333046 on Build GRCh38
rs151236255 Chr.21: 44333090-44333090 on Build GRCh38
rs756791423 Chr.21: 44333154-44333154 on Build GRCh38
rs891927048 Chr.21: 44332968-44332968 on Build GRCh38
rs902909024 Chr.21: 44333018-44333018 on Build GRCh38
rs763933450 Chr.21: 44333351-44333351 on Build GRCh38
rs909115679 Chr.21: 44333055-44333055 on Build GRCh38
rs988484468 Chr.21: 44333337-44333337 on Build GRCh38
rs1038025763 Chr.21: 44333311-44333311 on Build GRCh38
rs77970521 Chr.21: 44333420-44333420 on Build GRCh38
rs766585233 Chr.21: 44333076-44333076 on Build GRCh38
rs773361360 Chr.21: 44333207-44333208 on Build GRCh38
rs947037010 Chr.21: 44333008-44333008 on Build GRCh38
rs772829403 Chr.21: 44333295-44333295 on Build GRCh38
rs199617249 Chr.21: 44333161-44333161 on Build GRCh38
rs775984914 Chr.21: 44333219-44333219 on Build GRCh38
rs754351767 Chr.21: 44333083-44333083 on Build GRCh38
rs755945783 Chr.21: 44333112-44333112 on Build GRCh38
rs771203756 Chr.21: 44333278-44333278 on Build GRCh38
rs780027455 Chr.21: 44333169-44333169 on Build GRCh38
rs771024688 Chr.21: 44333197-44333197 on Build GRCh38
rs896067502 Chr.21: 44333406-44333406 on Build GRCh38
rs752269920 Chr.21: 44333010-44333010 on Build GRCh38
rs111434965 Chr.21: 44333391-44333391 on Build GRCh38
rs201576990 Chr.21: 44333114-44333114 on Build GRCh38
rs9976610 Chr.21: 44333056-44333056 on Build GRCh38
rs746027663 Chr.21: 44333286-44333286 on Build GRCh38
rs766874630 Chr.21: 44333159-44333159 on Build GRCh38
rs938437742 Chr.21: 44333254-44333254 on Build GRCh38
rs767869403 Chr.21: 44333117-44333121 on Build GRCh38
rs947121368 Chr.21: 44333367-44333367 on Build GRCh38
rs927457782 Chr.21: 44333389-44333389 on Build GRCh38
rs754823280 Chr.21: 44333253-44333253 on Build GRCh38
rs950577318 Chr.21: 44333131-44333131 on Build GRCh38
rs964493051 Chr.21: 44333302-44333302 on Build GRCh38
rs748224537 Chr.21: 44333121-44333121 on Build GRCh38
rs954564347 Chr.21: 44333329-44333329 on Build GRCh38
rs960406567 Chr.21: 44333002-44333002 on Build GRCh38
rs201278533 Chr.21: 44333066-44333066 on Build GRCh38
rs922950652 Chr.21: 44333144-44333144 on Build GRCh38
rs750105410 Chr.21: 44333160-44333160 on Build GRCh38
rs11552070 Chr.21: 44333126-44333126 on Build GRCh38
rs192880944 Chr.21: 44333023-44333023 on Build GRCh38
rs763623409 Chr.21: 44333087-44333087 on Build GRCh38
rs1056901460 Chr.21: 44333271-44333271 on Build GRCh38
rs995480793 Chr.21: 44333356-44333356 on Build GRCh38
rs755022406 Chr.21: 44333179-44333179 on Build GRCh38
rs553278564 Chr.21: 44333146-44333146 on Build GRCh38
rs769039082 Chr.21: 44333060-44333071 on Build GRCh38
rs753752452 Chr.21: 44333247-44333247 on Build GRCh38
rs778791287 Chr.21: 44333259-44333259 on Build GRCh38
rs769014966 Chr.21: 44333294-44333294 on Build GRCh38
rs760097234 Chr.21: 44333301-44333301 on Build GRCh38
rs200049699 Chr.21: 44333228-44333228 on Build GRCh38
rs760073135 Chr.21: 44333143-44333143 on Build GRCh38
rs758224762 Chr.21: 44333269-44333269 on Build GRCh38

View Full Product Details