Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100749233-100749771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI3BP
Entrez Gene ID
25890
Gene Name
ABI family member 3 binding protein
Gene Aliases
NESHBP, TARSH
UniGene
Hs.477015
Gene Chromosome Location
Chr.3: 100749335-100993508 on Build GRCh38
Transcripts
NM_015429.3 XM_005247281.2 XM_005247282.2 XM_005247283.2 XM_005247284.2 XM_005247285.2 XM_005247287.2 XM_005247288.2 XM_005247289.2 XM_005247290.2 XM_005247291.2 XM_005247293.2 XM_005247294.2 XM_005247295.2 XM_005247296.2 XM_005247298.2 XM_005247299.3 XM_005247300.2 XM_005247301.2 XM_005247302.2 XM_005247303.2 XM_005247305.2 XM_005247306.2 XM_005247309.1 XM_006713569.2 XM_006713570.2 XM_006713571.2 XM_006713572.2 XM_011512639.1 XM_011512640.1 XM_011512641.1 XM_011512643.1 XM_011512644.1 XM_011512645.1 XM_011512646.1 XM_011512647.1 XM_011512648.1 XM_011512649.1 XM_011512651.1 XM_011512652.1 XM_011512653.1 XM_011512654.1 XM_011512656.1 XM_011512657.1 XM_011512658.1 XM_011512659.1 XM_011512660.1 XM_017006105.1 XM_017006106.1 XM_017006107.1 XM_017006108.1

SNP Detail

SNP ID Chromosome Location
rs560968115 Chr.3: 100749539-100749540 on Build GRCh38
rs978669625 Chr.3: 100749320-100749322 on Build GRCh38
rs16334 Chr.3: 100749563-100749564 on Build GRCh38
rs942710014 Chr.3: 100749609-100749609 on Build GRCh38
rs895931577 Chr.3: 100749311-100749311 on Build GRCh38
rs573536899 Chr.3: 100749611-100749612 on Build GRCh38
rs918763456 Chr.3: 100749591-100749591 on Build GRCh38
rs947694166 Chr.3: 100749337-100749337 on Build GRCh38
rs980495412 Chr.3: 100749353-100749353 on Build GRCh38
rs554488770 Chr.3: 100749454-100749454 on Build GRCh38
rs182554215 Chr.3: 100749378-100749378 on Build GRCh38
rs975103397 Chr.3: 100749326-100749326 on Build GRCh38
rs71625569 Chr.3: 100749567-100749568 on Build GRCh38
rs71132531 Chr.3: 100749262-100749263 on Build GRCh38
rs1042203001 Chr.3: 100749395-100749395 on Build GRCh38
rs1032913963 Chr.3: 100749407-100749407 on Build GRCh38
rs1027704 Chr.3: 100749586-100749586 on Build GRCh38
rs908001313 Chr.3: 100749337-100749341 on Build GRCh38
rs3057219 Chr.3: 100749535-100749538 on Build GRCh38
rs1039712633 Chr.3: 100749610-100749610 on Build GRCh38
rs139406979 Chr.3: 100749742-100749742 on Build GRCh38
rs549260284 Chr.3: 100749421-100749422 on Build GRCh38
rs888454029 Chr.3: 100749427-100749427 on Build GRCh38
rs564550727 Chr.3: 100749630-100749630 on Build GRCh38
rs1010702694 Chr.3: 100749697-100749697 on Build GRCh38
rs541196098 Chr.3: 100749504-100749504 on Build GRCh38
rs397690887 Chr.3: 100749568-100749569 on Build GRCh38
rs1055747273 Chr.3: 100749373-100749374 on Build GRCh38
rs914906954 Chr.3: 100749324-100749324 on Build GRCh38
rs752525957 Chr.3: 100749358-100749358 on Build GRCh38
rs565573002 Chr.3: 100749398-100749398 on Build GRCh38
rs373031555 Chr.3: 100749664-100749664 on Build GRCh38
rs1006857775 Chr.3: 100749458-100749458 on Build GRCh38
rs1020156765 Chr.3: 100749318-100749319 on Build GRCh38
rs543315711 Chr.3: 100749569-100749570 on Build GRCh38
rs553317103 Chr.3: 100749528-100749528 on Build GRCh38
rs1057220230 Chr.3: 100749384-100749384 on Build GRCh38
rs5851222 Chr.3: 100749525-100749528 on Build GRCh38
rs998291438 Chr.3: 100749631-100749631 on Build GRCh38
rs574928819 Chr.3: 100749578-100749578 on Build GRCh38
rs116584266 Chr.3: 100749649-100749649 on Build GRCh38
rs533495818 Chr.3: 100749337-100749337 on Build GRCh38
rs367708432 Chr.3: 100749488-100749488 on Build GRCh38
rs565331756 Chr.3: 100749730-100749730 on Build GRCh38
rs373574068 Chr.3: 100749257-100749257 on Build GRCh38
rs150914812 Chr.3: 100749600-100749600 on Build GRCh38
rs181136155 Chr.3: 100749587-100749587 on Build GRCh38
rs989815452 Chr.3: 100749493-100749493 on Build GRCh38
rs545390226 Chr.3: 100749345-100749347 on Build GRCh38
rs542985602 Chr.3: 100749527-100749527 on Build GRCh38
rs1014485409 Chr.3: 100749312-100749313 on Build GRCh38
rs185165011 Chr.3: 100749403-100749403 on Build GRCh38
rs976581162 Chr.3: 100749274-100749274 on Build GRCh38
rs781227660 Chr.3: 100749539-100749539 on Build GRCh38
rs1048364 Chr.3: 100749656-100749656 on Build GRCh38
rs937634717 Chr.3: 100749496-100749496 on Build GRCh38
rs895887079 Chr.3: 100749374-100749374 on Build GRCh38
rs530817674 Chr.3: 100749399-100749399 on Build GRCh38
rs989182839 Chr.3: 100749322-100749322 on Build GRCh38
rs199973526 Chr.3: 100749273-100749274 on Build GRCh38
rs962459539 Chr.3: 100749604-100749604 on Build GRCh38
rs955221739 Chr.3: 100749432-100749432 on Build GRCh38
rs901251712 Chr.3: 100749474-100749474 on Build GRCh38
rs747399919 Chr.3: 100749747-100749747 on Build GRCh38
rs868220830 Chr.3: 100749402-100749402 on Build GRCh38
rs1022674250 Chr.3: 100749703-100749703 on Build GRCh38
rs71719432 Chr.3: 100749564-100749565 on Build GRCh38
rs956823732 Chr.3: 100749289-100749289 on Build GRCh38
rs555538704 Chr.3: 100749316-100749319 on Build GRCh38
rs760699918 Chr.3: 100749528-100749529 on Build GRCh38
rs937330225 Chr.3: 100749355-100749355 on Build GRCh38
rs66530346 Chr.3: 100749258-100749259 on Build GRCh38
rs958874327 Chr.3: 100749615-100749615 on Build GRCh38
rs922326997 Chr.3: 100749331-100749332 on Build GRCh38
rs386397410 Chr.3: 100749565-100749566 on Build GRCh38
rs72928695 Chr.3: 100749598-100749598 on Build GRCh38
rs963976819 Chr.3: 100749409-100749409 on Build GRCh38
rs549630297 Chr.3: 100749477-100749477 on Build GRCh38
rs536809745 Chr.3: 100749487-100749487 on Build GRCh38
rs145209312 Chr.3: 100749405-100749406 on Build GRCh38
rs5851223 Chr.3: 100749584-100749587 on Build GRCh38
rs201734724 Chr.3: 100749315-100749316 on Build GRCh38
rs759517780 Chr.3: 100749524-100749525 on Build GRCh38
rs1048406 Chr.3: 100749562-100749562 on Build GRCh38
rs200206121 Chr.3: 100749257-100749258 on Build GRCh38
rs1030953807 Chr.3: 100749275-100749275 on Build GRCh38
rs969027541 Chr.3: 100749513-100749513 on Build GRCh38
rs547345487 Chr.3: 100749380-100749380 on Build GRCh38
rs4061093 Chr.3: 100749586-100749589 on Build GRCh38
rs560548416 Chr.3: 100749348-100749349 on Build GRCh38
rs1034368606 Chr.3: 100749310-100749310 on Build GRCh38
rs1018411892 Chr.3: 100749469-100749469 on Build GRCh38
rs869050457 Chr.3: 100749345-100749345 on Build GRCh38
rs201230953 Chr.3: 100749259-100749259 on Build GRCh38
rs969270264 Chr.3: 100749719-100749719 on Build GRCh38
rs36170845 Chr.3: 100749510-100749511 on Build GRCh38
rs150308181 Chr.3: 100749311-100749312 on Build GRCh38
rs202007036 Chr.3: 100749263-100749263 on Build GRCh38
rs536302010 Chr.3: 100749601-100749601 on Build GRCh38
rs1029362291 Chr.3: 100749427-100749428 on Build GRCh38
rs761969042 Chr.3: 100749319-100749320 on Build GRCh38
rs879155784 Chr.3: 100749559-100749560 on Build GRCh38
rs2126541 Chr.3: 100749273-100749273 on Build GRCh38
rs67862637 Chr.3: 100749274-100749275 on Build GRCh38
rs139253819 Chr.3: 100749425-100749426 on Build GRCh38
rs927677086 Chr.3: 100749364-100749364 on Build GRCh38
rs571283315 Chr.3: 100749346-100749346 on Build GRCh38
rs1001507087 Chr.3: 100749307-100749307 on Build GRCh38
rs927750826 Chr.3: 100749535-100749535 on Build GRCh38
rs530998240 Chr.3: 100749259-100749260 on Build GRCh38
rs1027705 Chr.3: 100749489-100749489 on Build GRCh38
rs899742197 Chr.3: 100749586-100749587 on Build GRCh38
rs1048612199 Chr.3: 100749415-100749415 on Build GRCh38

View Full Product Details