Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129315771-129316268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H1FX
Entrez Gene ID
8971
Gene Name
H1 histone family member X
Gene Aliases
H1.10, H1X
UniGene
Hs.75307
Gene Chromosome Location
Chr.3: 129314771-129316277 on Build GRCh38
Transcripts
NM_006026.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209252 c.91T>C p.S31P Chr.3: 129315812-129315812 on Build GRCh38
4639151 c.18G>A p.E6E Chr.3: 129315885-129315885 on Build GRCh38
1172932 c.16G>C p.E6Q Chr.3: 129315887-129315887 on Build GRCh38
7370615 c.10G>T p.E4* Chr.3: 129315893-129315893 on Build GRCh38
116070 c.64A>C p.T22P Chr.3: 129315839-129315839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009262540 Chr.3: 129316168-129316168 on Build GRCh38
rs940721950 Chr.3: 129316048-129316048 on Build GRCh38
rs569041413 Chr.3: 129316186-129316186 on Build GRCh38
rs148496535 Chr.3: 129315823-129315823 on Build GRCh38
rs551925974 Chr.3: 129316120-129316120 on Build GRCh38
rs984344595 Chr.3: 129316193-129316193 on Build GRCh38
rs201051457 Chr.3: 129315890-129315890 on Build GRCh38
rs563400918 Chr.3: 129315966-129315966 on Build GRCh38
rs980632511 Chr.3: 129315949-129315949 on Build GRCh38
rs779085535 Chr.3: 129315945-129315945 on Build GRCh38
rs752124513 Chr.3: 129315816-129315816 on Build GRCh38
rs150299913 Chr.3: 129315828-129315828 on Build GRCh38
rs758161539 Chr.3: 129315797-129315799 on Build GRCh38
rs933581647 Chr.3: 129316237-129316237 on Build GRCh38
rs765977489 Chr.3: 129315801-129315801 on Build GRCh38
rs541322886 Chr.3: 129315896-129315896 on Build GRCh38
rs967040408 Chr.3: 129315849-129315849 on Build GRCh38
rs11537834 Chr.3: 129316136-129316136 on Build GRCh38
rs754154984 Chr.3: 129315939-129315939 on Build GRCh38
rs372867471 Chr.3: 129316233-129316233 on Build GRCh38
rs370057680 Chr.3: 129315851-129315851 on Build GRCh38
rs756494435 Chr.3: 129315919-129315919 on Build GRCh38
rs746781043 Chr.3: 129315875-129315875 on Build GRCh38
rs980651940 Chr.3: 129316180-129316180 on Build GRCh38
rs984865564 Chr.3: 129315976-129315976 on Build GRCh38
rs14323 Chr.3: 129316188-129316188 on Build GRCh38
rs753275312 Chr.3: 129315806-129315806 on Build GRCh38
rs1050557705 Chr.3: 129316129-129316129 on Build GRCh38
rs142987251 Chr.3: 129316087-129316087 on Build GRCh38
rs1024637797 Chr.3: 129316170-129316170 on Build GRCh38
rs892189383 Chr.3: 129316161-129316161 on Build GRCh38
rs759057153 Chr.3: 129315809-129315809 on Build GRCh38
rs541919742 Chr.3: 129315965-129315965 on Build GRCh38
rs111660250 Chr.3: 129315867-129315867 on Build GRCh38
rs766706789 Chr.3: 129315924-129315924 on Build GRCh38
rs13438 Chr.3: 129316178-129316178 on Build GRCh38
rs757542462 Chr.3: 129315818-129315818 on Build GRCh38
rs375731165 Chr.3: 129315841-129315841 on Build GRCh38
rs939955378 Chr.3: 129315978-129315978 on Build GRCh38
rs932501128 Chr.3: 129316064-129316064 on Build GRCh38
rs568278431 Chr.3: 129316149-129316149 on Build GRCh38
rs755085596 Chr.3: 129315944-129315944 on Build GRCh38
rs902137991 Chr.3: 129316079-129316079 on Build GRCh38
rs773020278 Chr.3: 129315863-129315863 on Build GRCh38
rs748144767 Chr.3: 129315857-129315857 on Build GRCh38
rs909226146 Chr.3: 129316035-129316035 on Build GRCh38
rs558807659 Chr.3: 129315829-129315829 on Build GRCh38
rs760325269 Chr.3: 129315799-129315799 on Build GRCh38
rs1041253036 Chr.3: 129315987-129315987 on Build GRCh38
rs562618010 Chr.3: 129316185-129316185 on Build GRCh38
rs758288851 Chr.3: 129315954-129315954 on Build GRCh38
rs937432852 Chr.3: 129315971-129315971 on Build GRCh38
rs922441576 Chr.3: 129315995-129315995 on Build GRCh38
rs552717915 Chr.3: 129316239-129316239 on Build GRCh38
rs775005336 Chr.3: 129315907-129315907 on Build GRCh38
rs1032067767 Chr.3: 129316152-129316152 on Build GRCh38
rs908421481 Chr.3: 129315927-129315927 on Build GRCh38
rs909327049 Chr.3: 129316205-129316205 on Build GRCh38
rs977590245 Chr.3: 129316228-129316228 on Build GRCh38
rs748207658 Chr.3: 129315951-129315951 on Build GRCh38
rs575204959 Chr.3: 129315827-129315827 on Build GRCh38
rs553326631 Chr.3: 129316249-129316249 on Build GRCh38
rs1001035078 Chr.3: 129316140-129316140 on Build GRCh38
rs1036307636 Chr.3: 129316071-129316071 on Build GRCh38
rs776320908 Chr.3: 129315888-129315888 on Build GRCh38
rs564798063 Chr.3: 129316123-129316123 on Build GRCh38
rs1049821333 Chr.3: 129316092-129316092 on Build GRCh38
rs6795995 Chr.3: 129315915-129315915 on Build GRCh38
rs910728935 Chr.3: 129316247-129316247 on Build GRCh38
rs868668280 Chr.3: 129315993-129315993 on Build GRCh38
rs150684541 Chr.3: 129316133-129316133 on Build GRCh38
rs749366252 Chr.3: 129315832-129315832 on Build GRCh38
rs888903100 Chr.3: 129315813-129315813 on Build GRCh38
rs147849516 Chr.3: 129315810-129315810 on Build GRCh38
rs370917641 Chr.3: 129315916-129315916 on Build GRCh38
rs557823733 Chr.3: 129316219-129316219 on Build GRCh38
rs770554907 Chr.3: 129315879-129315879 on Build GRCh38
rs111249161 Chr.3: 129316248-129316248 on Build GRCh38
rs923236310 Chr.3: 129316234-129316234 on Build GRCh38

View Full Product Details