Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130566623-130567124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A6
Entrez Gene ID
131873
Gene Name
collagen type VI alpha 6
Gene Aliases
-
UniGene
Hs.591282
Gene Chromosome Location
Chr.3: 130516709-130678135 on Build GRCh38
Transcripts
NM_001102608.1 XM_005247121.4 XM_011512423.2 XM_011512424.2 XM_011512425.2 XM_011512426.2 XM_011512428.2 XM_011512431.2 XM_011512432.2 XM_017005711.1 XM_017005712.1 XM_017005713.1 XM_017005714.1 XM_017005715.1 XM_017005716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237938 c.1554A>G p.A518A Chr.3: 130566973-130566973 on Build GRCh38
6282634 c.1598G>A p.R533Q Chr.3: 130567017-130567017 on Build GRCh38
4113460 c.1503G>A p.K501K Chr.3: 130566922-130566922 on Build GRCh38
4896971 c.1647C>T p.S549S Chr.3: 130567066-130567066 on Build GRCh38
5444650 c.1395G>A p.M465I Chr.3: 130566814-130566814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780921807 Chr.3: 130566942-130566942 on Build GRCh38
rs551912849 Chr.3: 130566950-130566950 on Build GRCh38
rs778201799 Chr.3: 130566905-130566905 on Build GRCh38
rs11921769 Chr.3: 130566801-130566801 on Build GRCh38
rs745557112 Chr.3: 130566961-130566961 on Build GRCh38
rs763062555 Chr.3: 130567017-130567017 on Build GRCh38
rs752810157 Chr.3: 130566812-130566820 on Build GRCh38
rs771149039 Chr.3: 130566988-130566988 on Build GRCh38
rs1020649956 Chr.3: 130566981-130566981 on Build GRCh38
rs764160244 Chr.3: 130567023-130567023 on Build GRCh38
rs759747410 Chr.3: 130567009-130567009 on Build GRCh38
rs201682544 Chr.3: 130566839-130566839 on Build GRCh38
rs768246772 Chr.3: 130566658-130566659 on Build GRCh38
rs755821962 Chr.3: 130567044-130567044 on Build GRCh38
rs899824706 Chr.3: 130566809-130566809 on Build GRCh38
rs200095455 Chr.3: 130566662-130566662 on Build GRCh38
rs373909064 Chr.3: 130566742-130566742 on Build GRCh38
rs779001805 Chr.3: 130566778-130566778 on Build GRCh38
rs753943734 Chr.3: 130566870-130566870 on Build GRCh38
rs750228240 Chr.3: 130567040-130567040 on Build GRCh38
rs931417794 Chr.3: 130567062-130567062 on Build GRCh38
rs995600884 Chr.3: 130566721-130566721 on Build GRCh38
rs756964248 Chr.3: 130566934-130566934 on Build GRCh38
rs1039829413 Chr.3: 130566802-130566802 on Build GRCh38
rs746722067 Chr.3: 130566686-130566686 on Build GRCh38
rs774968132 Chr.3: 130566840-130566840 on Build GRCh38
rs771930771 Chr.3: 130566674-130566674 on Build GRCh38
rs751376141 Chr.3: 130566912-130566912 on Build GRCh38
rs765546572 Chr.3: 130567010-130567010 on Build GRCh38
rs1004100910 Chr.3: 130566821-130566821 on Build GRCh38
rs199669411 Chr.3: 130566818-130566818 on Build GRCh38
rs957927770 Chr.3: 130566952-130566952 on Build GRCh38
rs114818609 Chr.3: 130566842-130566842 on Build GRCh38
rs777747583 Chr.3: 130566789-130566789 on Build GRCh38
rs767928089 Chr.3: 130566743-130566743 on Build GRCh38
rs765475073 Chr.3: 130566891-130566891 on Build GRCh38
rs181863743 Chr.3: 130567055-130567055 on Build GRCh38
rs747661431 Chr.3: 130567020-130567021 on Build GRCh38
rs760901421 Chr.3: 130566850-130566850 on Build GRCh38
rs1037851640 Chr.3: 130567003-130567003 on Build GRCh38
rs371953457 Chr.3: 130566968-130566968 on Build GRCh38
rs200975034 Chr.3: 130566990-130566990 on Build GRCh38
rs766489465 Chr.3: 130566764-130566764 on Build GRCh38
rs931343998 Chr.3: 130566812-130566812 on Build GRCh38
rs946178422 Chr.3: 130566740-130566740 on Build GRCh38
rs375571886 Chr.3: 130566984-130566984 on Build GRCh38
rs36113896 Chr.3: 130566949-130566950 on Build GRCh38
rs376855447 Chr.3: 130567090-130567090 on Build GRCh38
rs764691137 Chr.3: 130566739-130566739 on Build GRCh38
rs748799802 Chr.3: 130567047-130567047 on Build GRCh38
rs754994346 Chr.3: 130566661-130566661 on Build GRCh38
rs370634287 Chr.3: 130566772-130566772 on Build GRCh38
rs776921608 Chr.3: 130567070-130567070 on Build GRCh38
rs371128941 Chr.3: 130566820-130566820 on Build GRCh38
rs758918919 Chr.3: 130566719-130566719 on Build GRCh38
rs368961411 Chr.3: 130567060-130567060 on Build GRCh38
rs769406385 Chr.3: 130566963-130566963 on Build GRCh38
rs368134547 Chr.3: 130566967-130566967 on Build GRCh38
rs367786873 Chr.3: 130566848-130566848 on Build GRCh38
rs563018482 Chr.3: 130566667-130566667 on Build GRCh38
rs746754743 Chr.3: 130566790-130566790 on Build GRCh38
rs747514989 Chr.3: 130567064-130567064 on Build GRCh38
rs773523160 Chr.3: 130566970-130566970 on Build GRCh38
rs143108657 Chr.3: 130567039-130567039 on Build GRCh38
rs906366900 Chr.3: 130566894-130566894 on Build GRCh38
rs776250966 Chr.3: 130566694-130566694 on Build GRCh38
rs369313731 Chr.3: 130567098-130567098 on Build GRCh38
rs753975650 Chr.3: 130566769-130566769 on Build GRCh38
rs374174918 Chr.3: 130566898-130566898 on Build GRCh38
rs770456089 Chr.3: 130566690-130566690 on Build GRCh38
rs759679482 Chr.3: 130566871-130566871 on Build GRCh38
rs749555717 Chr.3: 130566996-130566996 on Build GRCh38
rs376463408 Chr.3: 130566744-130566744 on Build GRCh38
rs59021909 Chr.3: 130567085-130567085 on Build GRCh38
rs777798650 Chr.3: 130566900-130566900 on Build GRCh38
rs371268873 Chr.3: 130566892-130566892 on Build GRCh38
rs780840044 Chr.3: 130566998-130566999 on Build GRCh38
rs189434004 Chr.3: 130566965-130566965 on Build GRCh38
rs760999441 Chr.3: 130566763-130566763 on Build GRCh38
rs1025226709 Chr.3: 130566726-130566726 on Build GRCh38
rs751405197 Chr.3: 130567035-130567035 on Build GRCh38
rs977562851 Chr.3: 130566751-130566751 on Build GRCh38
rs775728744 Chr.3: 130567016-130567016 on Build GRCh38
rs374279156 Chr.3: 130566788-130566788 on Build GRCh38
rs747991460 Chr.3: 130566664-130566664 on Build GRCh38
rs773017155 Chr.3: 130566679-130566679 on Build GRCh38
rs766657514 Chr.3: 130566851-130566851 on Build GRCh38
rs112472535 Chr.3: 130567036-130567036 on Build GRCh38
rs771433710 Chr.3: 130567066-130567066 on Build GRCh38

View Full Product Details