Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149740337-149740833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD2
Entrez Gene ID
51122
Gene Name
COMM domain containing 2
Gene Aliases
HSPC042
UniGene
Hs.432729
Gene Chromosome Location
Chr.3: 149737160-149752507 on Build GRCh38
Transcripts
NM_016094.3 NR_135149.1

SNP Detail

SNP ID Chromosome Location
rs1021131069 Chr.3: 149740733-149740733 on Build GRCh38
rs540810328 Chr.3: 149740594-149740596 on Build GRCh38
rs941952449 Chr.3: 149740517-149740517 on Build GRCh38
rs1032564421 Chr.3: 149740757-149740757 on Build GRCh38
rs900426920 Chr.3: 149740548-149740548 on Build GRCh38
rs900704938 Chr.3: 149740390-149740390 on Build GRCh38
rs766808538 Chr.3: 149740514-149740514 on Build GRCh38
rs966828712 Chr.3: 149740738-149740738 on Build GRCh38
rs1037542528 Chr.3: 149740532-149740532 on Build GRCh38
rs567640407 Chr.3: 149740742-149740742 on Build GRCh38
rs966641849 Chr.3: 149740761-149740761 on Build GRCh38
rs979720605 Chr.3: 149740359-149740359 on Build GRCh38
rs925528428 Chr.3: 149740377-149740377 on Build GRCh38
rs927006150 Chr.3: 149740581-149740581 on Build GRCh38
rs747588465 Chr.3: 149740453-149740454 on Build GRCh38
rs547788533 Chr.3: 149740657-149740657 on Build GRCh38
rs1015236741 Chr.3: 149740520-149740520 on Build GRCh38
rs765391187 Chr.3: 149740502-149740502 on Build GRCh38
rs949525159 Chr.3: 149740463-149740463 on Build GRCh38
rs933505260 Chr.3: 149740393-149740393 on Build GRCh38
rs917983713 Chr.3: 149740461-149740461 on Build GRCh38
rs890198223 Chr.3: 149740605-149740605 on Build GRCh38
rs750117747 Chr.3: 149740476-149740476 on Build GRCh38
rs995829088 Chr.3: 149740570-149740570 on Build GRCh38
rs1051928021 Chr.3: 149740432-149740432 on Build GRCh38
rs751990458 Chr.3: 149740542-149740542 on Build GRCh38
rs759640079 Chr.3: 149740420-149740420 on Build GRCh38
rs868792320 Chr.3: 149740612-149740612 on Build GRCh38
rs117586858 Chr.3: 149740387-149740387 on Build GRCh38
rs1013433386 Chr.3: 149740724-149740724 on Build GRCh38
rs879903297 Chr.3: 149740703-149740704 on Build GRCh38
rs1054082 Chr.3: 149740801-149740801 on Build GRCh38
rs533050769 Chr.3: 149740752-149740752 on Build GRCh38
rs978074663 Chr.3: 149740775-149740775 on Build GRCh38
rs1051887214 Chr.3: 149740595-149740595 on Build GRCh38
rs753009060 Chr.3: 149740553-149740554 on Build GRCh38
rs531294471 Chr.3: 149740620-149740620 on Build GRCh38
rs1010022394 Chr.3: 149740686-149740686 on Build GRCh38
rs991240040 Chr.3: 149740428-149740428 on Build GRCh38
rs551930946 Chr.3: 149740418-149740418 on Build GRCh38
rs188187372 Chr.3: 149740499-149740499 on Build GRCh38

View Full Product Details