Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183945761-183946268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC5
Entrez Gene ID
10057
Gene Name
ATP binding cassette subfamily C member 5
Gene Aliases
ABC33, EST277145, MOAT-C, MOATC, MRP5, SMRP, pABC11
UniGene
Hs.368563
Gene Chromosome Location
Chr.3: 183919934-184018010 on Build GRCh38
Transcripts
NM_001320032.1 NM_005688.3 XM_005247058.4 XM_005247059.4 XM_011512314.1 XM_011512315.1 XM_017005492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584135 c.3497A>G p.Y1166C Chr.3: 183945857-183945857 on Build GRCh38
362251 c.3422G>T p.G1141V Chr.3: 183945932-183945932 on Build GRCh38
1041851 c.3437C>T p.T1146M Chr.3: 183945917-183945917 on Build GRCh38
6590444 c.3467G>A p.R1156Q Chr.3: 183945887-183945887 on Build GRCh38
3330314 c.3481G>T p.E1161* Chr.3: 183945873-183945873 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780357789 Chr.3: 183945888-183945888 on Build GRCh38
rs747444612 Chr.3: 183945801-183945801 on Build GRCh38
rs781735401 Chr.3: 183945908-183945908 on Build GRCh38
rs923888629 Chr.3: 183946038-183946038 on Build GRCh38
rs752637397 Chr.3: 183945861-183945861 on Build GRCh38
rs764783298 Chr.3: 183945950-183945950 on Build GRCh38
rs962846072 Chr.3: 183946150-183946150 on Build GRCh38
rs905868529 Chr.3: 183946122-183946122 on Build GRCh38
rs1044507773 Chr.3: 183946104-183946104 on Build GRCh38
rs879665551 Chr.3: 183945998-183945998 on Build GRCh38
rs769428747 Chr.3: 183945819-183945819 on Build GRCh38
rs58764680 Chr.3: 183946040-183946040 on Build GRCh38
rs896340378 Chr.3: 183946074-183946074 on Build GRCh38
rs1006632014 Chr.3: 183946129-183946129 on Build GRCh38
rs28365023 Chr.3: 183945832-183945832 on Build GRCh38
rs368799148 Chr.3: 183945859-183945859 on Build GRCh38
rs950875894 Chr.3: 183945809-183945809 on Build GRCh38
rs974301782 Chr.3: 183945802-183945802 on Build GRCh38
rs1025859744 Chr.3: 183946210-183946210 on Build GRCh38
rs761346073 Chr.3: 183945815-183945815 on Build GRCh38
rs774068512 Chr.3: 183945987-183945987 on Build GRCh38
rs1054277435 Chr.3: 183946005-183946005 on Build GRCh38
rs750568717 Chr.3: 183945883-183945883 on Build GRCh38
rs12493172 Chr.3: 183946117-183946117 on Build GRCh38
rs1044245766 Chr.3: 183945856-183945856 on Build GRCh38
rs187629097 Chr.3: 183946132-183946132 on Build GRCh38
rs951464372 Chr.3: 183946246-183946246 on Build GRCh38
rs371593634 Chr.3: 183945824-183945824 on Build GRCh38
rs866072889 Chr.3: 183946092-183946092 on Build GRCh38
rs927116474 Chr.3: 183945881-183945881 on Build GRCh38
rs531505833 Chr.3: 183946076-183946076 on Build GRCh38
rs28365022 Chr.3: 183945813-183945813 on Build GRCh38
rs551903262 Chr.3: 183945984-183945984 on Build GRCh38
rs72551384 Chr.3: 183945912-183945913 on Build GRCh38
rs28365021 Chr.3: 183945800-183945800 on Build GRCh38
rs1051198451 Chr.3: 183946053-183946053 on Build GRCh38
rs564665731 Chr.3: 183945876-183945876 on Build GRCh38
rs375530575 Chr.3: 183945877-183945877 on Build GRCh38
rs765292844 Chr.3: 183945878-183945878 on Build GRCh38
rs767311011 Chr.3: 183945934-183945934 on Build GRCh38
rs777373401 Chr.3: 183946172-183946173 on Build GRCh38
rs995770896 Chr.3: 183946187-183946187 on Build GRCh38
rs547166914 Chr.3: 183945935-183945935 on Build GRCh38
rs759305243 Chr.3: 183945917-183945917 on Build GRCh38
rs755404743 Chr.3: 183945906-183945906 on Build GRCh38
rs769248670 Chr.3: 183945805-183945805 on Build GRCh38
rs376694775 Chr.3: 183945834-183945834 on Build GRCh38
rs2292998 Chr.3: 183946045-183946045 on Build GRCh38
rs528691810 Chr.3: 183945901-183945901 on Build GRCh38
rs770448224 Chr.3: 183945947-183945947 on Build GRCh38
rs372595266 Chr.3: 183945887-183945887 on Build GRCh38
rs375899840 Chr.3: 183945916-183945916 on Build GRCh38
rs890912690 Chr.3: 183946081-183946081 on Build GRCh38
rs373612396 Chr.3: 183945825-183945825 on Build GRCh38
rs935943928 Chr.3: 183945988-183945988 on Build GRCh38

View Full Product Details