Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186786892-186787401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A2
Entrez Gene ID
1974
Gene Name
eukaryotic translation initiation factor 4A2
Gene Aliases
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
UniGene
Hs.518475
Gene Chromosome Location
Chr.3: 186783572-186789897 on Build GRCh38
Transcripts
NM_001967.3 NR_002586.1

Gene Symbol
SNORA63
Entrez Gene ID
6043
Gene Name
small nucleolar RNA, H/ACA box 63
Gene Aliases
E3, E3-2, RNE3, RNU107, SNORA63A, elF-4AII
UniGene
Hs.518475
Gene Chromosome Location
Chr.3: 186787299-186787433 on Build GRCh38
Transcripts
NM_001967.3 NR_002586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207576 c.909+9T>C p.? Chr.3: 186787273-186787273 on Build GRCh38
6951343 c.794G>A p.C265Y Chr.3: 186787149-186787149 on Build GRCh38
1042370 c.854A>G p.K285R Chr.3: 186787209-186787209 on Build GRCh38
7177170 c.868A>G p.T290A Chr.3: 186787223-186787223 on Build GRCh38
6951344 c.800T>A p.L267* Chr.3: 186787155-186787155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562687342 Chr.3: 186786954-186786954 on Build GRCh38
rs375043979 Chr.3: 186787316-186787316 on Build GRCh38
rs904856305 Chr.3: 186787034-186787034 on Build GRCh38
rs763689805 Chr.3: 186787328-186787328 on Build GRCh38
rs149330304 Chr.3: 186787201-186787201 on Build GRCh38
rs774745060 Chr.3: 186787345-186787345 on Build GRCh38
rs777503818 Chr.3: 186787094-186787094 on Build GRCh38
rs141019170 Chr.3: 186787095-186787095 on Build GRCh38
rs533235495 Chr.3: 186786996-186786996 on Build GRCh38
rs895842895 Chr.3: 186786931-186786931 on Build GRCh38
rs186916940 Chr.3: 186787106-186787106 on Build GRCh38
rs749341711 Chr.3: 186787343-186787343 on Build GRCh38
rs761447929 Chr.3: 186787098-186787099 on Build GRCh38
rs373445043 Chr.3: 186787281-186787281 on Build GRCh38
rs754199367 Chr.3: 186787227-186787227 on Build GRCh38
rs780672201 Chr.3: 186787342-186787344 on Build GRCh38
rs755521285 Chr.3: 186787087-186787087 on Build GRCh38
rs764510573 Chr.3: 186787367-186787367 on Build GRCh38
rs551124747 Chr.3: 186787013-186787013 on Build GRCh38
rs1003188684 Chr.3: 186787055-186787055 on Build GRCh38
rs79277600 Chr.3: 186787185-186787185 on Build GRCh38
rs746893414 Chr.3: 186787243-186787243 on Build GRCh38
rs761978868 Chr.3: 186787108-186787108 on Build GRCh38
rs111802039 Chr.3: 186787112-186787112 on Build GRCh38
rs1018822079 Chr.3: 186786983-186786983 on Build GRCh38
rs556787465 Chr.3: 186787090-186787090 on Build GRCh38
rs140038024 Chr.3: 186787183-186787184 on Build GRCh38
rs759800638 Chr.3: 186787349-186787349 on Build GRCh38
rs201168774 Chr.3: 186787207-186787207 on Build GRCh38
rs900001553 Chr.3: 186786989-186786989 on Build GRCh38
rs1000369195 Chr.3: 186786981-186786981 on Build GRCh38
rs766456656 Chr.3: 186787268-186787269 on Build GRCh38
rs773527578 Chr.3: 186787111-186787111 on Build GRCh38
rs750299552 Chr.3: 186787104-186787108 on Build GRCh38
rs756687113 Chr.3: 186787293-186787293 on Build GRCh38
rs904750000 Chr.3: 186786949-186786949 on Build GRCh38
rs1045305141 Chr.3: 186786995-186786995 on Build GRCh38
rs553982170 Chr.3: 186787025-186787025 on Build GRCh38
rs765195435 Chr.3: 186787109-186787109 on Build GRCh38
rs773154716 Chr.3: 186787261-186787261 on Build GRCh38
rs540138705 Chr.3: 186787159-186787159 on Build GRCh38
rs202021088 Chr.3: 186787100-186787100 on Build GRCh38
rs34583258 Chr.3: 186787174-186787174 on Build GRCh38
rs377327346 Chr.3: 186787344-186787344 on Build GRCh38
rs75837460 Chr.3: 186787124-186787124 on Build GRCh38
rs775701790 Chr.3: 186787362-186787362 on Build GRCh38
rs772078879 Chr.3: 186786940-186786940 on Build GRCh38
rs375685562 Chr.3: 186787220-186787220 on Build GRCh38
rs777115501 Chr.3: 186787177-186787177 on Build GRCh38
rs377417923 Chr.3: 186787283-186787283 on Build GRCh38
rs759910219 Chr.3: 186787297-186787297 on Build GRCh38
rs745861242 Chr.3: 186787105-186787105 on Build GRCh38
rs777134828 Chr.3: 186787372-186787372 on Build GRCh38
rs998297264 Chr.3: 186787352-186787352 on Build GRCh38
rs778546320 Chr.3: 186787296-186787296 on Build GRCh38
rs540193992 Chr.3: 186787279-186787279 on Build GRCh38
rs983051783 Chr.3: 186787082-186787082 on Build GRCh38
rs995920329 Chr.3: 186787005-186787005 on Build GRCh38
rs760002459 Chr.3: 186787272-186787272 on Build GRCh38
rs1980299 Chr.3: 186787198-186787198 on Build GRCh38
rs771284823 Chr.3: 186787269-186787269 on Build GRCh38
rs369963925 Chr.3: 186787271-186787271 on Build GRCh38
rs774108248 Chr.3: 186787101-186787101 on Build GRCh38
rs769968288 Chr.3: 186787252-186787252 on Build GRCh38
rs764700631 Chr.3: 186787373-186787373 on Build GRCh38
rs751007895 Chr.3: 186787279-186787280 on Build GRCh38
rs766910389 Chr.3: 186787113-186787113 on Build GRCh38
rs763304093 Chr.3: 186787264-186787264 on Build GRCh38
rs954408803 Chr.3: 186787077-186787077 on Build GRCh38
rs747666670 Chr.3: 186787083-186787083 on Build GRCh38
rs754535553 Chr.3: 186787321-186787322 on Build GRCh38
rs370580137 Chr.3: 186787351-186787351 on Build GRCh38
rs1010226956 Chr.3: 186786932-186786932 on Build GRCh38
rs35344552 Chr.3: 186787187-186787187 on Build GRCh38
rs370862991 Chr.3: 186787315-186787315 on Build GRCh38
rs373250339 Chr.3: 186787319-186787319 on Build GRCh38
rs192115833 Chr.3: 186787119-186787119 on Build GRCh38
rs375328761 Chr.3: 186787116-186787116 on Build GRCh38
rs1030020973 Chr.3: 186787044-186787044 on Build GRCh38
rs754120978 Chr.3: 186787099-186787100 on Build GRCh38
rs572904172 Chr.3: 186787240-186787240 on Build GRCh38
rs781118391 Chr.3: 186787342-186787342 on Build GRCh38
rs34750546 Chr.3: 186787155-186787155 on Build GRCh38
rs755679855 Chr.3: 186787174-186787174 on Build GRCh38
rs761308394 Chr.3: 186787363-186787363 on Build GRCh38
rs1045274054 Chr.3: 186787259-186787259 on Build GRCh38
rs112227828 Chr.3: 186786968-186786969 on Build GRCh38
rs753287035 Chr.3: 186787273-186787273 on Build GRCh38
rs554088706 Chr.3: 186787001-186787002 on Build GRCh38
rs566490580 Chr.3: 186787045-186787045 on Build GRCh38
rs747027943 Chr.3: 186787323-186787323 on Build GRCh38
rs992940450 Chr.3: 186787122-186787122 on Build GRCh38
rs951197056 Chr.3: 186787073-186787074 on Build GRCh38
rs1011088217 Chr.3: 186786968-186786968 on Build GRCh38
rs758402717 Chr.3: 186787105-186787106 on Build GRCh38
rs946917118 Chr.3: 186787255-186787255 on Build GRCh38
rs373252426 Chr.3: 186787299-186787299 on Build GRCh38
rs1035182637 Chr.3: 186787061-186787061 on Build GRCh38
rs963586792 Chr.3: 186787110-186787110 on Build GRCh38
rs987521596 Chr.3: 186787195-186787195 on Build GRCh38
rs765135371 Chr.3: 186787101-186787106 on Build GRCh38

View Full Product Details