Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7695523-7696038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1438696 c.923C>T p.P308L Chr.5: 7695805-7695805 on Build GRCh38
3776858 c.954C>G p.L318L Chr.5: 7695836-7695836 on Build GRCh38
278664 c.888C>T p.I296I Chr.5: 7695770-7695770 on Build GRCh38
6307897 c.970C>T p.Q324* Chr.5: 7695852-7695852 on Build GRCh38
332087 c.937C>A p.H313N Chr.5: 7695819-7695819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562797165 Chr.5: 7695627-7695627 on Build GRCh38
rs1025425951 Chr.5: 7695999-7695999 on Build GRCh38
rs778489668 Chr.5: 7695716-7695716 on Build GRCh38
rs1014488292 Chr.5: 7695553-7695553 on Build GRCh38
rs771546545 Chr.5: 7695667-7695667 on Build GRCh38
rs749839415 Chr.5: 7695659-7695659 on Build GRCh38
rs1002972736 Chr.5: 7695646-7695646 on Build GRCh38
rs929518793 Chr.5: 7695558-7695558 on Build GRCh38
rs202062473 Chr.5: 7695896-7695896 on Build GRCh38
rs953879314 Chr.5: 7695707-7695707 on Build GRCh38
rs760696204 Chr.5: 7695865-7695865 on Build GRCh38
rs775371186 Chr.5: 7695863-7695863 on Build GRCh38
rs758019160 Chr.5: 7695891-7695891 on Build GRCh38
rs764774720 Chr.5: 7695887-7695887 on Build GRCh38
rs535824046 Chr.5: 7695588-7695588 on Build GRCh38
rs764758148 Chr.5: 7695948-7695948 on Build GRCh38
rs11134252 Chr.5: 7695915-7695915 on Build GRCh38
rs781423182 Chr.5: 7695911-7695911 on Build GRCh38
rs370623770 Chr.5: 7695726-7695726 on Build GRCh38
rs368207400 Chr.5: 7695734-7695734 on Build GRCh38
rs948899363 Chr.5: 7695598-7695598 on Build GRCh38
rs558775105 Chr.5: 7696003-7696003 on Build GRCh38
rs538939893 Chr.5: 7695947-7695947 on Build GRCh38
rs889667593 Chr.5: 7695934-7695934 on Build GRCh38
rs938747450 Chr.5: 7695882-7695882 on Build GRCh38
rs370821588 Chr.5: 7695762-7695762 on Build GRCh38
rs756877878 Chr.5: 7695883-7695883 on Build GRCh38
rs894264127 Chr.5: 7695925-7695925 on Build GRCh38
rs866223493 Chr.5: 7695927-7695927 on Build GRCh38
rs111286347 Chr.5: 7695618-7695618 on Build GRCh38
rs750075297 Chr.5: 7695889-7695889 on Build GRCh38
rs934012110 Chr.5: 7695597-7695597 on Build GRCh38
rs940645931 Chr.5: 7695873-7695873 on Build GRCh38
rs972403101 Chr.5: 7695906-7695906 on Build GRCh38
rs73047981 Chr.5: 7695556-7695556 on Build GRCh38
rs1019842187 Chr.5: 7695974-7695974 on Build GRCh38
rs986799555 Chr.5: 7695842-7695842 on Build GRCh38
rs549346695 Chr.5: 7695916-7695916 on Build GRCh38
rs957501804 Chr.5: 7695589-7695589 on Build GRCh38
rs189185684 Chr.5: 7695938-7695938 on Build GRCh38
rs1021533197 Chr.5: 7695682-7695682 on Build GRCh38
rs192786032 Chr.5: 7695806-7695806 on Build GRCh38
rs200439471 Chr.5: 7695770-7695770 on Build GRCh38
rs999412230 Chr.5: 7695724-7695724 on Build GRCh38
rs371883348 Chr.5: 7695625-7695625 on Build GRCh38
rs977651428 Chr.5: 7695573-7695573 on Build GRCh38
rs773282692 Chr.5: 7695742-7695742 on Build GRCh38
rs1013936378 Chr.5: 7695685-7695685 on Build GRCh38
rs756826596 Chr.5: 7695712-7695712 on Build GRCh38
rs753380336 Chr.5: 7695801-7695801 on Build GRCh38
rs114403062 Chr.5: 7695568-7695568 on Build GRCh38
rs1019450766 Chr.5: 7695709-7695709 on Build GRCh38
rs777442996 Chr.5: 7695711-7695711 on Build GRCh38
rs144532126 Chr.5: 7695595-7695595 on Build GRCh38
rs374469258 Chr.5: 7695809-7695809 on Build GRCh38
rs184161358 Chr.5: 7695868-7695868 on Build GRCh38
rs1045447411 Chr.5: 7695963-7695963 on Build GRCh38
rs866113571 Chr.5: 7695803-7695803 on Build GRCh38
rs909502493 Chr.5: 7695819-7695819 on Build GRCh38
rs994307311 Chr.5: 7695987-7695987 on Build GRCh38
rs749434410 Chr.5: 7695749-7695749 on Build GRCh38
rs753816462 Chr.5: 7695874-7695874 on Build GRCh38
rs113227937 Chr.5: 7695708-7695708 on Build GRCh38
rs965261492 Chr.5: 7695984-7695984 on Build GRCh38
rs1024554522 Chr.5: 7695695-7695695 on Build GRCh38
rs768943451 Chr.5: 7695729-7695729 on Build GRCh38
rs556436108 Chr.5: 7695730-7695730 on Build GRCh38
rs1055518564 Chr.5: 7695921-7695921 on Build GRCh38
rs11134251 Chr.5: 7695732-7695732 on Build GRCh38
rs911820377 Chr.5: 7695623-7695623 on Build GRCh38
rs749912077 Chr.5: 7695979-7695979 on Build GRCh38
rs35317146 Chr.5: 7695588-7695589 on Build GRCh38
rs898895715 Chr.5: 7695652-7695652 on Build GRCh38
rs755295293 Chr.5: 7695596-7695596 on Build GRCh38
rs752612307 Chr.5: 7695705-7695705 on Build GRCh38
rs767242746 Chr.5: 7695833-7695833 on Build GRCh38
rs1044930515 Chr.5: 7695840-7695840 on Build GRCh38
rs147839766 Chr.5: 7695651-7695651 on Build GRCh38

View Full Product Details